Search
Filtration is an integral piece of nearly every workflow of every lab. whether its cell culture, protein research, water testing, bioprocess design, or QC/QC testing of beverages – filtration will play a role in your lab’s daily routine. And Thermo Fisher Scientific has the tools you need.
![]() |
1. Printed with lot numbers for traceability 2. Easy to read graduations 3. Membranes up to 90mm for maximum flowrate and throughput 4. Color-coded by membrane 5. Featuring Nalgene's famous leak-proof seal 6. Ergonomic handgrips
|
Laboratory filtration of culture media does not affect cell growth when the correct filter device and membrane are used.
Yes, you can increase efficiency in the time it takes to filter, enhance your speed with faster flow rates, and improve the stability of your media to save time in the lab by choosing the right filtration device.
The lower the extractables, the less chance of those compounds leaching into your filtered sample. Thermo Scientific Nalgene Rapid-Flow Receiver Bottles have lower extractables present compared to all other equivalent filtration devices. We source only virgin resins from high-quality suppliers to ensure consistency and quality. We also optimize our products and processes to avoid the use of various additives and slip agents whenever possible.
Image depicting the results from the Rapid-Flow filter unit receiver bottle compared to other equivalent receiver bottles. Results include the total organic carbon (TOC), absorbance, and metals analysis.
In a study designed to look at the flow rates of the Thermo Scientific Nalgene Rapid-Flow Filter units, the column-based membrane support design had up to 38% faster flow rates than competitors using a radial spoke membrane support design.
Nalgene Rapid-Flow Sterile Single Use Filter Units have a column-based membrane support plate. |
![]() | ![]() |
The radial spoke design used by other suppliers can result in suboptimal flow rates. | |
| Consistently consistent All Nalgene filters now have the Rapid-Flow multi-column membrane-support system. This proprietary system provides a uniform, consistent separation between touchpoints with the membrane, minimizing gap stress to maintain optimal flow. |
← Membrane |
Mind the gaps Other filters use a radial spoke support system. The gaps between spokes lack uniformity and consistency in membrane support, leading to increased stress and distortion. The result? Suboptimal flow rate and throughput. |
|
Rapid-Flow Sterile Single Use Filter Units can ave up to 38% faster flow rate than units from other suppliers.
Need technical support? Contact our expert team for technical and application support of Laboratory Products
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.