GLPKit-2new-230
You can handle anything
The leader in pipetting technology—From the first variable volume micropipette to the first to offer an interlock Thermo Scientific™ ClipTip™ interface—Thermo Scientific manual pipetting systems deliver unparalleled ergonomics, dependability and quality for consistent and reproducible results.
New to the lab or just looking to improve on your pipetting technique? We can help!
View our good laboratory pipetting guide›
Select the single channel pipette that's right for you
F1-ClipTip Pipettes
Sealed in security. Feel the difference.
  • Tips lock in place with a light touch
  • A complete seal on every channel
  • Maintain precise sample volumes
F1-ClipTipSingleChannel
Finnpipette F1 Pipettes
Ergonomic and Safe Performance
  • Remarkably lightweight
  • Reduce strain with low plunger and ejection forces
  • Ergonomic comfort
Finnpipette-F1-SingleChannel
Finnpipette F2 Pipettes
Robust, Durable Design
  • Fully autoclavable
  • Withstands exposure to harsh chemical applications
  • Resistant to UV light
Finnpipette-F2-Manual-Full
Did you know we have Electronic Single Channel pipettes?
E1CT-index230
Ideal for repetitive tasks, choose from our Electronic pipette models with index finger pipetting action and electronic tip ejection. A great choice when pipetting with microcentrifuge or test tubes.
Don't forget the pipette tips!
ClipTipReloads
Increase reliability and reproducibility with tips precisely manufactured and designed to work together as a system with your pipette. Thermo Scientific ClipTip and Finntip pipette tips are available in a variety of volumes and packaging configurations.
View all Handheld Pipette Tips
What is ClipTip Technology?
Friction sealing systems
Pipettes with a friction seal system, rely on a user’s force to attach tips to the pipette, which varies from user to user.
friction-cliptip-system-sm
Increased tip attachment and ejection forces
Loose tips which can drop off or leak air
Decreased confidence in reproducibility of results
Premature tip cone wear
ClipTip system
ClipTip interlocking technology has an interface between the pipette fitting and tips that seal by clipping into place.
interlocking-cliptip-system-new-sm
  • Extremely low tip attachment and ejection forces
  • Complete seal on every channel
  • No loose tips
  • Reproducibility 
Worry-free pipetting is finally here with the only tip that clips
The F1-ClipTip manual pipettes ensure you have a complete seal on each channel when the tips are locked into place. Whether you’re using a single or multichannel pipette, you will get the volume you expect to accelerate discovery.
Learn more
Related products
ClipTip Pipetting Systems
Breakthrough Thermo Scientific™ ClipTip™ technology makes daily pipetting a totally new experience.  
Finnpipette Systems
Delivering years of unmatched productivity and ergonomics across thousands of labs and applications.
Electronic Pipettes
The proven performance and productivity of index-finger pipetting action, electronic tip ejection, and Thermo Scientific ClipTip interlock technology.
Pipette Tips
Designed, developed and manufactured to perfectly fit Thermo Scientific pipettes. A pipette’s performance is directly related to the quality of the pipette tip for optimal liquid handling results.