Chip
Sample set
AGCC ARR/CEL
AGCC ARR/CEL/CHP
Human Gene 1.0 ST Array
Tissues
Download (131 MB)
Download (134 MB)
Human Gene 1.0 ST Array
Tissue mixture
Download (134 MB)
Download (136 MB)
Mouse Gene 1.0 ST Array
Tissues
Download (144 MB)
Download (147 MB)
Rat Gene 1.0 ST Array
Tissues
Download (144 MB)
Download (147 MB)
Description of downloads
  • AGCC ARR/CEL: These are zipped files converted with DEC from the GCOS Flat Files. These zip files contain richly annotated sample files (*.ARR) and AGCC-formatted probe cell intensity data (*.CEL). These data can be parsed with the Fusion SDK and are easily imported into GeneChip Command Console or Expression Console.
  • ARR/CEL/CHP: These are zipped study files from Expression Console v1.1. These zip files contain richly annotated sample files (*.ARR), AGCC formatted probe cell intensity data (*.CEL), and AGCC-formatted probe level summarization data (*.CHP). CHP files were generated using RMA and for exon arrays using only the core probes. These data can be parsed with the Fusion SDK and are easily imported into GeneChip Command Console Software or Expression Console.
Description of sample sets
This data set contains a collection of 11 tissues. The RNA samples are from a commercial source. Each tissue has three biological replicates; there are no technical replicates. The 11 tissues are listed below:
Tissues
Brain
Liver
Testis
Breast (human)
Lung (mouse, rat)
Thymus (mouse, rat)
Embryo (mouse, rat)
Pancreas (human)
Thyroid (human)
Heart
Prostate
Kidney
Skeletal Muscle
Mixtures
Mix 1
Mix 2
Mix 3
Mix 4
Mix 5
Mix 6
Mix 7
Mix 8
Mix 9
Brain
0.00
0.05
0.10
0.25
0.50
0.75
0.90
0.95
1.00
Heart
1.00
0.95
0.90
0.75
0.50
0.25
0.10
0.05
0.00