This data set is expected to be useful for the development and evaluation of low-level analysis methods for making genotype calls from probe intensity data. It consists of 30 trios each analyzed on both the Xba and Hind array (so a total of 30x3x2=180 hybridizations). The trios are CEPH trios also used in the International HapMap Project .
Of particular use is the fact that the HapMap Project has made available a large number of reference genotypes which can be used in conjunction with this data set. The HapMap data access policy  limits redistribution rights on these genotypes so they cannot be made available directly by Thermo Fisher Scientific, but the reference data can be downloaded directly from the HapMap Project. As of HapMap release 16c1, a total of 30,000 SNPs have reference genotypes available for the samples shared here. These numbers are steadily increasing with each HapMap update.
The details of the analysis method used by GDAS to determine genotype calls based on probe intensity data have been published in Bioinformatics.
The data set has been split into 11 parts for convenient download. These can be unzipped on top of one another. The file with the word 'base' in the file name is required, the other 10 zip files each contain distinct collections of chip data and users wanting to download only a subset of the data may pick a subset of these zips.
The data is provided in two versions. Each version contains the same data but in different file formats. Version 1 (in table 1) contains raw CEL, CHP and EXP files and is suitable for use outside of the GCOS/GTYPE framework. It is expected to be mainly of interest for users interested in low-level probe analysis. Version 2 (in table 2) contains DTT format files for integration with the GCOS/GTYPE framework and is expected to be mainly of interest for users wishing to integrate the data with these applications.
In either case there is a file named README.txt provided in the 'base' file with detailed instructions on how to use the data. Md5 checksums are provided in the tables below for verification of the integrity of downloaded data.
Version 1 release of data: CEL, CHP and EXP format
(Suitable for use outside of GCOS/GTYPE framework)
File
Size
md5 checksum
Description
100K_trios-base.zip
37 MB
f70cb636002b4ac8bc807f0639dae691
Documentation and library files for entire data set
100K_trios-hind-1.zip
229 MB
6d593c94260804ada939952ac4df2503
Probe intensities and genotype calls for 18 samples on Hind Array
100K_trios-hind-2.zip
228 MB
84691de89bff1aa38704f41682c85940
Probe intensities and genotype calls for 18 samples on Hind Array
100K_trios-hind-3.zip
230 MB
fb1efb1ed2f713c7b077b57146880903
Probe intensities and genotype calls for 18 samples on Hind Array
100K_trios-hind-4.zip
228 MB
e0aeaada50ec4dc23ed75b2154f3de75
Probe intensities and genotype calls for 18 samples on Hind Array
100K_trios-hind-5.zip
230 MB
a29dcc1780ba66ebdd8342faa01c46fa
Probe intensities and genotype calls for 18 samples on Hind Array
100K_trios-xba-1.zip
245 MB
941c0e36785440b85b2e9127ae91c26d
Probe intensities and genotype calls for 18 samples on Xba Array
100K_trios-xba-2.zip
232 MB
4ff4aecb882b9e30c6fbebff84080489
Probe intensities and genotype calls for 18 samples on Xba Array
100K_trios-xba-3.zip
228 MB
71dc53123fd26d5da441d48c2dd21e6c
Probe intensities and genotype calls for 18 samples on Xba Array
100K_trios-xba-4.zip
228 MB
34b85d8188276f529a2ad3e273c8dce9
Probe intensities and genotype calls for 18 samples on Xba Array
100K_trios-xba-5.zip
230 MB
4f48983e00f4940ab77cf4c6ec46a75e
Probe intensities and genotype calls for 18 samples on Xba Array
Version 2 release of data: DTT format
(Intended for use within GCOS/GTYPE framework)
File
Size
md5 checksum
Description
100K_trios-base.zip
38 MB
f70cb636002b4ac8bc807f0639dae691
Documentation and library files for entire data set
100K_trios-hind-dtt-1.zip
229 MB
8a28150d937fa81f29431b71da36706b
Archived DTT files for 18 samples on Hind Array
100K_trios-hind-dtt-2.zip
215 MB
705be96f555a85bdc38d5a24c7c8b15b
Archived DTT files for 18 samples on Hind Array
100K_trios-hind-dtt-3.zip
203 MB
19ddd1744b6f12b3b20c2401295858b2
Archived DTT files for 18 samples on Hind Array
100K_trios-hind-dtt-4.zip
228 MB
d9ba75c0ae6de144738ffa683e9e9f34
Archived DTT files for 18 samples on Hind Array
100K_trios-hind-dtt-5.zip
230 MB
746aeec3022a632c516883d3f021e33d
Archived DTT files for 18 samples on Hind Array
100K_trios-xba-dtt-1.zip
245 MB
a0f8b6af3a21243f816e6bad2a5e94a9
Archived DTT files for 18 samples on Xba Array
100K_trios-xba-dtt-2.zip
232 MB
51fd6cffda8856d21718372dd04cf137
Archived DTT files for 18 samples on Xba Array
100K_trios-xba-dtt-3.zip
228 MB
cf717614f6e708b81ab1e8f01577aa9d
Archived DTT files for 18 samples on Xba Array
100K_trios-xba-dtt-4.zip
228 MB
e04f50ec919854b86b9c87fe0cb037d6
Archived DTT files for 18 samples on Xba Array
100K_trios-xba-dtt-5.zip
230 MB
ad1dfa43fee7766625930e01334fc229
Archived DTT files for 18 samples on Xba Array