quantstudio-3d-showcase
The Applied Biosystems QuantStudio 3D Digital PCR System is a simple, chip-based digital PCR platform used in a number of applications where absolute quantification is critical. While the system is capable of detecting both TaqMan and SYBR Green chemistries, gold-standard TaqMan Assays are ideal for the QuantStudio 3D system as they minimize false positives in quantitative PCR experiments and eliminate the need for melt curve analysis.
Featured dPCR application: Mutation detection using liquid biopsy analysis
TaqMan Liquid Biopsy dPCR assays       
Absolute quantification on the QuantStudio 3D dPCR
system using sealed-chip technology for a reliable
method and precise, sensitive data. 
data-analysis-brochure
Liquid Biopsy Analysis
Accelerating the future of liquid biopsies with digital PCR and next-generation sequencing
Find the perfect predesigned TaqMan Assay for your QuantStudio 3D dPCR application
High-precision copy number variation
Detect and quantify small percent copy number differences with a high degree of precision
Next-generation sequencing library quantification
Absolute quantification of NGS libraries and validation of sequencing results, without reference standards
Rare mutation analysis
Detect and quantify rare mutations for low-prevalence targets in cancer research samples
Differential gene expression
Detect gene expressions changes of ±10% for absolute transcript quantification without a reference gene
Viral load quantification
Absolute quantification of bacterial and viral loads, and absolute pathogen counts
Low-level pathogen detection
Detect low-level pathogens that cause human illnesses through contaminated food and water supplies
Reference and standard quantification
Generate absolute reference standards for genetic measurements, metrology, and cross-lab comparisons
GMO detection
Enables sensitive detection and absolute quantification of plant mutations and genetically modified organisms
QuantStudio 3D Digital PCR System
QuantStudio-3D-crop
Absolute data with high precision and sensitivity—detect and count individual molecules
Simple workflow—just load and go for digital data from chip-based system
Affordable platform—start doing digital PCR immediately, and at an affordable cost
Compatible—use your existing TaqMan real-time PCR assays
Sealed system—helps limit contaminants using a sealed chip without exposed sample-transfer steps
Wide dynamic range—achieve linearity over 5 logs
Intuitive—view data directly on touch screen and analyze efficiently using Applied Biosystems AnalysisSuite Digital PCR Software 
Learn more