Invitrogen Platinum technology is used with several different types of DNA polymerases. Platinum technology consists of proprietary antibodies, specific to each DNA polymerase, that inhibit polymerase activity at ambient temperatures.
Invitrogen Platinum DNA Polymerases are provided in an inactive form, due to specific binding of the antibodies. The polymerases are activated in a temperature-dependent manner (at 94°C) during the start of PCR. Once dissociated from the antibodies, the DNA polymerases regain full activity.
Which Platinum DNA Polymerase is right for you?
  PlatinumTaq DNA Polymerase Platinum SuperFi DNA Polymerase Platinum Taq DNA Polymerase High Fidelity Platinum Pfx DNA Polymerase Platinum GenoType Tsp DNA Polymerase Platinum Multiplex PCR Master Mix Platinum Tfi DNA Polymerase Platinum Tfi Exo(-) DNA Polymerase
Fidelity (compared to Taq) 1x >100x 6x 26x 1x 1x 1x 1x
Amplicon size 5 kb or less* 20 kb or more** 20 kb or less 20 kb or less 2.5 kb or less 1 kb or less 1 kb or less 1 kb or less
Resulting Amplicon ends 3′-A Blunt Mixed Blunt Blunt 3′-A 3′-A 3′-A
Exonuclease activity 5′→ 3′ 3′→ 5′ 5′→ 3′
3′→ 5′
3′→ 5′ None 5′→ 3′ 5′→ 3′ None
Supermix or master mix available        
Direct gel loading available            
Applications
Routine PCR          
High fidelity          
High specificity
High yield      
Long amplicon          
GC-rich targets            
PCR genotyping            
Multiplex              
* Optional KB Extender provides additional versatility and PCR amplification up to 10 kb.
** Amplification of >20 kb fragment sizes is possible (up to 40 kb), but may require additional optimization of reaction conditions and primer design.
Ordering information
Resources
PCR education
Learn about the history of PCR, setup considerations, importance of DNA polymerase choice, common methods and applications, and troubleshooting.
PCR and cDNA Synthesis Support Center
Find tips, troubleshooting help, and resources for your end-point PCR and cDNA applications.