The Applied Biosystems MicroAmp TriFlex 3 x 32-Well PCR Reaction Plate and Applied Biosystems MicroAmp 32-Well Clear Adhesive Film help to lower the cost of consumables and reduces waste. They are designed specifically for the Applied Biosystems ProFlex 3 x 32-Well PCR System and are compatible with all other Applied Biosystems 0.2 mL, 96-well thermal cyclers.
Stop spending precious time forcing your plastics to fit your reaction size. Just tear off what you need and get on with your work.
- One plate fits all—designed for use with the ProFlex 3 x 32-well block and all standard Applied Biosystems 96-well block thermal cyclers
- Easy-to-tear dual side tabs—simplifies plate segmentation
- Alpha-numeric labeling on each plate segment—permits easy well identification
- 32-well adhesive film—reduces sample evaporation and well-to-well contamination
MicroAmp TriFlex 3 x 32-Well PCR Reaction Plate
All Applied Biosystems thermal cyclers and PCR plastics are tested to help ensure optimal PCR performance, and are “engineer approved”—designed and built for reliability and precision to enable optimal PCR results.
MicroAmp TriFlex 3 x 32-Well Reaction Plates are:
Designed for use with the ProFlex
3 x 32-Well PCR System
Compatible with all standard Applied Biosystems 0.2-mL, 96-well block thermal cyclers
MicroAmp 32-Well Clear Adhesive Film
MicroAmp 32-Well Clear Adhesive Film for evaporation-free plate sealing
MicroAmp TriFlex 3 x 32-Well PCR Reaction Plate
All Applied Biosystems thermal cyclers and PCR plastics are tested to help ensure optimal PCR performance, and are “engineer approved”—designed and built for reliability and precision to enable optimal PCR results.
MicroAmp TriFlex 3 x 32-Well Reaction Plates are:
Designed for use with the ProFlex
3 x 32-Well PCR System
Compatible with all standard Applied Biosystems 0.2-mL, 96-well block thermal cyclers
MicroAmp 32-Well Clear Adhesive Film
MicroAmp 32-Well Clear Adhesive Film for evaporation-free plate sealing
Available exclusively for Applied Biosystems thermal cyclers to provide optimal PCR results
Get the most from your low- and standard-throughput PCR instruments by choosing the MicroAmp TriFlex 3 x 32-Well PCR Reaction Plate for 96 or 32 wells—one plate fits all!
For Research Use Only. Not for use in diagnostic procedures.