I am having problems with...
Abnormal Amplification
Your curves are sigmoidal or amplification occurs later than you expected.
NTC Positive Amplification
You have an amplification product in your no template control (NTC).
No Amplification
You have no detectable PCR product.
Poor PCR Efficiency
If the slope of your standard curve is below -3.6, you have poor PCR efficiency.
Normal Amplification Curve Example
Your amplification curve should look something like this