QuantStudio7Pro-XLshowcase
Smart connected and smarter productivity at your fingertips
The Applied Biosystems QuantStudio 6 and 7 Pro real-time PCR systems are designed to offer high quality and excellent reliability with features to deliver a smart workflow experience that brings lab-changing qPCR innovations to your research. These systems feature an interactive 12.1-in touchscreen interface, intuitive software, pre-optimized protocol templates, and access to your data anytime, anywhere with our Connect cloud-based platform. Additional smart features are included to enable a personalized experience that improves efficiency and helps streamline your workflow.
Request quote     Flyer     Interactive brochure
Featured video (see more)
QuantStudio 6 and 7 Pro System Features
Take the next step forward in genetic analysis where every component is designed to improve productivity by enhancing the user experience.
Researcher quote (see more)
"It’s SO small. Impressed by the very user friendly touchscreen so we do not need a computer next to it… having it integrated into the cloud is quite convenient."
—Michael Kublista
TATAA Biocenter AB, Sweden
  • Smart features—hands-free functionality includes facial authentication login, voice commands, and RFID reader
  • Manual error reduction—compatible with TaqMan Array Plates with RFID
  • Interchangeable blocks—flexibility to change block formats between 96- (0.1 and 0.2 mL) and 384-well, and TaqMan Array Cards* via simply pushing a button or using a voice command
  • Connect—access, analyze, and share data anytime, anywhere with Connect, our cloud-based platform for real-time information access, secure data storage, and online results sharing
  • Remote access—monitor experiments and access results remotely using the Instrument Connect app
  • Small footprint—minimal bench space required: only 54.7 x 33.8 x 52.5 cm (H x W x D) and 38 kg (wt)
  • Multiplexing—5 or 6 optical channels (21 filter combinations for QuantStudio 7 Pro system)
  • Automation compatible—increase walkaway time and productivity by adding automatic loading of one or two QuantStudio 7 Pro systems with the Orbitor RS2 Microplate Mover
  • Results you can trust—10 logarithmic units of dynamic range to detect differences as small as 1.5-fold
  • Easy access to help—embedded video help from the instrument
  • Peace of mind—superior services and support with Smart Help and Smart Remote Support
QuantStudio Pro systems are compatible with Apple Mac computers.
Download specification sheet ›
Download green features flyer ›
As a qPCR pro, working smarter is your mantra. The new QuantStudio Pro systems introduce a way to incorporate smarter workflows throughout your day whether you are working at your desk, at the bench, or checking in from a remote location.
See a day in the life of a qPCR pro below and find out how your workflow can improve.
Quantstudio-graphic1
At your desk
Working at your desk has never been easier. You can:
  • Plan your experiments, design your protocols, and send them directly to your instrument of choice
  • Collaborate with people on-site or in remote locations
  • Retrieve and analyze your data at your desk; no need to go to the instrument to download or to have a dedicated computer
  • Check to see if the instrument is available and reserve future run times for optimal efficiency
At-the-bench
At the bench
Improve your day at the bench with simple, time-saving, and error-reducing features
  • Get started without removing your gloves
  • Use voice commands for common tasks such as “Start run” and “Open/close door”
  • Reduce data entry and streamline your workflow when using the TaqMan Array Plate with RFID tag
  • Change the block using the on-screen walkthrough
  • Get high-quality qPCR data seamlessly delivered to your preferred destination
Quantstudio-graphic2
Remote location
Taking a coffee break or checking the status of your experiment after work hours is easy with:
  • Remote monitoring that shows run status and amplification plots in real-time
  • Data sharing and analysis features using the cloud-enabled Connect platform
  • Planning and scheduling your next experiment without delay for ultimate productivity
Be the ultimate qPCR pro in your lab or anywhere you may be
QuantStudio 6 Pro system
QuantStudio 7 Pro system
Flexible format Interchangeable blocks with enhanced motorized change mechanism
VeriFlex Technology
3 zones
6 zones
TAC* block
upgradeable
Efficiency & productivity Small footprint
Automation for 24/7 auto plate-loading
upgradeable
Large, full-function touchscreen
Connect  platform-compatible
Features to assist with 21 CFR part 11 compliance**
upgradeable
included
Maximize uptime Smart Help
Help videos
Minimal calibration for easy maintenance
* TaqMan array microfluidic card (TAC) block available in 2020
** Module available in 2020
You can access, analyze, and share data anytime, anywhere* with the Connect cloud-based platform.
  • Remotely monitor your runs in real time
  • Analyze sophisticated data sets in minutes
  • Monitor instrument status and schedule a run from anywhere
  • Securely store data
  • Share results online with colleagues across institutions and around the world
Instrument Connect remote monitoring app
Stay connected to cloud-enabled instruments and benchtop devices anytime, anywhere, and from any mobile device to see availability, run progress, and more.
  • Check the availability of your Connect platform-enabled, network-connected device
  • Monitor run progress of a basic PCR run
  • View amplification plots in real time
  • View plots and filter by sample or target in real time
Learn more
*Where the Web is accessible
If your facility does not allow you to use the Connect cloud-based platform, don’t worry. The QuantStudio 6 and 7 Pro Real-Time PCR Systems will still improve your workflow.
You can still maximize uptime with Smart Help and Smart Remote Support features to streamline technical support help.  TaqMan Array Plates with RFID will also work to help reduce manual errors in loading your assay plate settings and loading the plate setup. And finally, the large touchscreen, ability to use the instrument without an attached computer and data delivery to your preferred location will be fully enabled to assist you.
QuantStudio 6 and 7 Pro connectivity and the internet requirements
Instrument Connect (cloud) required
Instrument internet connection required
Notes
Facial authentication
No
No
Facial authentication resides on the individual system and does not require any connectivity
Smart Help
Yes
Yes
Instrument needs to be connected to the internet to upload and download files and to be connected to the cloud
Smart Remote Support
No
No
1. AR video session requires smartphone/tablet with WiFi or cellular data connectivity to the internet to create a session between the customer and a technical specialist.
2. Desktop remote session requires a computer connected to the internet to allow a technical specialist to remotely access the customer’s computer.
QuantStudio 6 and 7 Pro Real-Time PCR Systems Support
Explore the “Getting Started” and “Troubleshooting” sections for solutions to top inquiries and common problems. Browse through our "Guides and Tools" section to access a comprehensive portfolio of instrument-related support resources.
The QuantStudio 6 and 7 Pro Real-Time PCR Systems are supported with:
  • Installation conducted on-site by professionally trained field service engineers
  • SmartStart Orientation hands-on training package conducted by field applications scientists
  • Equivalence consulting to help transition from an existing Applied Biosystems qPCR instrument
  • Smart Help on-screen, push-button help requests for technical support
  • Smart Remote Support immersive collaboration tool for faster troubleshooting and resolution
  • Qualification and validation services to help meet quality standards
On-site service plans at a glance
AB Assurance, 2PM
AB Assurance 1PM
AB Maintenance Plus
Standard on-site response time
Guaranteed 2–3 business days*
Guaranteed 2–3 business days*
2–3 business days**
Scheduled on-site planned maintenance (PM)
2 per year
1 per year
1 per year
Remote instrument diagnostics
Priority access to technical support
(Monday–Friday, standard business hours)
Parts, labor, and travel for repair
10% discount on T&M
Priority access to remote service engineer
Smart Help
Smart Remote Support
* Guaranteed response times vary by region.
** After receipt of a purchase order. Target response times vary by region.
For more information on service plans, qualification services, and training specific to your region, please visit thermofisher.com/instrumentservices or request a quote for service.
QuantStudio Pro systems
Additional blocks for both QuantStudio 6 and 7 Pro systems
Note: For interchangeable block usage, order an additional block(s) with the full system.
Automation compatible with QuantStudio 7 Pro system
Note: Barcode scanner is integrated into Orbitor arm.
TaqMan Array Plates with RFID
Product
# of samples
Cat. No.
Custom TaqMan RFID Tag
N/A
A43823
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 8
12
4413266
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 16
6
4413264
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 16 Plus
6
4413265
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 32
3
4391528
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 32 Plus
3
4391529
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 48
2
4391526
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 48 Plus
2
4391527
Learn more ›
TaqMan Array, Standard, 96-well plate;  Format 96
1
4391524
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 96 Plus
1
4391525
Learn more ›
Service and support packages
Name
Cat. No.
Price
QuantStudio 6 Pro Real-Time PCR System, 96-well, 0.2 mL block, SmartStart Orientation, 1-year AB Assurance, 1 PM†
A44288
Request a quote ›
QuantStudio 7 Pro Real-Time PCR System, 96-well, 0.2 mL block, SmartStart Orientation, 1-year AB Assurance, 1 PM†
A44289
Request a quote ›
QuantStudio 6 Pro Real-Time PCR System, 384-well block, SmartStart Orientation, 1-year AB Assurance, 1 PM†
A45582
Request a quote ›
QuantStudio 7 Pro Real-Time PCR System, 384-well block, SmartStart Orientation, 1-year AB Assurance, 1 PM†
A45583
Request a quote ›
† Packages are not available in all regions. Contact your services and support representative for availability in your region.
  • Smart features—hands-free functionality includes facial authentication login, voice commands, and RFID reader
  • Manual error reduction—compatible with TaqMan Array Plates with RFID
  • Interchangeable blocks—flexibility to change block formats between 96- (0.1 and 0.2 mL) and 384-well, and TaqMan Array Cards* via simply pushing a button or using a voice command
  • Connect—access, analyze, and share data anytime, anywhere with Connect, our cloud-based platform for real-time information access, secure data storage, and online results sharing
  • Remote access—monitor experiments and access results remotely using the Instrument Connect app
  • Small footprint—minimal bench space required: only 54.7 x 33.8 x 52.5 cm (H x W x D) and 38 kg (wt)
  • Multiplexing—5 or 6 optical channels (21 filter combinations for QuantStudio 7 Pro system)
  • Automation compatible—increase walkaway time and productivity by adding automatic loading of one or two QuantStudio 7 Pro systems with the Orbitor RS2 Microplate Mover
  • Results you can trust—10 logarithmic units of dynamic range to detect differences as small as 1.5-fold
  • Easy access to help—embedded video help from the instrument
  • Peace of mind—superior services and support with Smart Help and Smart Remote Support
QuantStudio Pro systems are compatible with Apple Mac computers.
Download specification sheet ›
Download green features flyer ›
As a qPCR pro, working smarter is your mantra. The new QuantStudio Pro systems introduce a way to incorporate smarter workflows throughout your day whether you are working at your desk, at the bench, or checking in from a remote location.
See a day in the life of a qPCR pro below and find out how your workflow can improve.
Quantstudio-graphic1
At your desk
Working at your desk has never been easier. You can:
  • Plan your experiments, design your protocols, and send them directly to your instrument of choice
  • Collaborate with people on-site or in remote locations
  • Retrieve and analyze your data at your desk; no need to go to the instrument to download or to have a dedicated computer
  • Check to see if the instrument is available and reserve future run times for optimal efficiency
At-the-bench
At the bench
Improve your day at the bench with simple, time-saving, and error-reducing features
  • Get started without removing your gloves
  • Use voice commands for common tasks such as “Start run” and “Open/close door”
  • Reduce data entry and streamline your workflow when using the TaqMan Array Plate with RFID tag
  • Change the block using the on-screen walkthrough
  • Get high-quality qPCR data seamlessly delivered to your preferred destination
Quantstudio-graphic2
Remote location
Taking a coffee break or checking the status of your experiment after work hours is easy with:
  • Remote monitoring that shows run status and amplification plots in real-time
  • Data sharing and analysis features using the cloud-enabled Connect platform
  • Planning and scheduling your next experiment without delay for ultimate productivity
Be the ultimate qPCR pro in your lab or anywhere you may be
QuantStudio 6 Pro system
QuantStudio 7 Pro system
Flexible format Interchangeable blocks with enhanced motorized change mechanism
VeriFlex Technology
3 zones
6 zones
TAC* block
upgradeable
Efficiency & productivity Small footprint
Automation for 24/7 auto plate-loading
upgradeable
Large, full-function touchscreen
Connect  platform-compatible
Features to assist with 21 CFR part 11 compliance**
upgradeable
included
Maximize uptime Smart Help
Help videos
Minimal calibration for easy maintenance
* TaqMan array microfluidic card (TAC) block available in 2020
** Module available in 2020
You can access, analyze, and share data anytime, anywhere* with the Connect cloud-based platform.
  • Remotely monitor your runs in real time
  • Analyze sophisticated data sets in minutes
  • Monitor instrument status and schedule a run from anywhere
  • Securely store data
  • Share results online with colleagues across institutions and around the world
Instrument Connect remote monitoring app
Stay connected to cloud-enabled instruments and benchtop devices anytime, anywhere, and from any mobile device to see availability, run progress, and more.
  • Check the availability of your Connect platform-enabled, network-connected device
  • Monitor run progress of a basic PCR run
  • View amplification plots in real time
  • View plots and filter by sample or target in real time
Learn more
*Where the Web is accessible
If your facility does not allow you to use the Connect cloud-based platform, don’t worry. The QuantStudio 6 and 7 Pro Real-Time PCR Systems will still improve your workflow.
You can still maximize uptime with Smart Help and Smart Remote Support features to streamline technical support help.  TaqMan Array Plates with RFID will also work to help reduce manual errors in loading your assay plate settings and loading the plate setup. And finally, the large touchscreen, ability to use the instrument without an attached computer and data delivery to your preferred location will be fully enabled to assist you.
QuantStudio 6 and 7 Pro connectivity and the internet requirements
Instrument Connect (cloud) required
Instrument internet connection required
Notes
Facial authentication
No
No
Facial authentication resides on the individual system and does not require any connectivity
Smart Help
Yes
Yes
Instrument needs to be connected to the internet to upload and download files and to be connected to the cloud
Smart Remote Support
No
No
1. AR video session requires smartphone/tablet with WiFi or cellular data connectivity to the internet to create a session between the customer and a technical specialist.
2. Desktop remote session requires a computer connected to the internet to allow a technical specialist to remotely access the customer’s computer.
QuantStudio 6 and 7 Pro Real-Time PCR Systems Support
Explore the “Getting Started” and “Troubleshooting” sections for solutions to top inquiries and common problems. Browse through our "Guides and Tools" section to access a comprehensive portfolio of instrument-related support resources.
The QuantStudio 6 and 7 Pro Real-Time PCR Systems are supported with:
  • Installation conducted on-site by professionally trained field service engineers
  • SmartStart Orientation hands-on training package conducted by field applications scientists
  • Equivalence consulting to help transition from an existing Applied Biosystems qPCR instrument
  • Smart Help on-screen, push-button help requests for technical support
  • Smart Remote Support immersive collaboration tool for faster troubleshooting and resolution
  • Qualification and validation services to help meet quality standards
On-site service plans at a glance
AB Assurance, 2PM
AB Assurance 1PM
AB Maintenance Plus
Standard on-site response time
Guaranteed 2–3 business days*
Guaranteed 2–3 business days*
2–3 business days**
Scheduled on-site planned maintenance (PM)
2 per year
1 per year
1 per year
Remote instrument diagnostics
Priority access to technical support
(Monday–Friday, standard business hours)
Parts, labor, and travel for repair
10% discount on T&M
Priority access to remote service engineer
Smart Help
Smart Remote Support
* Guaranteed response times vary by region.
** After receipt of a purchase order. Target response times vary by region.
For more information on service plans, qualification services, and training specific to your region, please visit thermofisher.com/instrumentservices or request a quote for service.
QuantStudio Pro systems
Additional blocks for both QuantStudio 6 and 7 Pro systems
Note: For interchangeable block usage, order an additional block(s) with the full system.
Automation compatible with QuantStudio 7 Pro system
Note: Barcode scanner is integrated into Orbitor arm.
TaqMan Array Plates with RFID
Product
# of samples
Cat. No.
Custom TaqMan RFID Tag
N/A
A43823
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 8
12
4413266
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 16
6
4413264
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 16 Plus
6
4413265
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 32
3
4391528
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 32 Plus
3
4391529
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 48
2
4391526
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 48 Plus
2
4391527
Learn more ›
TaqMan Array, Standard, 96-well plate;  Format 96
1
4391524
Learn more ›
TaqMan Array, Standard, 96-well plate; Format 96 Plus
1
4391525
Learn more ›
Service and support packages
Name
Cat. No.
Price
QuantStudio 6 Pro Real-Time PCR System, 96-well, 0.2 mL block, SmartStart Orientation, 1-year AB Assurance, 1 PM†
A44288
Request a quote ›
QuantStudio 7 Pro Real-Time PCR System, 96-well, 0.2 mL block, SmartStart Orientation, 1-year AB Assurance, 1 PM†
A44289
Request a quote ›
QuantStudio 6 Pro Real-Time PCR System, 384-well block, SmartStart Orientation, 1-year AB Assurance, 1 PM†
A45582
Request a quote ›
QuantStudio 7 Pro Real-Time PCR System, 384-well block, SmartStart Orientation, 1-year AB Assurance, 1 PM†
A45583
Request a quote ›
† Packages are not available in all regions. Contact your services and support representative for availability in your region.
Don’t forget to choose from our wide portfolio of quality assays, reagents, and plastics to run your experiments.