These kits are used to maintain your ViiA™ 7 System. Each of the block-specific calibration kits include calibration plates to perform a spectral calibration with FAM™, VIC®, ROX™, SYBR® Green, TAMRA™, NED™ dyes, plates to perform region-of-interest (ROI) calibration, normalization calibration, and to run an RNase P verification.
We recommend calibrating and verifying ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex Systems every six months; calibrating and verifying QuantStudio™ 12K Flex every 12 months.