Matched Antibody Pair Kits for ELISA
We offer a broad range of matched, pre-titered, and fully optimized antibody pair kits to accurately quantify cytokines, chemokines, growth factors, signaling pathway targets, and proteins associated with immunology, inflammation, cancer, cardiovascular, and neurodegenerative disease research. From prequalified antibody pairs to buffers and detection reagents, we’ll help you find the reagents you need to develop your own immunoassays when studying human, mouse, rat, swine, and chicken samples.
Popular antibody pairs
Human TNF alpha Antibody Pair Kit
Human IL-8 Antibody Pair Kit
Human IL-6 Antibody Pair Kit
Advantages of antibody pairs for ELISA
  • Quality—reliable antibodies, pre-matched antibody pairs, and proven detection reagents
  • Ease of use—simplified protocol and optimized reagents
  • Flexibility—multiple detection technologies (fluorescence, absorbance, or chemiluminescence) are possible
  • Cost savings—several times more ecomonical than complete, ready-to-use ELISA kits that contain pre-coated plates
Build and customize your own immunoassays
Our antibody pair kits enable you to build your own ELISA or any other assay platform that utilizes a matched antibody pair. The kits are designed for use with a variety of sample types such as serum, plasma, cell culture supernatant, cell lysate, tissue homogenate, urine, and cerebrospinal fluid (CSF). For convenience, we also offer a buffer set for antibody pairs that contains premade, easy-to-use buffers and solutions that are optimized for use with antibody pair kits. Each antibody pair kit supplies sufficient reagents for 10 ELISA plates (5 plates for intracellular targets).
Using Novex Antibody Pairs to make an ELISA
Typical kit components:
  • Capture antibody
  • Detector antibody
  • Recombinant standard
  • HRP conjugate
Learn more: