Buffer kits for antibody pairs
We offer several buffer kits and components to enable you to build your own immunoassay using our matched capture and detection antibody pairs. Combined with our antibody pair kits, our buffer kits work with a variety of sample types, including serum, plasma, cell culture supernatant, cell lysate, tissue homogenate, urine, and cerebrospinal fluid (CSF).
Buffer kits to help build your own immunoassay
The Buffer Kit for Antibody Pairs (CNB0011) supplies sufficient reagents for 10 ELISA plates. It includes the following components, some of which are available individually:
  • Assay buffer (5X)
  • Coating buffer A
  • Coating buffer B
  • Wash buffer (25X)
  • Stabilized chromogen (substrate)
  • Stop solution
For a complete selection of coating buffers, blocking buffers, wash buffers, substrates, coated and uncoated microplates, and reservoir trays for ELISA, see ELISA reagents and accessories.
bulk-pod-showcase
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Please click on the button below to fill out our bulk and custom ordering form. 
Complete request form
Protein quantitation immunoassays
Free white paper