Matched antibody pairs
Our matched, pre-titered, and fully optimized capture and detection antibody pair kits allow researchers studying immunology, inflammation, cancer/oncology, and cardiovascular disorders to create their own immunoassay to measure levels of protein in their biological samples.
Matched capture and detection antibody pairs
For human, mouse, rat, swine (porcine), and chicken samples, we offer several matched antibody pair options for the study of interferons, interleukins, growth factors, and more. Our antibody pairs, provided as kits, offer:
  • Quality—reliable antibodies, pre-matched antibody pairs, and proven detection reagents.
  • Ease of use—simplified protocols and optimized reagents.
  • Flexibility—suitability for fluorescence, absorbance, or chemiluminescence detection.
  • Cost savings—potential cost savings over ready-to-use ELISA kits
Our antibody pair kits enable you to build your own ELISAs or to use any other compatible assay platforms that are suitable for matched antibody pairs. Each antibody pair kit supplies sufficient reagents for 10 ELISA plates (5 plates for intracellular targets) and includes capture antibody, detection antibody, recombinant standard, and HRP conjugate. The kits work with a wide variety of sample types, including  serum, plasma, cell culture supernatant, cell lysate, tissue homogenate, urine, and cerebrospinal fluid (CSF) types.
bulk-pod-showcase
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Please click on the button below to fill out our bulk and custom ordering form. 
Complete request form
Protein quantitation immunoassays
Free white paper