Guide to ELISA Kit Package Types
Let us answer your questions about which ELISA kit is right for your research.  Whether you want to optimize your own assay or prefer a pre-optimized, ready-to-use kit for the fastest time to answer, we have an option for your research needs.
ELISA kit package types
For most targets, the detection antibody is biotinylated, in which case the HRP Conjugate is streptavidin-HRP.
Kit Contents Antibody
Pair Kits
Reagent
Kits
Precoated
Plate Kits
Coating Antibody X X Coated
Detection Antibody X X X
Calibrated Standard X X X
HRP Conjugate   X X
TMB Substrate Solution   X X
Stop Solution   X X
Standard-Sample Diluent     X
Wash Buffer     X
HRP Dilution Buffer     X
Adhesive Plate Covers     X
Precoated 96-well Strip Plate     X
Package Sizes Sufficient For: 40
plates
5
plates
1, 2 or 5
plates
ELISA Kits (with pre-coated plate) are complete, ready-to-use kits that include a detailed protocol and all the required reagents to perform a quantitative sandwich ELISA with a variety of samples (e.g. serum, plasma, cell culture supernatants). Each kit has lot-optimized reagents titrated to the pre-coated plate to ensure maximum sensitivity and ease-of-use. Any changes to the protocol may affect the ELISA results and will require additional optimization to yield the best results.
Antibody Pair Kits include validated matched pairs of antibodies and calibrated standard for custom ELISA development. Using the recommended buffers described in the detailed protocol, these kits provide bulk antibody pairs for analyzing five to forty 96-well plates. The reagents are optimized for reliable performance using the specific antibody concentrations described in the protocol, but researchers can modify and re-optimize the protocol as needed.
Reagent Kits include pre-titered coating and detection antibodies, recommended buffers and a detailed protocol for preparing a quantitative sandwich ELISA. They require users to separately purchase plates and buffers and to coat the capture antibody at the specified concentration. Reagent Kits are designed to give optimal results for cell culture supernatants with the specific protocol. However, the reagent format provides flexibility to researchers to potentially modify and re-optimize the protocol as needed.
bulk-pod-showcase
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Please click on the button below to fill out our bulk and custom ordering form. 
Complete request form
Protein quantitation immunoassays
Free white paper