Featured-microplates-ELISA-
We offer a wide selection of high-performance surface coated plates (pre-coated and pre-blocked polystyrene 96-well and 384-well microplates) in clear, white, and black. These coated microplates can be used for ELISA development and other plate-based assays with standard or fluorescent plate readers.
Popular products
NeutrAvidin® Coated Plates—Standard Capacity
NeutrAvidin® Coated Plates—High Capacity
NeutrAvidin® Coated Plates—High Sensitivity
Featured coated microplates for ELISA
Thermo Scientific Pierce biotin coated plates can be used in immunoassays with NeutrAvidin, streptavidin, avidin, or other biotin-binding proteins.
Protein A, G, A/G, and L coated plates provide alternatives to direct, passive adsorption methods for immobilizing antibodies for ELISA and other plate-based assay techniques. 
Anti-GST coated polystyrene microplates with mouse monoclonal anti-GST antibody and pre-blocked for immediate use.
Glutathione, nickel, and copper coated plates to capture and detect fusion proteins; glutathione-coated plates capture and detect glutathione-S-transferase (GST) fusion proteins, while nickel and copper bind his-tagged fusion proteins.
Microplates for binding peptides or other molecules containing either amine or sulfhydryl groups for use in binding assays or when the molecules are difficult to coat onto polystyrene plates.
Pre-blocked streptavidin and NeutrAvidin ready-to-use coated plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays. Also, check our biotin-binding plate sample pack for establishing which variety of streptavidin coated plate is best for particular assay involving biotinylated molecules.
Our antibody coated plates are useful for binding assays when available antibodies are in low quantities, denatured, and become inactive upon direct adsorption to polystyrene plates.
Protein quantitation immunoassays
Free white paper