PB4500_Luminex_Instrument_Support
We offer a variety of accessories for use with assays run on Luminex® systems as well as reagents to help you run your Luminex® protein quantitation assays. We also supply sheath fluid, calibration kits, power cords, and extended warranties.  We are committed to ensuring that you have all the necessary components you need to run successful Luminex® protein quantitation assays on MAGPIX, Luminex®200, or FlexMAP 3D® Luminex® systems.
If your Luminex® instrument needs attention or you require technical assistance with your Luminex® assays or xPONENT® software for data acquisition and analysis, contact our Technical Support team or email LuminexFAS@lifetech.com.
Featured instrument accessory: Magnetic 96-well Separator
The Magnetic 96-Well Separator is designed for fast and simple removal of supernatant from samples bound to magnetic particles and may be used manually or as an integrated part of an automated workstation. The Magnetic 96-Well Separator is an ideal tool for use in singleplex and multiplex biological assays based on the Luminex® xMAP® Technology. These assays make use of Luminex® MagPlex™, or magnetic microspheres, and the MAGPIX®, Luminex® 200™, or FLEXMAP 3D® instruments.
Find additional accessories by category
Luminex® Assays
See our selection of Singleplex Luminex® Assays and Multiplex Luminex® Assays. Or search our catalog for immunoassays using the Immunoassay Selection Guide.
If you can't find the assay that matches your experiment, contact us about a custom Luminex® assay.
Protein quantitation immunoassays
Free white paper