Rapid protein gel staining and semi-dry blotting with one device
The Thermo Scientific™ Pierce™ Power System is a device that can be used for both fast Coomassie dye staining/destaining of protein gels and rapid semi-dry transfer of proteins from gels to membranes. Choose the entire system—power station, stain cassette and blot cassette—to accomplish both rapid staining and blotting. Alternatively, select the Pierce™ Power Stainer or Pierce™ Power Blotter. You can easily expand functionality later by purchasing the appropriate cassette.
Order now  
Rapid and efficient protein staining or protein transfer at your fingertips
The complete Pierce™ Power System consists of three Thermo Scientific Pierce components:
  • Power Station—features an integrated power supply, easy-to-use color LCD touch screen interface, and ready-to-run electrophoretic gel staining and protein transfer programs; also enables custom settings to be quickly created, saved and run
  • Power Stain Cassette—facilitates effective Coomassie dye staining (including background destaining) of protein midi or mini gels in just 6–11 minutes when used with the optimized Pierce™ Power Staining Kits
  • Power Blot Cassette—enables efficient semi-dry transfer of protein from midi or mini gels to nitrocellulose or PVDF membranes in about 10 minutes when used with the optimized Pierce™ 1-Step Transfer Buffer; also effective for standard 30–60 minute semi-dry transfer protocols using traditional Towbin transfer buffers
Pierce Power System buying guide
Choose the right Thermo Scientific™ Pierce™ Power System products and accessories for your needs.
Learn more Learn more Learn more
Pierce Power Stainer
Pierce Power Blotter
Pierce Power System
Protein gel application(s) Stain proteins with Coomassie dye Transfer proteins to membrane Stain proteins with Coomassie dye and transfer proteins to membrane
Power system contents Power station
Stain cassette
Power station
Blot cassette
Power station
Stain cassette
Blot cassette
Simultaneous processing capacity 1 midi gel
or 2 mini gels
2 midi gel blots
or 4 mini gel blots
1 midi gel or 2 mini gels;
2 midi gel blots or 4 mini gel blots
Recommended consumables Pierce Power Staining Kits (22839, 22840) Pierce 1-Step Transfer Buffer (84731)
Blotting Filter Paper (84738, 84784)
Pierce Power Staining Kits (22839, 22840)
Pierce 1-Step Transfer Buffer (84731)
Blotting Filter Paper (84738, 84784)
Already own a Thermo Scientific™ Pierce™ G2 Fast Blotter (Catalog #62288)?
Upgrade now and start rapid staining, too!
Pierce Power System:
One station + two cassettes = powerful possibilities
Rapid protein staining (complete with background destaining) and protein transfer using the Thermo Scientific™ Pierce™ Power System. The system combines features of the Pierce™ Power Stainer and the Pierce™ Power Blotter into a single time-saving system.
One system for faster results in two protein detection workflows
The Thermo Scientific Pierce Power System helps to accomplish a complete workflow (gel electrophoresis, protein staining and western blot detection) in about 5 hours. A. Serial dilutions of HeLa cell lysate (10 to 0.0625 µg) were loaded in two 4-20% Tris-Glycine gels and separated by electrophoresis (70 minutes). B. One gel was washed and then stained using the Pierce Power Stainer and the Pierce Mini Gel Power Staining Kit (~12 minute protocol). The second gel was transferred to nitrocellulose membrane using Pierce Power Blotter and 1-Step Transfer Buffer (10 minute protocol). C. The blot was washed and stained with Pierce Reversible Protein Stain (18 minute protocol), imaged (~1 minute). The blot was cleared of stain (8 minute protocol), blocked (30 minutes), incubated in HDCA1 polyclonal (60 minutes), washed (30 minute protocol), incubated with goat-anti-rabbit-HRP conjugated secondary antibody (30 minutes) and washed (30 minutes). The blot was developed in Thermo Scientific™ SuperSignal™ West Dura Chemiluminescent Substrate (5 minutes) and D. imaged on myECL Imager (5 minute protocol). Total protocol time: ~5 hr.
Video of the Pierce Power System in action!
Summary of the Thermo Scientific™ Pierce™ Power System
Includes:
  • Pierce Power Station
  • Pierce Power Stain Cassette
  • Pierce Power Blot Cassette
  • Western Blot Roller
  • Power Cord with C/13 Connector
Applications:
  • Coomassie staining and destaining of proteins in SDS-PAGE gels in 6-11 minutes
  • Semi-dry Western blot transfer of 10-300 kDa proteins from SDS-PAGE gels to nitrocellulose or PVDF membranes in 5-10 minutes
Requires:
Pierce Power System Welcome Pack:
This special package offers everything you need for rapid Coomassie protein staining or rapid protein transfer. With the Pierce Power System welcome pack, you’ll receive:
  • Pierce Power System, Catalog #22830
  • Pierce Mini Gel Power Staining Kit, Catalog #22840
  • Pierce 1-Step Transfer Buffer, Catalog #84731
  • Western Blotting Filter Paper, 7 cm x 8.4 cm, 100 sheets, Catalog #84783
Order the welcome pack