File Name: WesternV2.00.sim
Version: 2.0
Date Published: 9/15/10
Language: English
Download size: 29 MB
Download
Changes to Previous Firmware Version (1.8)
  • The new firmware (version 2.0) adds a manual PAUSE function during a run. This can be used to pause the instrument during a run so that individual steps in the protocol may be performed manually.
  • The new firmware also specifies that the liquid flow of an aborted run is directed to waste, rather than to all the outlets.
Download Requirements:
USB Flash Drive—40 MB available space (minimum)
Instructions:
  1. Download the firmware to the USB flash drive.
  2. With the power off, insert the USB flash drive into the BenchPro™ 4100.
  3. Turn the power on; the BenchPro™ 4100 will automatically recognize the firmware file.
  4. Follow the directions on the BenchPro™ 4100 screen.