On this page:
Choose western blot buffers based on application needs and performance requirements such as sensitivity and reproducibility. Compare transfer, blocking, wash, and stripping buffers to reduce background and improve signal clarity.
Overview of western blot buffers
Western blotting success depends on more than antibodies and detection reagents—the buffers used at each step of the workflow directly influence sensitivity, background, and reproducibility. From efficient protein transfer to clean antibody detection and membrane reprobing, selecting the right buffer is important for generating reliable, publication-quality results.
Western blot buffer types
Western blotting workflows typically use multiple types of buffers, each designed to support a specific step in the process, and can have an impact on transfer efficiency, background noise, and signal quality.
  • Transfer buffers—support efficient protein transfer from gel to membrane
  • Blocking buffers—minimize nonspecific binding before antibody incubation
  • Wash buffers—remove unbound antibody prior to detection
  • Stripping buffers—strip/remove antibodies from membrane for membrane reprobing
Each buffer type contributes to overall assay performance by influencing protein stability, antibody specificity, detection sensitivity, and reproducibility. Selecting the appropriate buffer for each step from the beginning results in less downstream optimization and improves confidence in experimental results.
The table below summarizes when to use each buffer type, typical formulations, and commonly used options within the western blotting workflow.
Step Transfer buffers Blocking buffers Wash buffers Stripping buffers
When to use Gel-to-membrane transfer Reduce background before antibody binding Remove unbound antibody Reprobing membranes
Buffer type Tris-glycine (Towbin), Bis-Tris, Tris-acetate Milk, BSA, casein, specialized blocker TBS, TBS-T, PBS, PBS-T Mild or harsh stripping buffer
Common composition With or without methanol/SDS 3–5% in TBS-T/PBS-T 0.05–0.1% Tween-20 Various
Related products
Need a running buffer? Protein electrophoresis buffers and reagents.
Western blot transfer buffers guide
Quick selection guide: Which transfer buffer should you use?
  • Using Bis-Tris gels (Bolt or NuPAGE)? Choose Bolt or NuPAGE Bis-Tris transfer buffer
  • Working with small proteins (<40 kDa)? Choose Novex Tris-Glycine transfer buffer
  • Working with large proteins (>150 kDa)? Choose NuPAGE Bis-Tris transfer buffer
  • Need fast transfer? Choose semi-dry 1-step transfer buffers
How to choose the right transfer buffer
Selecting the right transfer buffer depends on your gel chemistry, protein size, and transfer method.
  1. Match your gel chemistry (Tris-Glycine, Tricine, Bis-Tris, or Tris-Acetate)
  2. Select transfer method (wet = flexibility, semi-dry = speed)
  3. Optimize conditions (standard vs. high ionic strength buffers)
Compare western blot transfer buffers
The table below summarizes recommended transfer buffers based on gel chemistry, protein size, and transfer method.
  Wet transfer Semi-dry transfer
Transfer buffer Novex Tris-Glycine transfer buffer Bolt Bis-Tris transfer buffer
Reduced samples: Antioxidant*
NuPAGE Bis-Tris transfer buffer
Reduced samples: Antioxidant*
Power Blotter 1-Step Transfer Buffer (5X)** Pierce 1-Step Transfer Buffer (1X)**
When to use General-purpose wet transfer for broad protein size ranges Controlled transfer of low-molecular-weight proteins resolved on Tricine gels Optimized transfer performance for Bolt Bis-Tris Plus gel systems Consistent transfer optimized for NuPAGE Bis-Tris gel chemistry Efficient transfer of high-molecular-weight proteins from Tris-Acetate gels High-ionic-strength buffer enabling rapid semi-dry protein transfer Ready-to-use buffer for fast, reproducible semi-dry transfers
Gel chemistry Novex Tris-Glycine Novex Tricine Bolt Bis-Tris Plus NuPAGE Bis-Tris NuPAGE Tris-Acetate All gel chemistries
*Antioxidant prevents re-oxidation of reduced proteins during transfer, improving band integrity
**1-Step Transfer buffer is a high ionic strength formulation which allows for 5-minute to 12-minute protein transfer when used with compatible semi-dry blotting systems. 1-Step Transfer Buffer is compatible with Power Blotter and other protein semi-dry transfer devices, when they are paired with a suitable high-current power supply. Such devices should help provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.
What do western blot transfer buffers do? 
Protein transfer is a critical step in western blotting. Transfer buffers help provide the ionic strength and pH required to move proteins efficiently from the gel to a membrane while preserving protein integrity and epitope accessibility. The buffer composition directly affects transfer efficiency, speed, and uniformity, particularly across different molecular weight ranges. Transfer buffers also affect compatibility with nitrocellulose or PVDF membranes and determine whether proteins remain bound during downstream detection steps.
Why transfer buffers matter
Using a premixed transfer buffer matched to your gel chemistry and workflow helps to:
  • Support efficient electrotransfer under wet and semi-dry conditions
  • Maintain protein solubility and structural integrity
  • Improve consistency across low-, broad-, and high-molecular-weight proteins
  • Reduce incomplete transfer or protein loss
When to choose ready-to-use vs. DIY transfer buffers
Thermo Fisher Scientific offers premixed, concentrated, and specialized transfer buffers optimized for different gel chemistries and transfer systems, helping ensure consistent results while reducing preparation time.
  • Choose ready-to-use transfer buffers when consistency, speed, and reproducibility are critical, especially in high-throughput or multi-user labs.
  • Prepare transfer buffers from scratch when protocols require customization or cost control is a priority.
Tip: For most workflows, matching a ready-to-use transfer buffer to your gel system helps provide the fastest path to reliable results.
The following transfer buffer recipes are provided to allow preparation of buffers from scratch.
25X Tris-Glycine transfer buffer
1. Dissolve the following reagents in 400 mL ultrapure water.
Reagent Amount Concentration (1X)
Tris Base 18.2 g 12 mM
Glycine 90 g 96 mM
2. Mix well and adjust volume to 500 mL with ultrapure water.
3. Before western transfer, dilute to 1X with water.
4. Buffer is stable for 6 months at room temperature.
20X Bis-Tris transfer buffer
1. Dissolve the following reagents in 100 mL ultrapure water.
Reagent Amount Concentration (1X)
Bicine 10.2 g 25 mM
Bis-Tris (free base) 13.1 g 25 mM
EDTA 0.75 g 1 mM
2. Mix well and adjust volume to 125 mL with ultrapure water.
3. Before western transfer, dilute to 1X with water.
4. Buffer is stable for 6 months when stored at 4°C.
Western blot blocking buffers guide
Quick selection guide: Which blocking buffer should you use?
  • General chemiluminescent western blotting? Choose StartingBlock Blocking Buffer.
  • High background or low signal with milk? Switch to Blocker Casein.
  • Detecting phosphoproteins or using biotin/streptavidin systems? Choose Blocker BSA.
  • Fluorescent western blotting or imaging? Choose Blocker FL Fluorescent Blocking Buffer.
How to choose the right blocking buffer
Selecting the right blocking buffer depends on your detection method, nature of the target protein, background levels, and signal intensity.
  1. Choose your detection method (chemiluminescent vs fluorescent western blotting).
  2. Consider your target protein (phosphoproteins, biotin/streptavidin systems, or routine targets).
  3.  Evaluate background and sensitivity (high background = switch blockers; weak signal = adjust blocking conditions).
Compare western blot blocking buffers
The table below summarizes recommended blocking buffers based on detection method, target type, and experimental conditions.
Chemiluminescent western blotting (HRP/AP)
Fluorescent western blotting
Blocking buffer
StartingBlock
Blocker Casein
Blocker BSA
Blocker FL
Why use
General-purpose blocker for chemiluminescent detection
Improved sensitivity, high-performance replacement for milk-based blockers
Phosphoproteins or biotin-based chemiluminescent detection
Performing fluorescent western detection
Blocking agent
Serum- and biotin-free single purified protein
Purified protein
Purified bovine serum albumin
Single purified protein
Highlights
  • Low background across wide range of antibodies and antibody combinations.
  • Compatible with streptavidin systems
  • Blocks in less than 15 minutes
Single protein blocking buffer helps provide fewer chances of cross-reaction with assay components than serum or milk solutions
  • 10% solutions of high-quality bovine serum albumin
  • Single purified protein reduces interference from phosphoproteins and biotin
  • Blocks excess nonspecific binding sites to help reduce background fluorescence
  • Works with both nitrocellulose and low fluorescence PVDF membranes
  • Detergent-free
  • Blocks in 15–30 minutes
Available formats
PBS, TBS, PBST, TBST
PBS, TBS
PBS, TBS
10X concentrate
User guides
StartingBlock blocking buffers user guide
Blocker casein blocking buffers user guide
Blocker BSA blocking buffers user guide
Blocker FL 10X fluorescent blocking buffer user guide
Tip: If changing antibody concentration doesn’t improve results, change the blocking buffer before changing the antibody.
What do western blot blocking buffers do? 
After protein transfer, membranes contain both immobilized target proteins and unoccupied binding sites. If these sites are not blocked, primary or secondary antibodies can bind nonspecifically, leading to elevated background and reduced signal-to-noise ratio. Blocking buffers coat these remaining unoccupied binding sites so antibodies bind selectively to their intended targets, preventing nonspecific binding, reducing background and improving signal clarity.
An effective blocking buffer binds nonspecific interaction sites without masking the target epitope or interfering with antibody binding.
Which Blocking Buffer Should I Use for Each Detection Method?
Detection method Recommended Thermo Fisher Blocking Buffers Why
Chemiluminescent (HRP)
StartingBlock Blocking Buffer
Compatible with HRP detection and reduces nonspecific binding
Alkaline phosphatase (AP) SuperBlock Blocking Buffer Phosphate inhibits AP; use TBS-based formulations
Fluorescent (NIR) Blocker FL Fluorescent Blocking Buffer Minimizes autofluorescence and imaging background
Multiplex fluorescence Blocker FL Fluorescent Blocking Buffer Supports low-background detection of multiple targets
Biotin/streptavidin-based detection StartingBlock Blocking Buffer; SuperBlock Blocking Buffer; Protein-Free Blocking Buffer Avoids endogenous biotin interference
Blocking buffer formats and options
Thermo Fisher provides blocking buffer in multiple formats to support different workflows:
  • Ready-to-use blocking buffers for convenience and consistency
  • Concentrated formats for flexible dilution
  • Blocking buffers optimized for chemiluminescent and fluorescent detection
  • Single-protein blockers designed to reduce cross-reactivity or improve sensitivity
These formats support applications such as:
  • Low-background imaging
  • Multiplex fluorescence detection
  • High-sensitivity chemiluminescence
Blocking buffer timing and conditions
Typical blocking times range from 15 to 60 minutes at room temperature, depending on the blocking buffer used. Increasing blocking time does not always reduce background and may reduce sensitivity.
5% nonfat milk (for routine chemiluminescent detection)
Nonfat dry milk 2.5 g
1X TBST or 1X PBST Up to 50 mL
Filter to remove particulates  
Ready-to-use alternative:
3% BSA (phosphoproteins or biotin-based assays)
BSA 1.5 g
1X TBST or 1X PBST Up to 50 mL
Filter to remove particulates  
Ready-to-use Thermo Fisher blocking buffers are recommened when reproducibility, convenience, or reduced optimization time is critical.
Ready-to-use alternatives: Pierce Blocker BSA (10X) in TBS and Pierce Blocker BSA (10X) in PBS.
Western blot wash buffers guide
Quick selection guide: Which wash buffer should you use?
  • Standard HRP-based western blot washing? Use PBS or PBST (PBS + Tween-20).
  • Need to reduce background (HRP or AP workflows)? Use TBST (TBS + Tween-20).
  • Alkaline phosphatase (AP) detection? Use TBS (avoid PBS, which inhibits AP).
  • High background? Use TBST or PBST with 0.05–0.1% Tween-20.
  • Persistent background? Increase Tween-20 concentration (up to ~0.2%).
  • Fluorescent western blotting? Use detergent-free buffers (TBS or PBS).
How to choose the right wash buffer
Choosing the right wash buffer depends on your detection chemistry, need for detergent, and background vs. signal balance.
  1. Match your detection chemistry (HRP → PBS or TBS; AP → TBS only).
  2. Decide if detergent is needed (high background → add Tween-20; weak signal → reduce or omit detergent).
  3. Optimize detergent concentration (Typical: 0.05–0.1% Tween-20; increase for high background, decrease for weak signal).
  4. Consider imaging requirements (fluorescent detection → avoid detergents during blocking; use only after blocking step, as needed).
Compare western blot wash buffers
The table below summarizes common wash buffers and formats based on detection method, background control, and workflow requirements.
Choose from dry blend packs or concentrated solutions (10X or 20X) of common wash buffers used in western blotting.
TBS
PBS
TBST (+ Tween-20)
PBST (+ Tween-20)
Best for
Alkaline phosphatase (AP) –based detection and detergent-free washing
General HRP-based chemiluminescent western blot washing
Reducing nonspecific background in HRP or AP detection workflows
Routine HRP detection for removing potential high background/unbound reagents
Dry blend
BupH Tris Buffered Saline Packs
BupH Phosphate Buffered Saline Packs
Liquid conc.
Pierce 20X TBS Buffer
Pierce 20X Phosphate Buffered Saline
Pierce 20X TBS Tween 20 Buffer 
Pierce 20X PBS Tween 20 Buffer 
Formulation
  • 25 mM Tris
  • 0.15 M NaCl
  • pH 7.2
  • 10 mM sodium phosphate
  • 0.15 M NaCl
  • pH 7.5
  • 25 mM Tris
  • 0.15 M NaCl
  • 0.05% Tween-20
  • pH 7.5
  • 10 mM sodium phosphate
  • 0.15 M NaCl
  • 0.05% Tween-20
  • pH 7.5
For use with fluorescent systems
Yes
Yes
No
Tip: If background remains high, increase wash number or duration before increasing detergent concentration.
What do western blot wash buffers do?
Wash buffers remove unbound antibodies and reagents between incubation steps, reducing background while preserving specific signal. Insufficient washing produces high background, while excessive washing may result in decreased sensitivity caused by stripping of the antibody and/or antigen from the blot.
Washing is performed in physiological buffers such as Tris-buffered saline (TBS) or phosphate-buffered saline (PBS). Detergents such as Tween-20 can be added to the buffer to help remove nonspecifically bound material. The amount of Tween-20 (0.05%–0.2%) will vary depending on the strength of the antibodies used. Weak binding antibodies may be washed away by too much detergent.
Why wash buffers matter
Effective washing helps to:
  • Reduce nonspecific signal by removing loosely or nonspecifically bound antibodies that contribute to background
  • Improve assay sensitivity by increasing the signal-to-noise ratio, making true target bands easier to detect
  • Maintain consistent signal across replicates by ensuring uniform removal of excess reagents
Insufficient washing can result in high background and false-positive signals, while overly stringent washing may reduce sensitivity by removing weakly bound antibodies or target proteins.
Wash protocol recommendations
  • Typical washes: 3–5 washes, 5–10 minutes each
  • Increase detergent or wash number for high background
  • Reduce detergent concentration if signal is weak
Tris-buffered saline with Tween 20 (TBST)
10X TBS 100 mL
Tween 20 1 mL
Deionized water to 1,000 mL
Phosphate buffered saline with Tween 20 (PBST)
10X TBS 100 mL
Tween 20 1 mL
Deionized water to 1,000 mL
Western blot stripping buffers guide
Quick selection guide: Which stripping buffer should you use?
  • Need gentle stripping for sensitive or low-affinity antibodies? Use Restore Western Blot Stripping Buffer.
  • Removing high-affinity antibodies or performing multiple reprobes? Use Restore PLUS Western Blot Stripping Buffer.
  • Fluorescent (NIR) western blot detection? Use Restore Fluorescent Western Blot Stripping Buffer.
  • Working with valuable or limited samples? Choose Restore Stripping Buffer to preserve immobilized protein.
  • Need faster or more complete antibody removal? Use Restore PLUS Stripping Buffer or extended protocol.
How to choose the right stripping buffer
Choosing the right stripping buffer depends on antibody affinity, detection method, and how well the membrane must be preserved.
  1. Determine if stripping is necessary (multiple targets, antibody optimization, or limited samples).
  2. Assess antibody affinity (low/moderate → mild buffer; high affinity → stronger buffer).
  3. Match your detection method (chemiluminescent → standard buffers; fluorescent → fluorescence-compatible buffers).
  4. Protect membrane integrity (minimize time, temperature, and number of stripping cycles).
Compare western blot stripping buffers
The table below summarizes stripping buffers based on antibody affinity, detection method, and membrane compatibility.
  Restore Stripping Buffer Restore PLUS Stripping Buffer Restore Fluorescent Western Blot Stripping Buffer
Type of buffer Standard/mild Strong Fluorescent-compatible
When to use For routine and sensitive/low-affinity antibodies For removing high-affinity primary antibodies For removing fluorescent antibodies
Features Gentle, odor-free • Robust yet gentle, odor free
•Transferred proteins remain viable and can strip the same blot up to 5 times
• Gentle and highly effective for removing primary and fluorescent secondary antibodies
• Optimized for fluorescent western blot workflows
Membrane NC and PVDF NC and PVDF Low-fluorescence PVDF membranes recommended
Time of incubation 15–30 min at 37°C 5–15 mins at RT or 37°C for high affinity antibodies 10–20 min at RT
User guide Restore Western Blot Stripping Buffer User Guide Restore Plus Western Blot Stripping Buffer User Guide Restore FL Western Blot Stripping Buffer User Guide
Technical tip: Strip and reprobe western blots
Follow step-by-step guidance for stripping antibodies and reprobing western blot membranes.
What do western blot stripping buffers do? 
Stripping buffers are designed to remove bound primary and secondary antibodies from western blots so that membranes can be reprobed under alternate conditions or with another antibody to detect a different protein target, without rerunning another western blot. Reprobing a western blot provides an efficient way to analyze additional targets while conserving samples and allowing optimization as needed.
Types of stripping buffers
Stripping buffers vary in strength and formulation to support different antibodies, detection methods, and membrane reuse needs. Choosing the appropriate type helps ensure effective antibody removal while preserving immobilized proteins.
  • Mild stripping buffers: For low- to moderate-affinity antibodies or sensitive targets, helping provide gentle antibody removal with minimal protein loss
    • Recommended for limited reprobing cycles
  • Strong stripping buffers: For high-affinity or difficult-to-remove antibodies, offering more aggressive stripping when complete antibody removal is required
    • Recommended when complete antibody removal is required and target protein abundance is sufficient
  • Fluorescent-compatible stripping buffers: Optimized for fluorescent western blotting, including near-infrared (NIR) detection, enabling efficient removal of dye-labeled antibodies while minimizing membrane autofluorescence
Tip: Start with the mildest stripping buffer that effectively removes antibodies and increase strength only if needed.
When to strip and reprobe
Stripping is recommended when:
  • Antibody optimization is required
  • Sample availability is limited
Advantages of stripping and reprobing blots
There are several major reasons to choose to strip and reprobe a western blot. Following are some of the most important:
  • Conserves sample: When the protein mixture is rare or valuable, reprobing conserves the sample and allows the membrane to be analyzed with the same or different antibodies.
  • Improves workflow efficiency: Reprobing allows for the detection of multiple proteins from the same blot, reducing the need to repeat earlier western blot workflow steps.
  • Facilitates assay optimization: The light emission intensity of high-sensitivity chemiluminescent substrates often require antibody concentration optimization to achieve the highest quality blot. Optimization is achieved easily by stripping the membrane and reprobing with different antibody concentration.
  • Allows for correcting mistakes: Immunoblotting requires many steps, giving ample opportunity for mistakes to occur. By stripping the membrane, the blot can be reused.
0.5 M Tris HCl, pH 6.8 12.5 mL
10% SDS 20 mL
2-mercaptoethanol 0.8 mL
Deionized water 67.5 mL
How-to videos
How do I probe a western blot?
Probing is a critical stage of western blotting that determines signal specificity, sensitivity, and background. In this video, you’ll learn how to correctly probe a western blot after protein transfer, including blocking, antibody incubation, and washing steps, to help ensure reliable and reproducible detection.
You’ll learn how to:
  • Select and apply the appropriate blocking buffer to reduce nonspecific binding
  • Prepare and dilute primary and secondary antibodies
  • Perform effective washing steps to minimize background and preserve signal
How do I strip and reprobe my western blot?
Stripping and reprobing allows a single western blot membrane to be reused for detection of multiple targets, helping save sample and reagents. In this video, you’ll learn how to safely remove bound antibodies and reprobe the membrane without compromising immobilized proteins or signal quality.
You’ll learn how to:
  • Select the appropriate stripping buffer based on antibody affinity and detection method
  • Optimize stripping conditions to remove antibodies while preserving target proteins
  • Handle membranes during stripping to preserve immobilized proteins
Ordering information
Catalog # Name Size Price Qty
BT0005 Each
41.65
Online exclusive
43.75 Save 2.10
BT0006 Each
35.65
Online exclusive
37.22 Save 1.57
BT00061 Each
240.65
Online exclusive
248.00 Save 7.35
NP0005 Each
42.84
NP0006 Each
36.81
NP00061 Each
243.00
NP0050 Each
138.65
Online exclusive
144.00 Save 5.35
LC3675 Each
57.25
PB7100 Each
107.65
Online exclusive
112.00 Save 4.35
PB7300 Each
341.65
Online exclusive
356.00 Save 14.35
84731 Each
100.00
84731X5 Each
406.00
Catalog # Name Size Price Qty
37515 Each
228.65
Online exclusive
276.00 Save 47.35
37516 Each
251.65
Online exclusive
303.00 Save 51.35
37517 Each
234.65
Online exclusive
255.00 Save 20.35
37518 Each
750.65
Online exclusive
904.00 Save 153.35
37520 Each
199.65
Online exclusive
224.00 Save 24.35
37525 Each
214.65
Online exclusive
241.00 Save 26.35
37525X3 3 x 200 mL
485.65
Online exclusive
546.00 Save 60.35
37527 Each
250.65
Online exclusive
279.00 Save 28.35
37527X3 Each
599.65
Online exclusive
666.00 Save 66.35
37528 Each
182.65
Online exclusive
208.00 Save 25.35
37530 Each
188.00
37532 Each
182.65
Online exclusive
208.00 Save 25.35
37535 Each
234.65
Online exclusive
283.00 Save 48.35
37536 Each
251.65
Online exclusive
303.00 Save 51.35
37537 Each
233.65
Online exclusive
254.00 Save 20.35
37538 Each
242.65
Online exclusive
285.00 Save 42.35
37538X3 3 x 1 L
548.65
Online exclusive
646.00 Save 97.35
37539 Each
259.65
Online exclusive
305.00 Save 45.35
37542 Each
242.65
Online exclusive
286.00 Save 43.35
37543 Each
257.65
Online exclusive
303.00 Save 45.35
37545 Each
188.65
Online exclusive
227.00 Save 38.35
37549 Each
231.00
37565 Each
175.00
37570 Each
233.65
Online exclusive
263.00 Save 29.35
37571 Each
265.65
Online exclusive
298.00 Save 32.35
37572 Each
244.65
Online exclusive
275.00 Save 30.35
37573 Each
262.65
Online exclusive
295.00 Save 32.35
37575 Each
326.00
37576 Each
186.00
37578 Each
52.65
Online exclusive
62.50 Save 9.85
37579 Each
52.65
Online exclusive
62.50 Save 9.85
37580 Each
52.65
Online exclusive
63.00 Save 10.35
37581 Each
52.65
Online exclusive
63.00 Save 10.35
37584 Each
55.65
Online exclusive
62.50 Save 6.85
37585 Each
55.65
Online exclusive
62.50 Save 6.85
37587 Each
202.65
Online exclusive
220.00 Save 17.35
46640 Each
322.65
Online exclusive
362.00 Save 39.35
46641 Each
66.65
Online exclusive
75.25 Save 8.60
Catalog # Name Size Price Qty
28320 Each
167.65
Online exclusive
184.00 Save 16.35
85115 Each
1,202.65
Online exclusive
1,322.00 Save 119.35
28341 Each
114.65
Online exclusive
127.00 Save 12.35
28344 Each
124.65
Online exclusive
139.00 Save 14.35
28346 Each
126.65
Online exclusive
141.00 Save 14.35
28348 Each
138.65
Online exclusive
154.00 Save 15.35
28352 Each
116.65
Online exclusive
130.00 Save 13.35
28354 Each
115.65
Online exclusive
128.00 Save 12.35
28355 Each
149.65
Online exclusive
166.00 Save 16.35
28358 Each
115.65
Online exclusive
128.00 Save 12.35
28360 Each
116.65
Online exclusive
130.00 Save 13.35
28362 Each
88.65
Online exclusive
99.00 Save 10.35
28363 Each
88.65
Online exclusive
99.00 Save 10.35
28372 Each
207.65
Online exclusive
233.00 Save 25.35
28374 Each
209.65
Online exclusive
235.00 Save 25.35
28376 Each
206.65
Online exclusive
232.00 Save 25.35
28379 Each
100.65
Online exclusive
113.00 Save 12.35
28380 Each
176.65
Online exclusive
199.00 Save 22.35
28382 Each
207.65
Online exclusive
233.00 Save 25.35
28384 Each
206.65
Online exclusive
232.00 Save 25.35
28390 Each
174.65
Online exclusive
196.00 Save 21.35
Catalog # Name Size Price Qty
21059 Each
181.65
Online exclusive
216.00 Save 34.35
21059X4 Each
576.65
Online exclusive
686.00 Save 109.35
21062 Each
54.65
Online exclusive
65.00 Save 10.35
21063 Each
755.65
Online exclusive
900.00 Save 144.35
46428 Each
53.65
Online exclusive
62.00 Save 8.35
46430 Each
189.65
Online exclusive
218.00 Save 28.35
46430X4 Each
601.65
Online exclusive
692.00 Save 90.35
62299 Each
58.65
Online exclusive
63.50 Save 4.85
62300 Each
198.65
Online exclusive
216.00 Save 17.35
62300X4 Each
597.65
Online exclusive
650.00 Save 52.35
Frequently asked questions
Blocking buffer selection depends on detection method and target protein. Milk-based blockers work well for routine chemiluminescent detection. BSA or purified-protein blockers are preferred for phosphoproteins or biotin-based assays. Fluorescent western blotting requires fluorescence-compatible blocking buffers to minimize background.
High-molecular-weight proteins transfer more efficiently when using gel chemistries and transfer buffers designed for large proteins, such as Tris-Acetate systems. For wet transfer, reduce methanol to 10–15% and add up to 0.02% SDS to improve mobility. Additionally, increased transfer time and using PVDF membranes can enhance binding and overall transfer efficiency.
PBS-T and TBS-T are suitable for most western blots. However, TBS-T is recommended for alkaline phosphatase (AP) detection and phosphorylation studies because phosphate in PBS can interfere with AP-based systems. Choose the buffer based on detection chemistry and background performance.
Most western blot workflows use 0.05–0.1% Tween-20 in TBS or PBS. If background is high increase detergent up to 0.2% if signal is weak reduce detergent 0.02–0.05%. Alternatively, adjust wash duration or frequency before significantly increasing detergent concentration.
Strip and reprobe a western blot when analyzing multiple targets, optimizing antibodies, or conserving limited samples. Use mild stripping buffers for low-affinity antibodies and stronger buffers for high-affinity antibodies. PVDF membranes are generally preferred for repeated stripping due to their durability.
Reusing transfer buffer is not recommended. Ion depletion, pH changes, methanol loss, and heat generation alter buffer performance during transfer. Using fresh buffer helps ensure consistent transfer efficiency and reproducible results. 
Consider using an antioxidant when you need enhanced sensitivity or improved band quality, particularly for low-abundance or difficult-to-transfer proteins or challenging targets. Antioxidants help improve protein transfer efficiency, which can result in stronger signal intensity, better sensitivity, and sharper, more defined bands.
Related products
Western blot educational resources
Access resources to help optimize western blot detection strategies, troubleshoot experiments, and improve data quality.
For Research Use Only. Not for use in diagnostic procedures.