Search
Choose western blot membranes and filter papers based on transfer system, membrane chemistry, and detection requirements. Compare membrane types, formats, and filter paper options to optimize detection performance.
Selecting the appropriate membrane chemistry and filter paper format directly impacts transfer efficiency, background signal, detection sensitivity, and the ability to strip and reprobe blots.
Protein transfer is easy and convenient with our pre-cut, pre-assembled western blot membrane/filter paper sandwiches designed to fit your gels. Alternatively, membrane rolls, sheets, and filters papers are available to enable building your own transfer sandwich.
Quick selection guide: Which membrane and format should you use?
How to choose the right membrane and format
The tables below compare membrane formats and transfer systems based on ease of use, flexibility, and compatibility with different western blot transfer systems.
Wet tank transfer membrane options
| Format | Great for | Ease of use | Nitrocellulose | PVDF | Low fluorescence PVDF |
|---|---|---|---|---|---|
| Roll | Flexible membrane dimensions | + | Nitrocellulose Membranes, 0.45 µm, 1 Roll | Low Fluorescence PVDF Transfer Membranes, 0.3 µm, 1 Roll | |
| Pre-cut sheets | Consistency, reduced risk of damage due to handling, reduced waste | ++ | PVDF Transfer Membranes, 0.45 µm, 10 x 10 cm | ||
| Membrane/filter paper sandwich | Ready-to-use for wet transfer, no assembly required | +++ |
For rapid dry and semi-dry transfer systems, like the iBlot 3 Western Blot Transfer System and Power Blotter Semi-Dry Transfer System, specialized transfer stacks are available.
For fast workflows, pre-configured transfer stacks and membrane systems are available for rapid transfer platforms such as iBlot and Power Blotter systems.
Rapid transfer system membrane options
| Transfer System | Best for | Ease of use | Nitrocellulose | PVDF | Low fluorescence PVDF |
|---|---|---|---|---|---|
| iBlot 3 system | Fastest setup, clean-up, and transfer time (3–8 minutes) | +++++ | |||
| Power Blotter | Rapid transfer (5–10 minutes), flexible membrane and filter paper options | ++++ | The low fluorescence PVDF membranes listed in the wet tank transfer membrane options table above are compatible with Power Blotter. |
The table below compares nitrocellulose, PVDF, and low-fluorescence PVDF membranes based on protein binding, background, durability, and compatibility with different detection methods.
How to choose between nitrocellulose and PVDF membranes
| Nitrocellulose membrane | PVDF membrane | Low-fluorescence PVDF membrane | |
| Chemiluminescent detection | +++ | +++ | +++ |
| Fluorescent detection | ++ | + | +++ |
| Total protein normalization | ++ | + | +++ |
| Requires activation? | No | Yes | Yes |
| Binding mechanism | Nitrogen dipole, H-bond, ionic, and hydrophobic interactions | Hydrophobic interactions | Hydrophobic interactions |
| Durability | Less durable | More durable | More durable |
Scoring key: +++ = best fit, ++ = good fit, + = acceptable fit
Low-fluorescence PVDF membranes are the preferred choice for fluorescent and near-infrared western blotting, minimizing autofluorescence to improve signal-to-noise ratios and detection sensitivity.
Key advantages of low-fluorescence PVDF
Low-fluorescence PVDF membranes are engineered to reduce autofluorescence across commonly used fluorescence channels, enabling more accurate detection and improved quantitative results.
Reduced autofluorescence across detection channels
Invitrogen low fluorescence PVDF membranes exhibit minimal auto-fluorescence across all fluorescent channels and result in reduced background compared to both nitrocellulose and standard PVDF membranes (Figure 1).
Figure 1. Invitrogen low fluorescence PVDF exhibits lower autofluorescence when compared to nitrocellulose and standard PVDF membranes. Unprocessed membranes were hydrated and imaged together in six different fluorescent channels on a compatible digital western blot imaging system. Within each channel, exposure times and contrast settings were consistent for each membrane.
Consistent membrane structure for reliable performance
Invitrogen low fluorescence PVDF membranes feature a 0.3 µm pore size with high consistency and uniformity across the membrane surface (Figure 2), that contributes to consistent protein binding and improved reproducibility across experiments.
Figure 2. Invitrogen low fluorescence PVDF has fibrous structure and a more uniform pore size distribution than many standard PVDF membranes. (A) Top and cross-section views of Invitrogen low fluorescence PVDF membrane (left) and standard PVDF (right) as imaged via scanning electron microscopy. (B) Pore size distribution of Invitrogen low fluorescence PVDF membrane (red) compared with standard PVDF (gray). A capillary porometer was used to evaluate 12 mm diameter circles of each respective membrane for average pore size and frequency.
Exceptional performance at lower wavelength channels
Compared to other commercially available low fluorescence PVDF membranes, Invitrogen low fluorescence PVDF membranes offer excellent performance when imaging in lower wavelength channels (Figure 3). Reduced autofluorescence is especially important at lower wavelengths, where background signal can significantly impact detection sensitivity.
Figure 3. Invitrogen low fluorescence PVDF exhibits lower autofluorescence when compared to other commercially available low fluorescence PVDF membranes. Unprocessed membranes were hydrated and imaged together in six different fluorescent channels on a compatible digital western blot imaging system using exposure times and contrasting consistent with standard western blot applications. Within each channel, contrasts and imaging time were consistent for each membrane. Membranes were hydrated in 100% methanol for 3 minutes and washed in diH2O for 5 minutes on an orbital shaker.
Improved signal-to-noise in multiplex experiments
Reduced background fluorescence enables detection of more protein targets per blot and supports higher-quality, publication-ready data.
Figure 4. Invitrogen low fluorescence PVDF achieves higher signal-to-noise ratio in western blotting compared to other commercially available low fluorescence PVDF membranes. Four lysates were 2-fold serially diluted from 20 µg–2.5 µg, loaded and electrophoresed on Tris-Glycine gels, and transferred to three different membranes. Blocking was completed for 1 hour at room temperature, followed by overnight 4°C incubation with primary antibodies against Hsp90, Calreticulin, beta-Actin, and p23. Secondary antibodies (GAR-800, GAM-647, and GAC-488) in TBST were applied for 1 hour at room temperature. Blots were imaged and contrasted on the iBright FL1500 Imaging System under identical conditions.
Together, these results demonstrate how low-fluorescence PVDF membranes improve sensitivity, reduce background, and enable more reliable fluorescent western blot workflows.
Filter papers support protein transfer by maintaining even contact and consistent buffer flow. Western blotting filter papers are pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins. High-quality western blotting filter papers are available as standard-thickness and extra-thick sheets that are pre-cut for convenience with mini gels and midi gels.
The table below summarizes filter paper thicknesses and formats based on gel size and transfer workflow.
| Thickness | Mini gels | Midi gels | Special formats |
|---|---|---|---|
| 0.83 mm (standard thickness) | Western Blotting Filter Paper, 7 x 8.4 cm | Western Blotting Filter Paper, 8 x 13.5 cm | Western Blotting Filter Papers, 8 x 10.5 cm |
| 2.5 mm (extra thick = high absorbency) |
Tip: Use fresh, fully saturated filter papers to ensure even transfer and prevent overheating
Nitrocellulose and PVDF membranes both produce high-quality western blot results, with nitrocellulose commonly preferred for routine chemiluminescent, colorimetric, and fluorescent detection methods, while PVDF membranes help provide increased durability for chemiluminescent applications that require stripping and reprobing.
Low-fluorescence PVDF membranes are engineered to minimize autofluorescence across fluorescent channels, enabling improved signal-to-noise ratio, longer exposure times, enhanced multiplexing, and more accurate quantitative analysis, making them ideal for fluorescent western blotting and total protein normalization workflows.
Low-fluorescence PVDF membranes perform well for both fluorescent and chemiluminescent detection, offering high protein-binding capacity, durability, and low background fluorescence, making them a versatile option for laboratories that use multiple detection methods.
Membrane pore size generally has modest impact for most proteins, but smaller pore sizes improve retention of low-molecular-weight or low-abundance proteins. Ultimately, the optimal choice may depend on the membrane type and manufacturer, and require optimization based on the target protein.
Western blot membranes can be dried after transfer and stored for later detection, with PVDF requiring reactivation in methanol or ethanol before blocking and antibody incubation steps; drying may strengthen protein binding and support stripping and reprobing, but may increase background in some applications and should be verified.
Nitrocellulose membranes can be striped and reprobed, but they are more fragile and typically support only one or two stripping cycles. For repeated stripping, PVDF membranes are preferred due to their greater mechanical durability.
Access resources to help optimize western blot detection strategies, troubleshoot experiments, and improve data quality.
For Research Use Only. Not for use in diagnostic procedures.