On this page:
Choose fluorescent western blot detection solutions based on multiplexing requirements, quantitative analysis needs, and target protein complexity. Compare fluorescent antibodies, reagents, and detection approaches to support multiplex protein detection, quantitative analysis, and efficient data generation. This page is for laboratories requiring simultaneous detection of multiple protein targets, quantitative analysis, or multiplex western blot workflows.
What is fluorescent western blot detection?
Fluorescent western blot detection is a protein analysis method that uses fluorophore-labeled antibodies to detect and quantify proteins. This method enables multiplexing, broad dynamic range, and stable signal detection, making it excellent for quantitative western blot workflows. Fluorescent western blotting allows researchers to measure multiple protein targets while maintaining high reproducibility and low background.
Fluorescent western blotting works by using fluorophore-labeled antibodies to generate signals when excited by specific wavelengths of light. These signals are captured using a fluorescence imaging system and are directly proportional to protein abundance.
When should you use fluorescent western blotting?
Multiplex western blotting enables simultaneous analysis of multiple protein targets on a single membrane, reducing sample consumption and improving experimental efficiency.
Use fluorescent detection when:
  • You need quantitative western blot data
  • You want to detect multiple proteins simultaneously (multiplexing)
  • You require a wide dynamic range for comparing protein expression levels
  • You want stable signal without time-dependent decay
Consider other methods if:
Fluorescent secondary antibodies
Fluorescent secondary antibodies are a commonly used approach for fluorescent western blot detection and are well suited for multiplex western blotting and quantitative protein analysis.
  • Enables simultaneous detection of multiple protein targets using species-specific fluorescent secondary antibodies labeled with distinct fluorophores
  • Enables signal amplification by binding multiple secondary antibodies to each primary antibody
  • Compatible with multiplex and quantitative fluorescent western blot workflows
Multiplex western blotting enables simultaneous detection and quantitation of multiple protein targets using spectrally distinct fluorescent secondary antibodies. When paired with low-fluorescence PVDF membranes, this approach minimizes background fluorescence while supporting reliable multiplex and quantitative western blot analysis.
Multiplex western blot detection using low Fluorescence PVDF membrane. A431, A549, HEK293, and HepG2 lysates were 2-fold serially diluted from 20 to 2.5 ug, loaded, electrophoresed, and transferred. After blocking with 1X Blocker FL, membrane was incubated with primary antibodies against Hsp90, Calreticulin, beta-Actin, and p23, then Alexa Fluor secondary antibodies goat anti-rabbit 800, goat anti-chicken 546, and goat anti-mouse 488. Blots were imaged and contrasted on the iBright FL1500 Imaging System.
Additional fluorescent detection options
While fluorescent secondary antibodies are the preferred choice for most fluorescent western blot workflows, additional approaches are available for specialized applications.
Conjugated primary antibodies
Well suited for applications requiring maximum specificity or streamlined assay design.
  • Reduces background and cross-reactivity
  • Enables detection without a secondary antibody step
  • Minimizes assay steps and improves reproducibility
Antibody labeling kits
Well suited for custom fluorescent labeling when conjugated antibodies are not commercially available.
  • Enables labeling of primary antibodies with fluorophores
  • Supports custom assay development and flexible experimental design
  • Useful when pre-conjugated antibodies are unavailable
Buffers for fluorescent western blotting
Fluorescent western blot buffers are optimized to reduce background fluorescence, improve signal-to-noise ratio, and support reliable quantitative detection.
Which buffers should you use for fluorescent western blot detection?
  • For sample buffers use dye-free, fluorescent-compatible sample buffers to avoid signal interference
  • For blocking buffers use optimized blocking buffers to minimize nonspecific binding and background fluorescence
  • For stripping buffers use fluorescent stripping buffers to enable reuse of membranes without compromising signal
Below are fluorescent-compatible sample buffers, blocking buffers, and stripping buffers that help ensure reliable results.
Membranes for fluorescent western blotting
Choosing the right membrane is critical for minimizing background fluorescence and maximizing signal detection.
Which membrane should you choose for fluorescent detection?
  • For optimal fluorescent western blot performance, use low-fluorescence PVDF membranes to minimize background fluorescence and maximize signal-to-noise ratio.
  • For multiplex and quantitative fluorescent western blotting, low-fluorescence PVDF membranes are the preferred choice due to their low background and broad fluorophore compatibility.
  • Nitrocellulose membranes may be used for fluorescent western blot workflows, but low-fluorescence PVDF membranes generally help provide the optimal performance for sensitive fluorescence detection.
Below are fluorescent-compatible membranes for western blotting.
Molecular weight markers for fluorescent detection
Imaging and analysis of fluorescent western blotting
Fluorescent western blot imaging enables multiplex detection, accurate quantitation, and reproducible analysis. By detecting multiple protein targets simultaneously, fluorescent imaging can improve workflow efficiency by reducing the need to strip and reprobe membranes.
The imaging system determines which fluorescence channels (e.g., visible fluorescence or near-infrared) are available, guiding antibody and reagent selection. Selecting the right imaging system and analysis approach is important for generating reliable data.
Fluorescent western blot imaging systems
Key highlights:
  • Enables multiplex detection of multiple protein targets on a single blot—no need to strip and reprobe
  • Supports quantitative analysis across multiple fluorescent channels
  • Helps provide reproducible imaging conditions for fluorescent workflows
No-Stain protein quantitation reagent in fluorescent western blotting
Key highlights:
  • Fast, easy-to-use reagent helps provide sensitive, linear detection
  • Total protein normalization is a more reliable method than housekeeping proteins
  • Important for correcting sample loading variability
Reprobing fluorescent western blots
Key highlights:
  • Enables efficient reuse of PVDF membranes for additional protein detection
  • Allows the same blot to be reprobed with different primary antibodies
  • Excellent when sample quantity is limited or additional target analysis is required
Chemiluminescent vs. fluorescent detection
Videos for fluorescent western blot detection
How do I perform multiplex fluorescent western blotting?
Fluorescent western blotting enables multiplex detection and quantitative protein analysis. In this webinar, you'll learn western blot workflow fundamentals, compare fluorescent and chemiluminescent detection methods, and explore practical tips for improving fluorescent western blot performance.
You'll learn how to:
  • Select gels, buffers, and protein ladders for protein separation
  • Choose transfer systems and membranes
  • Select fluorescent antibodies and fluorophores
  • Capture and analyze fluorescent western blot data
  • Improve fluorescent western blot workflow performance
Ordering information
Catalog # Name Size Price Qty
LC2570 Each
96.25
37565 Each
175.00
37542 Each
242.65
Online exclusive
Save 43.35
37578 Each
52.65
Online exclusive
Save 9.85
37538 Each
242.65
Online exclusive
Save 42.35
37579 Each
52.65
Online exclusive
Save 9.85
37538X3 3 x 1 L
548.65
Online exclusive
Save 97.35
37539 Each
259.65
Online exclusive
Save 45.35
37543 Each
257.65
Online exclusive
Save 45.35
37527 Each
250.65
Online exclusive
Save 28.35
37527X3 Each
599.65
Online exclusive
Save 66.35
62300 Each
198.65
Online exclusive
Save 17.35
62299 Each
58.65
Online exclusive
Save 4.85
62300X4 Each
597.65
Online exclusive
Save 52.35
84788 Each
55.75
Catalog # Name Size Price Qty
678001 Each
322.00
88018 Each
505.65
Online exclusive
Save 50.35
88025 Each
160.65
Online exclusive
Save 16.35
88014 Each
174.65
Online exclusive
Save 17.35
77010 Each
383.65
Online exclusive
Save 38.35
88024 Each
157.65
Online exclusive
Save 19.35
88013 Each
169.65
Online exclusive
Save 21.35
77012 Each
381.65
Online exclusive
Save 47.35
Continue the western blot workflow 
Explore additional solutions that support each step of the western blot workflow from protein detection to data analysis:
Western Blot Buffers and Consumables
Choose buffers, membranes, and consumables to reduce background and improve signal.
Antibodies for Western Blotting
Choose primary and secondary antibodies optimized for western blotting to support specific target detection.
Total Protein Normalization
Improve quantitative western blot accuracy by correcting for sample loading and transfer variability using total protein normalization.
Protein Gel Electrophoresis and Western Blot Imaging Systems
Explore gel documentation systems and blot imagers.
Frequently asked questions
Fluorescent western blot detection uses fluorophore-labeled antibodies to detect and quantify proteins. The method supports multiplex western blotting, broad dynamic range, and quantitative analysis, making it well suited for measuring multiple protein targets simultaneously.
Multiplex western blotting enables simultaneous detection and quantitation of multiple protein targets in a single experiment using spectrally distinct fluorophores. This approach helps conserve sample and improve workflow efficiency.
Fluorescent and chemiluminescent detection each offer distinct advantages. Fluorescent detection is preferred for multiplex western blotting and quantitative analysis of multiple protein targets, while chemiluminescent detection is well suited for applications requiring maximum sensitivity and detection of very low-abundance proteins.
Low-fluorescence PVDF membranes help minimize background fluorescence and maximize the signal-to-noise ratio, making them the preferred membrane choice for multiplex and quantitative fluorescent western blot workflows.
Near-infrared (NIR) western blot detection supports multiplex western blot workflows that use fluorophores which emit in the 700–800 nm range. NIR detection can reduce background fluorescence and improve signal-to-noise ratios.
Western blot educational resources
Access resources to help optimize western blot detection strategies, troubleshoot experiments, and improve data quality.
For Research Use Only. Not for use in diagnostic procedures.