How to transition to the Novex Wedgewell-Format Tris-Glycine gels.
You can expect the same high quality results with the latest Invitrogen Novex WedgeWell-Format Tris-Glycine gels as you get with your existing Invitrogen Novex Tris-Glycine gels. In addition, you can now enjoy the added benefits of longer shelf life and the wedge well convenience allowing you to load up to two times the sample volume compared to the traditional Tris-glycine gels. In the following comparisons, you can see equivalent performance of Novex WedgeWell-Format Tris-Glycine gels and the Novex Tris-Glycine gels.
 Novex Tris-Glycine gels, WedgeWell formatExisting Novex Tris-Glycine gels
Protein resolution, band qualityComparable
Average run time~60 min~90 min
Well formatHigh capacity wedge design, up to 37 µLStandard capacity
Gel dimensionsMini: 8 x 8 cm, 1 mm thickMini: 8 x 8 cm, 1 or 1.5 mm thick;
Midi: 8 x 13 cm, 1 mm thick
Shelf life Up to 12 months1–2 months
Pack sizes2 & 10 packs10 packs
Use our selection guide to find the equivalent Novex WedgeWell Tris-Glycine gel catalog number that replaces your existing gel or consult the table, below.
Equal performance with Novex WedgeWell Tris-Glycine gels
Equal performance with Novex WedgeWell Tris-Glycine gels. Protein ladders, purified proteins, and E. coli lysate were loaded on a 4–20% gradient WedgeWell Tris-Glycine gel (A) and a 4–20% gradient Novex Tris-Glycine gel (B). Similar band migration and similarly sharp and straight bands are observed on gels (A) and (B).  Lanes 1, 5, 10: 5 µL Thermo Scientific™ PageRuler™ Unstained Protein Ladder; lanes 2, 6, 9: 5 µL Invitrogen™ Novex™ Mark12™ Unstained Standard; lane 3: 10 µg E. coli lysate (10 µL sample volume); lane 4: 6 µg BSA (10 µL sample volume); lane 7: 6 µg hIgG (10 µL sample volume); lane 8: 20 µg E. coli lysate (20 µL sample volume).
Migration chart comparison of WedgeWell vs. original Novex Tris-Glycine gels. View migration data side by side for the Invitrogen Mark12 Unstained Standard run on Novex WedgeWell Tris-Glycine gels and Novex Tris-Glycine gels.
No records were found matching your criteria
Discontinued Cat. No.Discontinued product descriptionReplacement Cat. No.Replacement product description
EC6055BOXNovex 4% Tris-Glycine, 1.0mm, 10-wellXP00060BOXNovex WedgeWell 6% Tris-Glycine, 10-well
EC60552BOXNovex 4% Tris-Glycine, 1.0mm, 12-wellXP00062BOXNovex WedgeWell 6% Tris-Glycine, 12-well
EC6058BOXNovex 4% Tris-Glycine, 1.5mm, 10-wellXP00060BOXNovex WedgeWell 6% Tris-Glycine, 10-well
EC60585BOXNovex 4% Tris-Glycine, 1.5mm, 15-wellXP00065BOXNovex WedgeWell 6% Tris-Glycine, 15-well
EC6065BOXNovex 6% Tris-Glycine, 1.0mm, 10-wellXP00060BOXNovex WedgeWell 6% Tris-Glycine, 10-well
EC60652BOXNovex 6% Tris-Glycine, 1.0mm, 12-wellXP00062BOXNovex WedgeWell 6% Tris-Glycine, 12-well
EC60655BOXNovex 6% Tris-Glycine, 1.0mm, 15-wellXP00065BOXNovex WedgeWell 6% Tris-Glycine, 15-well
EC6068BOXNovex 6% Tris-Glycine, 1.5mm, 10-wellXP00060BOXNovex WedgeWell 6% Tris-Glycine, 10-well
EC60685BOXNovex 6% Tris-Glycine, 1.5mm, 15-wellXP00065BOXNovex WedgeWell 6% Tris-Glycine, 15-well
EC6015BOXNovex 8% Tris-Glycine, 1.0mm, 10-wellXP00080BOXNovex WedgeWell 8% Tris-Glycine, 10-well
EC60152BOXNovex 8% Tris-Glycine, 1.0mm, 12-wellXP00082BOXNovex WedgeWell 8% Tris-Glycine, 12-well
EC60155BOXNovex 8% Tris-Glycine, 1.0mm, 15-wellXP00085BOXNovex WedgeWell 8% Tris-Glycine, 15-well
EC6018BOXNovex 8% Tris-Glycine, 1.5mm, 10-wellXP00080BOXNovex WedgeWell 8% Tris-Glycine, 10-well
EC60185BOXNovex 8% Tris-Glycine, 1.5mm, 15-wellXP00085BOXNovex WedgeWell 8% Tris-Glycine, 15-well
EC6075BOXNovex 10% Tris-Glycine, 1.0mm, 10-wellXP00100BOXNovex WedgeWell 10% Tris-Glycine, 10-well
EC6075BX30Novex 10% Tris-Glycine, 1.0mm, 10-wellXP00100BOXNovex WedgeWell 10% Tris-Glycine, 10-well
EC60752BOXNovex 10% Tris-Glycine, 1.0mm, 12-wellXP00102BOXNovex WedgeWell 10% Tris-Glycine, 12-well
EC60755BOXNovex 10% Tris-Glycine, 1.0mm, 15-wellXP00105BOXNovex WedgeWell 10% Tris-Glycine, 15-well
EC6078BOXNovex 10% Tris-Glycine, 1.5mm, 10-wellXP00100BOXNovex WedgeWell 10% Tris-Glycine, 10-well
EC60785BOXNovex 10% Tris-Glycine, 1.5mm, 15-wellXP00105BOXNovex WedgeWell 10% Tris-Glycine, 15-well
EC6005BOXNovex 12% Tris-Glycine, 1.0mm, 10-wellXP00120BOXNovex WedgeWell 12% Tris-Glycine, 10-well
EC60052BOXNovex 12% Tris-Glycine, 1.0mm, 12-wellXP00122BOXNovex WedgeWell 12% Tris-Glycine, 12-well
EC60055BOXNovex 12% Tris-Glycine, 1.0mm, 15-wellXP00125BOXNovex WedgeWell 12% Tris-Glycine, 15-well
EC6006BOXNovex 12% Tris-Glycine, 1.0mm, 2D-wellEC6026BOXNovex 4-20% Tris-Glycine, 1.0mm, 2D-well
EC6008BOXNovex 12% Tris-Glycine, 1.5mm, 10-wellXP00120BOXNovex WedgeWell 12% Tris-Glycine, 10-well
EC60085BOXNovex 12% Tris-Glycine, 1.5mm, 15-wellXP00125BOXNovex WedgeWell 12% Tris-Glycine, 15-well
EC6485BOXNovex 14% Tris-Glycine, 1.0mm, 10-wellXP00140BOXNovex WedgeWell 14% Tris-Glycine, 10-well
EC64852BOXNovex 14% Tris-Glycine, 1.0mm, 12-wellXP00142BOXNovex WedgeWell 14% Tris-Glycine, 12-well
EC64855BOXNovex 14% Tris-Glycine, 1.0mm, 15-wellXP00145BOXNovex WedgeWell 14% Tris-Glycine, 15-well
EC6488BOXNovex 14% Tris-Glycine, 1.5mm, 10-wellXP00140BOXNovex WedgeWell 14% Tris-Glycine, 10-well
EC64885BOXNovex 14% Tris-Glycine, 1.5mm, 15-wellXP00145BOXNovex WedgeWell 14% Tris-Glycine, 15-well
EC6495BOXNovex 16% Tris-Glycine, 1.0mm, 10-wellXP00160BOXNovex WedgeWell 16% Tris-Glycine, 10-well
EC64952BOXNovex 16% Tris-Glycine, 1.0mm, 12-wellXP00162BOXNovex WedgeWell 16% Tris-Glycine, 12-well
EC64955BOXNovex 16% Tris-Glycine, 1.0mm, 15-wellXP00165BOXNovex WedgeWell 16% Tris-Glycine, 15-well
EC6498BOXNovex 16% Tris-Glycine, 1.5mm, 10-wellXP00160BOXNovex WedgeWell 16% Tris-Glycine, 10-well
EC64985BOXNovex 16% Tris-Glycine, 1.5mm, 15-wellXP00165BOXNovex WedgeWell 16% Tris-Glycine, 15-well
EC6505BOXNovex 18% Tris-Glycine, 1.0mm, 10-wellXP00160BOXNovex WedgeWell 16% Tris-Glycine, 10-well
EC65052BOXNovex 18% Tris-Glycine, 1.0mm, 12-wellXP00162BOXNovex WedgeWell 16% Tris-Glycine, 12-well
EC65055BOXNovex 18% Tris-Glycine, 1.0mm, 15-wellXP00165BOXNovex WedgeWell 16% Tris-Glycine, 15-well
EC6508BOXNovex 18% Tris-Glycine, 1.5mm, 10-wellXP00160BOXNovex WedgeWell 16% Tris-Glycine, 10-well
EC65085BOXNovex 18% Tris-Glycine, 1.5mm, 15-wellXP00165BOXNovex WedgeWell 16% Tris-Glycine, 15-well
EC6135BOXNovex 10-20% Tris-Glycine, 1.0mm, 10-wellXP10200BOXNovex WedgeWell 10-20% Tris-Glycine, 10-well
EC61352BOXNovex 10-20% Tris-Glycine, 1.0mm, 12-wellXP10202BOXNovex WedgeWell 10-20% Tris-Glycine, 12-well
EC61355BOXNovex 10-20% Tris-Glycine, 1.0mm, 15-wellXP10205BOXNovex WedgeWell 10-20% Tris-Glycine, 15-well
EC61385BOXNovex 10-20% Tris-Glycine, 1.5mm, 15-wellXP10205BOXNovex WedgeWell 10-20% Tris-Glycine, 15-well
EC6035BOXNovex 4-12% Tris-Glycine, 1.0mm, 10-wellXP04120BOXNovex WedgeWell 4-12% Tris-Glycine, 10-well
EC60352BOXNovex 4-12% Tris-Glycine, 1.0mm, 12-wellXP04122BOXNovex WedgeWell 4-12% Tris-Glycine, 12-well
EC60355BOXNovex 4-12% Tris-Glycine, 1.0mm, 15-wellXP04125BOXNovex WedgeWell 4-12% Tris-Glycine, 15-well
EC6036BOXNovex 4-12% Tris-Glycine, 1.0mm, 2D-wellEC6026BOXNovex 4-20% Tris-Glycine, 1.0mm, 2D-well
EC6038BOXNovex 4-12% Tris-Glycine, 1.5mm, 10-wellXP04120BOXNovex WedgeWell 4-12% Tris-Glycine, 10-well
EC60385BOXNovex 4-12% Tris-Glycine, 1.5mm, 15-wellXP04125BOXNovex WedgeWell 4-12% Tris-Glycine, 15-well
EC60249BOXNovex 4-20% Tris-Glycine, 1.0mm, 9-wellXP04200BOXNovex WedgeWell 4-20% Tris-Glycine, 10-well
EC6025BOXNovex 4-20% Tris-Glycine, 1.0mm, 10-wellXP04200BOXNovex WedgeWell 4-20% Tris-Glycine, 10-well
EC60252BOXNovex 4-20% Tris-Glycine, 1.0mm, 12-wellXP04202BOXNovex WedgeWell 4-20% Tris-Glycine, 12-well
EC60255BOXNovex 4-20% Tris-Glycine, 1.0mm, 15-wellXP04205BOXNovex WedgeWell 4-20% Tris-Glycine, 15-well
EC6028BOXNovex 4-20% Tris-Glycine, 1.5mm, 10-wellXP04200BOXNovex WedgeWell 4-20% Tris-Glycine, 10-well
EC60285BOXNovex 4-20% Tris-Glycine, 1.5mm, 15-wellXP04205BOXNovex WedgeWell 4-20% Tris-Glycine, 15-well
EC6045BOXNovex 8-16% Tris-Glycine, 1.0mm, 10-wellXP08160BOXNovex WedgeWell 8-16% Tris-Glycine, 10-well
EC60452BOXNovex 8-16% Tris-Glycine, 1.0mm, 12-wellXP08162BOXNovex WedgeWell 8-16% Tris-Glycine, 12-well
EC60455BOXNovex 8-16% Tris-Glycine, 1.0mm, 15-wellXP08160BOXNovex WedgeWell 8-16% Tris-Glycine, 15-well
EC6048BOXNovex 8-16% Tris-Glycine, 1.5mm, 10-wellXP08160BOXNovex WedgeWell 8-16% Tris-Glycine, 10-well
EC60485BOXNovex 8-16% Tris-Glycine, 1.5mm, 15-wellXP08165BOXNovex WedgeWell 8-16% Tris-Glycine, 15-well
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.