ZOOM IEF fractionator
The ZOOM IEF Fractionator offers a fast, reliable method to reduce sample complexity, enrich low abundance proteins, and increase the dynamic range of detection. Solution phase isoelectric focusing with the ZOOM IEF Fractionator provides reproducible separations in three hours. Fractionated samples are ready for further analysis by two dimensional gel electrophoresis (2DE), one dimensional gel electrophoresis (1DE), or two dimensional liquid chromatography/mass spectrometry (2D LC/MS).
View all products
The ZOOM IEF Fractionator combo kit
Components of this kit include:
The ZOOM IEF Fractionator—built for safety, reliability, and durability. It includes easy-to-assemble sample chambers, in-built buffer chambers, buffer trough, removable electrode assembly, and safety lid.
ZOOM Disks—precast polyacrylamide gels that eliminate the need for manual preparation and minimize any chance of cross contamination. These immobilized buffered disks are pre-labeled, disposable, and designed for single use, ensuring consistent and reproducible fractionation. They resolve samples into five equal fractions (using six disks of specific pH) in the pH range from 3–10.
ZOOM Reagents—proteomic grade reagents, including urea, thiourea, CHAPS, and ampholytes, that ensure optimal conditions for sample preparation.
Contents and storage:
The ZOOM Fractionator combo kit includes the ZOOM IEF Fractionator; one pack ZOOM Disks of each pH; and all ZOOM Reagents (urea, thiourea, CHAPS, carrier ampholytes pH 3–10, anode buffers, and cathode buffers). ZOOM Disks, Carrier Ampholytes, and Buffers are stored at 4°C.
Downstream processing by 2D gel electrophoresis
Following fractionation, the separated samples can be subjected to further analysis using narrow or broad range ZOOM Strips on the ZOOM IPGRunner System. Follow with second dimension analysis using neutral pH NuPAGE ZOOM Gels.
Technical resources
Manual
Quick Reference Card
References
  1. Zuo, X. and Speicher, D.W. (2000) Anal. Biochem. 284: 266-278.
  2. Zuo, et. al. (2001) Electrophoresis 22: 1603-1615.
  3. Zuo, X. and Speicher, D.W. (2002) Proteomics: 2: 58-68.
  4. Zuo, X., Hembach, P., Echan, L., and Speicher, D.W. (2002) Journal of Chromatography B 782: 253-265.
  5. Ali-Khan, N., Zuo, X., and Speicher, D.W. (2002) Current Protocols in Protein Science 22.1: 1-19.
  6. Zuo, X. and Speicher, D.W. (2000) Methods in Molecular Biology (Humana Press, P. Cutler, ed.) In press.
  7. Zuo, X. and Speicher D.W. 2004. Microscale solution isoelectrofocusing: A sample prefractionation method for comprehensive proteome analysis. In: Methods in Molecular Biology: Protein Purification Protocols. Humana Press, Totowa, NJ (P. Cutler, ed.), pp.361-375.
  8. Speicher, D.W. 2004. Overview of proteome analysis. In: Proteome Analysis: Interpreting the Genome (D. Speicher, ed.). Elsevier, The Netherlands, pp. 1-18.
  9. Zuo, X., Lee, K., Speicher, D.W. 2004. Electrophoretic prefractionation for comprehensive analysis of proteomes. In: Proteome Analysis: Interpreting the Genome (D. Speicher, ed.). Elsevier, The Netherlands, pp. 93-118.
  10. Speicher, D.W., Lee, K., Tang, H.-Y., Echan, L., Ali-Khan, N., Zuo, X., and Hembach, P. 2004. Current challenges in proteomics: Mining low abundance proteins and expanding protein profiling capacities. In: Advances in Mass Spectrometry, Vol. 16 (Ashcroft, A.E., Brenton, G, and Monaghan, J.J., ed.) pp. 37-57.
  11. Zuo, X., Lee, K., and Speicher, D.W. 2004. Fractionation of complex proteomes by microscale solution isoelectrofocusing using ZOOM IEF fractionators to improve protein profiling. In: The Proteome Handbook (Walker, J.M., ed.) In press.
The ZOOM IEF Fractionator was in part developed pursuant to a license from The Wistar Institute of Anatomy and Biology, located in Philadelphia, Pennsylvania. Established in 1892, The Wistar Institute was the first independent medical research facility in the United States.
Resources
Learning resources
Support resources