Improve your bioanalysis workflow
Overcome the challenges associated with traditional affinity purification methods using Thermo Scientific™ MSIA™ (Mass Spectrometric Immunoassay) microcolumns. Designed for analytical affinity purification, ready-to-use MSIA microcolumns are coated with target-specific affinity ligand to capture low abundant target analytes from even the most complex biological samples. Secure resourceful innovation that enables rapid assay development, provides high specificity and reduces variability to have data that you can trust.
Bioanalysis Innovation
tailored to you
Ready-to-use
affinity ligand coated microcolumns
Capture
analytes with speed, precision & confidence 
Elute
reliable results for
increased data confidence 
MSIA microcolumns can be used on both automated high throughput and semi-automated low throughput formats, enabling simple, precise affinity capture that fits your biomolecule analysis workflow.
MSIA microcolumns are available for:
Housed within pipette tips, MSIA microcolumns are coated with target specific affinity ligand providing highly specific affinity capture. 
MSIA microcolumns capture even the low abundant analytes with superior quantitation performance, down to the femtomole level.
Cleaner analyte of interest enables low background, high senstivity and enhanced reproducibility.
Empower your biomolecule analysis workflow with MSIA microcolumns - providing data you can trust.
MSIA vs. Traditional Methods
Select the best MSIA product for your application
MSIA D.A.R.T.’S, Streptavidin
MSIA
Streptavidin-
EVO
MSIA D.A.R.T.’S, Insulin 
*MSIA
D.A.R.T.’S, Custom
*MSIA D.A.R.T.’S,
Protein A, Protein G & Protein A/G 
Description
Based on the Streptavidin-biotin chemistry.
Enable a broad array of MSIA applications.
Based on the Streptavidin-biotin chemistry.
Enable a broad array of MSIA applications.
For purification of various insulin analogs, including endogenous human insulin, therapeutic analogs and non-human animal analogs. Allows direct 
linking of **affinity ligand of your choice to MSIA microcolumn.
A versatile option to bind and orientate a wide range of antibodies, enabling flexibility to use antibodies of choice specific to the application .
Low
Throughput
Finnpipette
Novus i
Multichannel Electronic Pipette
- - - -  Finnpipette
Novus i Multichannel Electronic Pipette
Finnpipette
Novus i Multichannel Electronic Pipette
Finnpipette
Novus i Multichannel Electronic Pipette
High
Throughput
Thermo Scientific
Versette
Automated Liquid Handler 
Tecan’s Freedom EVO Platform
with MCA96 liquid handling arm
Thermo Scientific Versette Automated Liquid Handler Thermo Scientific Versette
Automated Liquid Handler
Thermo Scientific Versette Automated Liquid Handler 
Binding 
Capacity
Immobilized Stretavidin: 4.0 µg/microcolumn (<15%)
Bindingcapacity:
4 µg/microcolumn (<15%) [based on biotinylated-IgG]
Immobilized Stretavidin: 10 µg/microcolumn (<15%)
Binding capacity:
7.5 µg/microcolumn (<15%) [based on biotinylated-IgG]
N/A N/A Protein A: 4 µg IgG/microcolumn
Protein G: 4 µg IgG/microcolumn
Protein A/G: 4 µg IgG/microcolumn
Application
For microscale purification of
target protein, peptide or antibody using target specific biotin conjugated affinity ligands. 
For microscale purification of target protein, peptide or antibody using target specificbiotin conjugated affinity ligands. Simultaneous quantitative and qualitative bioanalysis of insulin including endogenous and therapeutic
analogs.
Tailored to capture
& elute protein, peptide or antibody of your interest.
Allows use of untagged
antibodies for
your specific applications.
Relevant
Industries
Biopharma
CRO
Sports anti-doping
Translational research
Biopharma
CRO
Sports anti-doping
Translational research
Biopharma
CRO
Forensic
Sports anti-doping
Biopharma
Clinical/Translational
CRO
Sports anti-doping
Biopharma
CRO
Sports anti-doping
Translational research
*Direct linking of affinity ligand for the target analyte to the microcolumn.
**Affinity ligand must meet specific selection criteria. Either provide the affinity ligand or request us to directly buy from the vendor.
MSIA products
MSIA™ D.A.R.T.'S™ for the Versette and Novus i Platform
Enabled through proprietary MSIA D.A.R.T.'S™ technologies, these microcolumns have demonstrated pg/mL level detection while providing simultaneous qualitative/quantitative data content from complex biological matrix, such as human plasma.
MSIA™ Microcolumns for the Tecan Freedom EVO Platform
MSIA affinity micro columns, designed for Freedom EVO platform (MCA 96 head), provide innovative enablement in the areas ranging from bioanalysis of therapeutic mAbs, antibody-drug conjugates (ADC), biotransformation [in-vivo modifications, drug-antibody ratio (DAR)], preclinical and clinical drug trials (PKPD, DMPK, and ADME) to biomarker and proteins analysis. Learn more
Finnpipette™ Novus i Multichannel Electronic Pipette
Combine Thermo Scientific™ Finnpipette™ Novus i Multichannel Electronic pipettes with our adjustable pipette stand for an economical semi-automated system for rapid development and application of Mass Spectrometric Immunoassay (MSIA) methods. Each Novus i comes with a custom MSIA menu for further simplification of pipette programming that is complementary to MSIA applications.
Equipment and instruments related to MSIA microcolumns
Versette™ Pipetting Workstation
Support a wide range of applications with the Thermo Scientific™ Versette™ Automated Liquid Handler, a compact handler featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads.
UltiMate™ 3000 RSLCnano  System
The Thermo Scientific™ UltiMate™ 3000 RSLCnano LC system offers better confidence: confidence in performance, confidence in results and confidence in ease of operation. Our 25-year experience in nano LC has resulted in a system that provides better results than ever before, setting a new standard in nano LC applications for Proteomics.
Q Exactive™ Hybrid Quadrupole-Orbitrap Mass Spectrometer
The Q Exactive Mass Spectrometer is equally useful for untargeted or targeted screening and a broad range of qualitative and quantitative applications in drug discovery, proteomics, environmental and food safety, clinical research and forensic toxicology. Learn more
Q Exactive™ Plus Hybrid Quadrupole-Orbitrap™ Mass Spectrometer
The Q Exactive™ Plus Hybrid Quadrupole-Orbitrap™ Mass Spectrometer takes Quanfirmation to the next level with increased performance and new options that enhance applications from confident DMPK qual/quan screening studies to characterization of intact monoclonal antibodies. Learn more
For more resources visit the MSIA Resource Library ›