Search
We offer strong cation exchange (SCX) and strong anion exchange (SAX) resins and membranes. Our ion exchange (IEX) resins are rigid polymeric beads with covalent surface chemistries, which enable easier handling and packing while offering superior physical and chemical stability—resulting in a robust manufacturing process.
POROS™ XS Strong Cation Exchange Resin and the POROS™ XQ Strong Anion Exchange Resin combine high binding capacity with a salt tolerance of up to 150 mM NaCl. The POROS™ HQ 50 µm Strong Anion Exchange Resin contains a quaternized polyethyleneimine functional group, yielding a high capacity, and is composed of a Perfusion Chromatography™ media designed for the separation and purification of biomolecules. These resins deliver unprecedented separation capability.
Pierce™ strong cation and anion exchange spin columns are membrane-based spin formats that eliminate the need for column packing, allow multiple samples to be processed simultaneously, and are ideal for working with low-volume buffer solutions.
Product links in the last row of each column
| Pierce (SCX) Spin Columns | POROS XS Resin | Pierce (SAX) Spin Columns | POROS XQ Resin | POROS 50 HQ Resin | |
| Surface charge | Strong cation exchange (SCX) | Strong cation exchange (SCX) | Strong anion exchange (SAX) | Strong anion exchange (SAX) | Strong anion exchange (SAX) |
| Format | membrane | resin | membrane | resin | resin |
| Binding capacity | 4 mg (Mini) 60–80 mg (Maxi) | >100 mg/mL (dynamic) | N/A | >140 mg/mL (dynamic) | 60–70 mg/mL (dynamic) |
| Salt tolerance | ≤25 mM | 150 mM | ≤25 mM | ≤50 mM | ≤150 mM |
| Recommended processing volumes | 0.5 mL (Mini) 20 mL (Maxi) | 10 mL–1000 L | 0.5 mL (Mini) 20 mL (Maxi) | 10 mL–1000 L | 10 mL–1000 L |
| Packaging options | 0.5 mL spin columns (Mini) 20 mL spin columns (Maxi) | loose resin | 0.5 mL spin columns (Mini) 20 mL spin columns (Maxi) | loose resin | loose resin |
| Order product(s) | 90008 (0.5 mL) 90009 (20 mL) | 82071 | 90010 (0.5 mL) 90011 (20 mL) | 82073 | 82077 |
When you need high performance and high throughput on a large scale, POROS bulk chromatography resins are the industry standard for process-scale bioseparations.
Browse all options for affinity purification of proteins and biomolecules, including activated resins for immobilization of ligands and columns for packing affinity resins.
Quickly remove salts and small molecules from protein solutions using Thermo Scientific™ Zeba™ Spin Columns, plates, and cartridges.
Browse reagents, kits, and devices that enable robust and reproducible protein extraction, enrichment, clean-up, digestion, and peptide enrichment and clean-up for proteomics mass spectrometry analysis.
For Research Use Only. Not for use in diagnostic procedures.