The Validated Stealth RNAi™ siRNA allow you to quickly and ffectively knock down gene expression using validated Stealth RNAi siRNA duplexes. Each Validated Stealth RNAi siRNA duplex has been functionally tested for knockdown in mammalian cells. This ensures that each duplex is highly active.
Functionally Validated Stealth RNAi siRNA
  • Bench-tested molecules--100% guaranteed to effectively knock down expression of your gene by greater than 70%*
  • Stealth RNAi siRNA technology—chemically-modified double-stranded RNA for the most effective knockdown of gene expression
  • Up to two non-overlapping Stealth RNAi siRNA molecules per gene--controls are compatible with the latest experimental guidelines for RNAi research
  • Full sequence information following purchase—simple online access using lot number information
Functional validation ensures results
With Validated Stealth RNAi siRNA, the work of designing and testing RNAi molecules has been done for you, saving time and effort. The development of these molecules begins by using the state-of-the-art BLOCK-iT™ RNAi Designer to design several Stealth RNAi siRNA duplexes that target a gene. These duplexes are then transfected into cells using Lipofectamine™ 2000 or RNAiMAX Transfection reagents to test their ability to knock down the targeted gene. Duplexes must knock down expression by at least 80% to be considered.
Ordering information
To order, go to the BLOCK-iT™ RNAi Express search engine.
Find and order the Validated Stealth RNAi™ siRNA for your genes of interest.  If a Validated Stealth RNAi siRNA is not available, you can also use this search engine to find a list of ready-to-order, predesigned Stealth™ Select RNAi target sequences to the majority of the human, mouse and rat transcriptomes.
More Information
Literature
Web Seminars and Guides
Custom Services
Need help with your RNAi research? Contact us for a consultation and quote today.
Questions
Send your questions to our top researchers at RNAiresearcher
@thermofisher.com