Heat Seals
Heat-sealed microplates are ideal for storage applications. Advantages over other plate sealing methods include time and cost savings and sealing integrity. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal. Heat sealing offers the capability of retrieving samples and then re-sealing the plate.
Thermo Scientific Heat Seals are:
  • Fast
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the ALPS 3000, ALPS 25 and ALPS 50 V
Single Sheet Heat Seals
Designed for reliable performance with Thermo Scientific ALPS 30 and ALPS 50 V manual heat sealers.
SKU Brand Color Pierce / Peel Properties Multiple seals Supported Plates
AB0559 ThermoSeal Silver NA Very strong seal, chemical resistant (DMSO compatible) No PP
AB0685 ClearSeal Clear NA Strong seal, optically transparent No PP
AB0745 Easy Peel Silver Peelable Long term storage, up to -200°F No PP, PE. COC
AB0757 Easy Pierce Silver Pierceable Chemical resistant (DMSO compatible) Yes PP, PE
AB0812 DiamondSeal Clear NA Optically transparent No PP, PS, PE, COC
AB1720 Easy Pierce 20μm Silver Pierceable Thin foil Yes PP, PE
AB1797 ClearSeal Clear Pierceable Thin, non-conductive No PP, PS, PE
Heat Seal Rolls
Designed for reliable performance with Thermo Scientific ALPS 3000 automated heat sealer.
SKU Brand Color Pierce / Peel Properties Multiple seals Supported Plates
AB3559 ThermoSeal Silver NA Very strong seal, chemical resistant (DMSO compatible) No PP
AB3686 ClearSeal Clear NA Strong seal, optically transparent No PP
AB3720 Easy Pierce 20μm Silver Pierceable Thin foil Yes PP, PE
AB3738 Easy Pierce Silver Pierceable Chemical resistant (DMSO compatible) Yes PP, PE
AB3739 Easy Peel Silver Peelable Long term storage, up to -200°F No PP, PE. COC
AB3797 ClearSeal Clear Pierceable Thin, non-conductive No PP, PS, PE
AB3799 DiamondSeal Clear Peelable Optically transparent No PP, PS, PE, COC
Single Sheet Heat Seals
Designed for reliable performance with Thermo Scientific ALPS 30 and ALPS 50 V manual heat sealers.
SKU Brand Color Pierce / Peel Properties Multiple seals Supported Plates
AB0559 ThermoSeal Silver NA Very strong seal, chemical resistant (DMSO compatible) No PP
AB0685 ClearSeal Clear NA Strong seal, optically transparent No PP
AB0745 Easy Peel Silver Peelable Long term storage, up to -200°F No PP, PE. COC
AB0757 Easy Pierce Silver Pierceable Chemical resistant (DMSO compatible) Yes PP, PE
AB0812 DiamondSeal Clear NA Optically transparent No PP, PS, PE, COC
AB1720 Easy Pierce 20μm Silver Pierceable Thin foil Yes PP, PE
AB1797 ClearSeal Clear Pierceable Thin, non-conductive No PP, PS, PE
Heat Seal Rolls
Designed for reliable performance with Thermo Scientific ALPS 3000 automated heat sealer.
SKU Brand Color Pierce / Peel Properties Multiple seals Supported Plates
AB3559 ThermoSeal Silver NA Very strong seal, chemical resistant (DMSO compatible) No PP
AB3686 ClearSeal Clear NA Strong seal, optically transparent No PP
AB3720 Easy Pierce 20μm Silver Pierceable Thin foil Yes PP, PE
AB3738 Easy Pierce Silver Pierceable Chemical resistant (DMSO compatible) Yes PP, PE
AB3739 Easy Peel Silver Peelable Long term storage, up to -200°F No PP, PE. COC
AB3797 ClearSeal Clear Pierceable Thin, non-conductive No PP, PS, PE
AB3799 DiamondSeal Clear Peelable Optically transparent No PP, PS, PE, COC
For fully automated sealing using heat seal rolls, select the ALPs 3000
We have plate sealers too!
For manual or semi-automated sealing using individual plate seals, choose between the ALPs 50V or the ALPs 30.
Learn more 
For fully automated sealing using heat seal rolls, select the ALPs  3000.