Storage Plate Cap Mat
96-well Cap Mats for Nunc and Abgene polypropylene plates designed to protect well contents from evaporation, outside contamination and cross-well contamination
  • Chemically resistant and pierceable; withstand low temperatures to -80°C
  • Pre-slit silicone well caps permit a pipette tip or probe to enter the well without damage to the cap
  • Colored options to allow for quick identification of storage plates
  • Silicone caps with pre-slit are solvent resistant; should not be subjected to 100% DMSO
Additionally, we offer Abgene Individual Storage Plate Caps and
Abgene Storage Plate Strip Caps
Notes: Cap mats fit both 96-well Polypropylene DeepWell and Standard Height Shared Wall Plates. Cat. No. 276011 preslit silicone caps can be pierced repeatedly.
Available 96-well Cap Mats
No records were found matching your criteria
Cat. No Under well shape Pierceable Color Sterile Brand
276000 Round Pierceable Natural Sterile Nunc
276002 Round Pierceable Natural No Nunc
276003 Round Pierceable Yellow No Nunc
276004 Round Pierceable Red No Nunc
276005 Round Pierceable Blue No Nunc
276011* Round Pierceable (preslit) Natural No Nunc
4410 Round No Natural No Nunc
4411-11 Round No Natural No Nunc
4412-11 Round No Natural Sterile Nunc
4413-11 Round No Natural No Nunc
4414-11 Round No Natural Sterile Nunc
4415 Round No Natural No Nunc
4416 Round No Natural Sterile Nunc
4420 Square No Natural No Nunc
4421 Square No Natural No Nunc
4422 Square No Natural Sterile Nunc
4430-11 Round No Natural No Nunc
4431-11 Round No Natural No Nunc
4432-11 Round No Natural Sterile Nunc
9503230 Square Pierceable Natural No Nunc
9503233 Square Pierceable Natural Sterile Nunc
AB0675 Square No Natural No (autoclavable) Abgene
AB0566 Round Pierceable Natural No Abgene
AB0674 Round No Natural No (autoclavable) Abgene
AB0566SP** Round Pierceable Natural No Abgene
*Preslit silicone caps can be pierced repeatedly
**Mat is slightly more rigid than AB-0566
Super Sealer Heat Sealer
Thermo Scientific SuperSealer
Ensure a proper seal on plates, blocks and storage tube racks with the Thermo Scientific SuperSealer.
Learn more