Storage Plate Lids
Microplate lids protect samples from contamination and
evaporation during assay processing, incubation or storage.
  • 96-well lids with individual condensation rings
  • Lids have notches for quick orientation
  • Universal lids can be used on any standard height plate on the market
Available storage plate lids
SKU Height (mm) Cut-off corners Condensation rings Evaporation barrier Color Sterile Supported brand
263339 Standard 7.9 Long side Yes Yes Clear No Nunc
264122 Standard 7.9 Long side Yes Yes Clear Yes Nunc
264623 Standard 7.9 Long side Yes Yes Clear Yes Nunc
264611 Standard 7.9 Long side No Yes Clear Yes Nunc
264612 Standard 7.9 Long side No Yes Clear No Nunc
264616 Standard 7.9 Long side No Yes Clear Yes Nunc
448143 Standard 7.9 Long side No No White No Nunc
250002 Standard 8.34 No No No Clear Yes Universal (Abgene)
250003 Standard 8.34 No No No Clear No Universal (Abgene)
250005 Standard 8.34 No No No Clear No Universal (Abgene)
253623 Low profile 4.6 No No Yes Clear No Universal (Abgene)
253624 Low profile 4.6 No No Yes Clear Yes Universal (Abgene)
4954 Automation friendly 8.9 No No Yes Clear Yes Universal (Abgene)
4955 Automation friendly 8.9 No No Yes Black Yes Universal (Abgene)