Storage Caps and Seals
Accommodate a range of application and storage requirements. Matrix and Nunc caps are available in a range of formats to allow for your manual or automated storage needs.
Don't forget the easy to use SepraSeal Cap Removal Tool
Matrix Tube Caps & Seals
Matrix SepraSeal Capping Systems
Matrix SepraSeal Capping Systems
Designed for storage to -20°C and available solid or pre-split as well as sterile and non-sterile formats.
Matrix DuraSeal Capping Systems
For storage down to -80°C, DuraSeals can also be used for repetitive piercing procedures with their pre-split v-shaped design
Matrix ScrewTop Tube Caps
Color-coded caps and inserts available racked in trays or in bulk to suit a variety of applications.
Nunc Cryovial Caps
Nunc Cryobank and Bank-It Replacement Caps
Replacement caps supplied racked for manual or automated replacement. Available in natural, green, red or blue for simplified sample tracking.
Nunc CryoTube Colored Caps
Thermo Scientific Nunc CryoTube Colored Caps are for use with Nunc internally threaded CryoTube vials and include a silicone gasket to provide the best seal.
Support
Need technical support? Contact our expert team for technical and application support of laboratory products.