SepraSeal Caps
Thermo Scientific Matrix SepraSeal septa enables individual tube capping plus the ability to apply 96 septa at once and store down to -20°C.
  • Available in sterile, non-sterile, solid, or pre-split, and a range of colors for quick visual sample identification
  • Solid seals can be removed for sample access with the Thermo Scientific Matrix SepraSeal cap removal tool, or can be pierced once
  • For applications involving repetitive sample access, a pre-split Matrix SepraSeal can be pierced by automated liquid handling equipment, requiring very little pressure, while maintaining seal integrity with self sealing closure
For best results, apply capmats with the Thermo Scientific SuperSealer
Ordering Information
  • Designed for storage down to -20°C
  • Solid seals can be removed for sample access with the Thermo Scientific Matrix SepraSeal cap removal tool, or can be pierced once
  • Designed for storage down to -20°C, sterile
  • Solid seals can be removed for sample access with the Thermo Scientific Matrix SepraSeal cap removal tool, or can be pierced once
  • Designed for storage down to -20°C
  • Pre-split Matrix SepraSeals can be pierced by automated liquid handling equipment, requiring very little pressure, while maintaining seal integrity with self sealing closure
  • Designed for storage down to -20°C, sterile
  • Pre-split Matrix SepraSeals can be pierced by automated liquid handling equipment, requiring very little pressure, while maintaining seal integrity with self sealing closure
  • Designed for storage down to -20°C
  • Solid seals can be removed for sample access with the Thermo Scientific Matrix SepraSeal cap removal tool, or can be pierced once
  • Designed for storage down to -20°C, sterile
  • Solid seals can be removed for sample access with the Thermo Scientific Matrix SepraSeal cap removal tool, or can be pierced once
  • Designed for storage down to -20°C
  • Pre-split Matrix SepraSeals can be pierced by automated liquid handling equipment, requiring very little pressure, while maintaining seal integrity with self sealing closure
  • Designed for storage down to -20°C, sterile
  • Pre-split Matrix SepraSeals can be pierced by automated liquid handling equipment, requiring very little pressure, while maintaining seal integrity with self sealing closure