Search
A major challenge for microbiologists and virologists is the prediction of patterns of evolution and emergence of disease agents. RNA viruses like SARS-CoV-2 share the biological feature of high genetic variability, which causes them to appear as clouds of mutants. Coronavirus variants also emerge through antigenic shift within animal reservoirs, such as bats and snakes.
Ion Torrent targeted next-generation sequencing (NGS) enables a streamlined research workflow for complete genome sequencing and epidemiological studies of SARS-CoV-2. Through the use of a set of highly specific, universal coronavirus primers in combination with a high-fidelity master mix, all genomic segments are amplified and the DNA amplicons are sequenced on any Ion Torrent system to deliver highly accurate coronavirus typing in under a day.
The Ion AmpliSeq SARS-CoV-2 Insight Research Assay consists of 2 pools with amplicons ranging from 125–275 bp in length for complete coverage of over 99% of the viral genome and variants. This assay is part of an end-to-end solution, including a plug-in suite developed in collaboration with scientists at the forefront of the SARS-CoV-2 crisis.
Benefits of using Ion Torrent NGS include:
White Paper: SARS-CoV-2 research applications using the Ion GeneStudio S5 system
In this webinar, Dr. Timothy J. Triche MD, PhD, Co-Director of Center for Personalized Medicine at Children’s Hospital Los Angeles (CHLA), shares how he and his team used sequencing data generated from the Ion AmpliSeq SARS-CoV-2 Research Panel to analyze phylogenetic relationships among a cluster of six SARS-CoV-2-positive samples. By identifying two distinct lineages between the two sets, the team was able to conclude that the source of transmission wasn’t shared. Comparative analysis showed that, not unexpectedly, the mother and son had nearly identical strains of the virus that originated in Utah, with links to Europe, while the four health care workers had strains only distantly related to each other and to the family. This confirmed that there was no transmission between the patient family and the health care workers. Further, the health care workers acquired the virus from community spread and not from the patient and family member. "The NGS sequence data permitted Children's Hospital Los Angeles to draw conclusions within 48 hours of sample procurement from the six individuals. This information gave leaders at our institution greater confidence in our ability to provide a safe environment for our patients and our team members."
Watch webinar ›
Read the case study: Beyond emergency testing: staying ahead of SARS-CoV-2 with NGS to help disrupt the current trajectory ›
In addition to targeted sequencing, which covers >99% of the viral genome and variants, Ion Torrent systems can be used for de novo (whole genome) sequencing of SARS-CoV-2 by NGS .
Up to 80 samples can be multiplexed on a single Ion 540 Chip:
Number of samples | Chip |
|---|---|
| 2–3 samples | Ion 510 Chip |
| 4–6 samples | Ion 520 Chip |
| 15–20 samples | Ion 530 Chip |
| 60–80 samples | Ion 540 Chip |
Annotate variants, build consensus sequences, and construct de novo sequences for SARS-CoV-2 samples
Perform basecalling and alignment for single samples, run plugins for secondary analysis and easily manage your NGS data
Design on demand with ease and order Ion AmpliSeq research panels focused on your genes and organisms of interest
![]() | Ion AmpliSeq SARS-CoV-2 Insight Research Assay GS, manualConsists of the Ion AmpliSeq SARS-CoV-2 Insight Research Panel and Ion AmpliSeq Library Kit Plus reagents for those performing manual library preparation |
|
![]() | Ion AmpliSeq SARS-CoV-2 Insight Research Assay GS, Chef-readyConsists of the Ion AmpliSeq SARS-CoV-2 Insight Research Panel and the Ion AmpliSeq Kit for Chef DL8 for those utilizing the Ion Chef System for library preparation |
|
eBook: Advances in epidemiological research using NGS ›
Case study: Children's Hospital Los Angeles goes beyond emergency testing: staying ahead of SARS-CoV-2 with NGS to help disrupt the current trajectory ›
White paper: SARS-CoV-2 research applications using the Ion GeneStudio S5 system
Press Release: Early NGS data suggests SARS-CoV-2 is slow to mutate, see what this means for vaccine research
Press Release: Rapid SARS-CoV-2 genome sequencing aids epidemiological investigations
Poster: Custom next-generation sequencing primer designs for targeted sequencing of multi strain viral targets
For Research Use Only. Not for use in diagnostic procedures.