Library Quantitation
SOLiD® Library TaqMan® Quantitation Kit
  • Using a TaqMan® qPCR mix with universal cycling conditions, this accurate and easy-to-use kit provides a robust mechanism to correctly quantify the required template ratio for efficient sequencing.
RNA Spike-Ins
Ambion® ERCC Spike-In Control Mixes
Based on a common set of external RNA controls developed by the External RNA Controls Consortium (ERCC), these convenient mixes can be used across a variety of RNA experiments to measure dynamic range or quantitate differential gene expression.
Support
NEW   Next-Generation Sequencing Support Center
Find tips, troubleshooting help, and resources for your next-gen sequencing applications.