Multiplexing enables the pooling of samples prior to emulsion PCR (ePCR) on the SOLiD™ System. By incorporating more samples per run, multiplexing allows for significant decreases in the cost and handling requirements of ePCR, enrichment, and deposition. This substantially benefits applications such as whole-genome sequencing of small genomes or targeted resequencing of enriched samples.
Both DNA and RNA barcodes are specifically designed to prevent overlapping and permit simple, easy-to-interpret data analysis. All of the kits are packaged in sets of 16 barcodes, with up to 96 distinct barcodes available.
RNA barcodes
SOLiD® RNA Barcoding Kits
  • These easy-to-use kits help maximize RNA sequencing runs by pooling libraries prior to ePCR with up to 96 barcodes.
Learn more
DNA barcodes
SOLiD® DNA Barcoding Kits
  • These practical kits make the most of the extraordinary sequencing capacity of the SOLiD® System by multiplexing up to 96 libraries with DNA barcodes.
Learn more
Visistat reference component