Frequently Asked Questions
Answers
1. Is StemPro® Neural Supplement serum free?
Yes
2. What are the intended uses of StemPro® Neural Supplement?
StemPro® Neural Supplement is an optimized serum-free additive for the growth and expansion of neural stem and progenitor cells. For research use only??
3. Do I need to add other supplements?
Mixing of StemPro® Neural Supplement with a basal medium, such as Knockout™ DMEM/F-12 and Neurobasal®, and the addition of appropriate quantities of recommended growth factors yields a complete medium.
4. What are the key applications of StemPro® Neural Supplement?
StemPro® Neural Supplement supports the growth and expansion of neural stem cells and progenitor cells. It also supports the survival of neurons and differentiation of progenitor cells to astrocytes, oligodendrocytes, and Neurons. Besides its application can also be extended to transplantation studies and screening of drug candidates.
5. What species can I use Stempro® Neural Supplement to culture?
It can be used for human and rat NSCs, GPCs, and some neuronal progenitors.