Description
Custom 5’ labeled primer pairs include fluorescently labeled oligos with a choice of dye on the 5’ end, along with an unlabeled oligo in each pair. These products can be used in a variety of applications.
Features include:
  • Choice of delivered scales
  • For use in fragment analysis, gene expression studies, genotyping and PCR
  • All oligos are desalted
5' label: 6-FAM™, TET™, VIC®, HEX™, NED™, PET™
Catalog # Size Description 5' Reporter dye Quantity Order
450056 10,000 pmol 5’ Labeled/Unlabeled Primer Pairs 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 2 tubes Order now
450059 80,000 pmol 5’ Labeled/Unlabeled Primer Pairs 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 2 tubes Order now
450062 300,000 pmol 5’ Labeled/Unlabeled Primer Pairs 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 2 tubes Order now
4304976 10,000 pmol 5’ Labeled/Unlabeled Di-Repeats 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 2 tubes Order now
4304977 80,000 pmol 5’ Labeled/Unlabeled Di-Repeats 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 2 tubes Order now
4304978 300,000 pmol 5’ Labeled/Unlabeled Di-Repeats 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 2 tubes Order now
4304979 10,000 pmol 5’ Labeled/Unlabeled Di-Repeat + Tail 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 2 tubes Order now
4304981 80,000 pmol 5’ Labeled/Unlabeled Di-Repeat + Tail 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 2 tubes Order now
4304982 300,000 pmol 5’ Labeled/Unlabeled Di-Repeat + Tail 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 2 tubes Order now
*Pairs come with the labeled and unlabeled oligos in separate tubes.
Specifications
General specifications
Shelf Life: 12 months from manufacturing date
Purification: Desalted
Format: Delivered either in solution or dried down
Green features: Sustainable packaging, less waste
Shipping conditions: Room temperature
Contents and storage
Storage
If in liquid, option of either 1X TE or water
Storage at –20°C
Support
Order support
Order support
Contact Order & Web Support, Search FAQs, View Order History and Track Your Order
Technical support
Contact a Technical Application Scientist, Search Product Documents & FAQs and Instrument Maintenance & Repair Requests