Description
Custom 5’ labeled primers are fluorescently labeled oligos with a choice of dye on the 5’ end. These products can be used in a variety of applications.
Features include:
  • Choice of delivered scales
  • For use in fragment analysis, gene expression studies, genotyping, and PCR
  • All oligos are desalted. Larger scales of labeled primers have an option for HPLC purification.
  • IMPORTANT: HPLC purification reduces the yield in half
5' label: 6-FAM™, TET™, VIC®, HEX™, NED™, PET™
Catalog # Size HPLC yield 5' Reporter Dye Quantity Order
450007 10,000 pmol N/A 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 1 tube Order now
450006 80,000 pmol 40,000 pmol minimum 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 1 tube Order now
450017 300,000 pmol 150,000 pmol minimum 6-FAM™, TET™, VIC®, HEX™, NED™, PET™ 1 tube Order now
Specifications
General specifications
Shelf Life: 12 months from manufacturing date
Purification: Desalted, option of HPLC purification with larger scale
Format: Delivered either in solution or dried down
Green features: Sustainable packaging, less waste
Shipping conditions: Room temperature
Contents and storage
Storage
If in liquid, option of either 1X TE or water
Storage at –20°C
Support
Order support
Order support
Contact Order & Web Support, Search FAQs, View Order History and Track Your Order
Technical support
Contact a Technical Application Scientist, Search Product Documents & FAQs and Instrument Maintenance & Repair Requests