custom-taqman-probes-230
Custom Applied Biosystems™ TaqMan® MGB (minor groove binder), TAMRA™, and QSY™ probes are dual-labeled probes that can be used in all of your real-time PCR applications.
Features include:
  • Choice of delivered scales
  • Suitable for use in any real-time PCR application
  • HPLC purification of all TaqMan® probes
Which probe is right for your application?
Custom TaqMan MGB probes Best all-around probe for most applications.
Custom TaqMan TAMRA probes Good for maintaining consistency in projects that have used TAMRA probes.
Custom TaqMan QSY probes Best for multiplexing applications when 3 or more dyes are needed.
TaqMan probes and primers are available together as predesigned TaqMan® Assays
Learn more about multiplex real-time PCR
Support
  • Order support
    Contact Order & Web Support, Search FAQs, View Order History and Track Your Order
  • Technical support
    Contact a Technical Application Scientist, Search Product Documents & FAQs and Instrument Maintenance & Repair Requests