Description
Sequence detection primers are unlabeled primers that can be used with TaqMan® probes in all of your real-time PCR applications.
Features include:
  • Choice of delivered scales
  • For use in any real-time PCR or PCR application
  • All sequence detection primers are desalted
Real-time PCR sequence detection primers are delivered either in solution or dried.
Catalog # Size Quantity
4304970 10,000 pmol 1 tube Order now
4304971 80,000 pmol 1 tube Order now
4304972 130,000 pmol 1 tube Order now
Specifications
General specifications
Shelf Life: 12 months from manufacturing date
Purification: Desalted
Format: Delivered either in solution or dried down
Green features: Sustainable packaging, less waste
Shipping conditions: Room temperature
Contents and storage
Storage
Real-time PCR Primer: 10,000 pmol unlabeled primer, purified for use in sequence detection, per tube. If in liquid, customer can choose either 1X TE or water.
Storage at –20°C
Support
Order support
Order support
Contact Order & Web Support, Search FAQs, View Order History and Track Your Order
Technical support
Contact a Technical Application Scientist, Search Product Documents & FAQs and Instrument Maintenance & Repair Requests