Did you know that we offer ASR oligos?
In addition to our standard high-quality oligonucleotide service, Thermo Fisher Scientific is a manufacturer of Class 1 Analyte Specific Reagents that may be used in the development of in vitro diagnostic (IVD) applications.
Let us assemble your next ASR oligos
ASR oligos are custom-made to meet your specifications. Synthesis scales are available at 200 nmol and greater. Modifications can be made at the 5′, 3′, and internal locations of the oligo. Flexible packaging options are also available.
Please email us at asr.oligo@invitrogen.com to:
  • Discuss your project
  • Request a quote
  • Learn more about our quality systems and capabilities
ASR oligos are manufactured in accordance with "Good Manufacturing Practices (GMP)" as defined by the Food and Drug Administration Regulations, Title 21 CFR, Subchapter H, Part 820 Quality System Regulation and in conformance with ISO 13485:2003.
For more information or to request a quote, please contact us at custom.services@invitrogen.com.
Analyte Specific Reagent. Analytical and performance characteristics are not established.
Ordering Details
DNA Tubes
Plates
Alternate Ordering Methods
Primer Design