Buffer
The buffer type refers to the extraction buffer used to elute your DNA oligos once it has been synthesized. If you chose to receive the oligos lyophilized, keep in mind that the salts of the TE or Tris, if you were to select one of those options, would be part of your product as well since the oligos are dried down after they are extracted from the solid support used for the synthesis.