Cell culture heroes banner
Sophie Cowman, PhD candidate
Institute of Integrative Biology, University of Liverpool, England, UK
Sophie Cowman profile picture
Connect with Sophie
Biography
Sophie is passionate about understanding how the tumour microenvironment can influence DNA repair, which is the focus of her current research. In 2011 Sophie commenced a BSc in Molecular Biology at the University of Liverpool. In 2012, she was awarded a competitive scholarship enabling her to undertake a summer research internship in the lab of Dr Geraint Parry, investigating the nuclear pore complex. It was during this time that she developed a deep interest in scientific research and a love of working at the lab bench. Throughout her degree, her interests were cell biology, cancer biology and mechanisms behind DNA repair. This led to her current role as a PhD student in Dr Violaine Sée's Lab at the Department of Biochemistry, Institute of Integrative Biology, in the University of Liverpool, where she is currently in her final year. Throughout her PhD, Sophie's research primarily utilises cell culture techniques, gene expression analysis and advanced confocal microscopy, to investigating DNA repair mechanism in hypoxic brain tumours.
Learn about Sophie’s research
Title: Investigating DNA repair mechanisms in hypoxic brain tumours and the link to treatment resistance
Learning objectives
  • Understand how hypoxia can cause changes to DNA repair mechanism in cultured brain tumour cells
  • Understand how we can use advanced microscopy techniques to determine the functional impact of hypoxia-induced changes in DNA repair mechanism
Glioblastoma (GBM) and Medulloblastoma (MB) are the most common adult and pediatric brain tumors, both of which can have devastating consequences. Patients diagnosed with GBM have a life expectancy of around 15 months, whereas, for MB, the survival rate is higher. However, commonly used treatments for MB can have a negative impact on a child’s developing brain. Therefore, it is imperative that further research is conducted to understand the complex cell biology of these tumors to enable us to improve current treatment protocols.
Both GBM and MB are defined as hypoxic as their O2 levels are lower than the physiological 5% O2 found in the brain. Tumor hypoxia is known to enhance the ability cancer cells to invade other tissues and form tumors at secondary sites (metastasis), as well as causing resistance to chemotherapy and radiotherapy. Currently, little is known about how the chronic hypoxic tumor environment causes long-lasting cellular adaptations within tumor cells resulting in their resistant phenotype.
To further understand this, our lab investigates how long-term hypoxia exposure impacts DNA repair mechanisms within brain tumor cell lines, and how these changes can affect the response of cells to DNA damaging agents such as chemotherapeutic drugs and X-ray irradiation. We use a variety of complementary techniques including cell culture, gene expression analysis and advanced confocal microscopy.
We have observed down-regulation of key DNA repair proteins induced by hypoxia, causing the cells not to ‘recognize' certain types of DNA damage. Therefore, the cells are less likely to trigger apoptosis after cancer treatment. Additionally, further changes in DNA repair genes may cause a reduction in the efficiency of DNA repair, influencing the cell response to cancer therapy. It is hoped that gaining a deeper understanding of the effect of hypoxia in GBM and MB will aid in the development of more successful treatment methods.
Watch the webinar
I want to be the next Gibco Cell Culture Hero
As a Gibco Cell Culture Hero you will be a part of a growing community of global PhD and postdoc researchers who promote education and drive tomorrow's breakthroughs.
Complete the form below for a chance to present your research to a global audience via webinar, share your story of success and perseverance to the world on thermofisher.com.
Must be a PhD or postdoc using cell culture to apply. Must be passionate about communicating science within your social media networks.
Submit application
Links to content or other Internet sites should not be construed as an endorsement of the organizations, entities, views or content contained therein. The opinions and/or views expressed on social media platforms represent the thoughts of the individual and online communities, and not those necessarily of Thermo Fisher Scientific.
Share