BioPath Online
Pathway Focus: The T Cell Receptor (TCR) Pathway
TCR Signaling Pathway Antibodies
TCR Signaling Pathway Antibodies—ABfinity™ Technology Ensures Lot-to-Lot Consistency
Recombinant mouse sonic hedgehog C25II Recombinant Mouse Sonic Hedgehog C25II
Measure IL-2 Production in T Cells with ELISA Measure IL-2 Production in T Cells with ELISA—Easy-to-Use Invitrogen™ ELISA kits
New Antibodies
New Immunoassays
New Molecular Probes® Products
TCR Signaling Pathway Antibodies—ABfinity™ Technology Ensures Lot-to-Lot Consistency
  • Unparalleled lot-to-lot consistency
  • Superior sensitivity and specificity
  • Extensive validation and characterization
The T cell receptor (TCR) is a complex of integral membrane proteins that participates in the activation of T cells in response to the presentation of antigen. Stimulation of the TCR is triggered by major histocompatibility complex (MHC) molecules on antigen-presenting cells, inducing a series of intracellular signaling cascades that ultimately result in cellular proliferation, differentiation, cytokine production, and/or activation-induced cell death.
ABfinity™ recombinant monoclonal antibodies are now available for various members of the TCR pathway, enabling sensitive, specific detection of changes in expression, activity level, and phosphorylation state.
ABfinity™ antibodies, produced by proprietary Invitrogen technology, are developed by immunizing animals, screening for desired functionality, and then cloning the immunogen-specific antibody genes into high-level expression vectors. The antibodies are produced on a large scale by proprietary expression methods followed by purification with affinity chromatography. ABfinity™ antibodies are produced under ISO conditions with the highest quality controls, and maximum efforts are taken to ensure that every lot performs consistently.
Figure 1. Flow cytometry analysis of Jurkat cells labeled with rabbit anti-ZAP-70 [pY315/pY319]. Jurkat cells were fixed and permeabilized using FIX & PERM® reagents. Cells were then stained with 2 μg/test anti-ZAP-70 [pY315/pY319] followed by Alexa Fluor® 488 goat anti-rabbit Ig (gray trace). Pre-incubation with the phosphopeptide used as an immunogen decreased the signal (red trace) while pre-incubation with non-phosphopeptide had no effect (blue trace). Note that blue and gray traces are nearly identical.
Figure 2. Immunocytochemistry of Jurkat cells labeled with rabbit anti-ZAP-70 [pY315/pY319]. Jurkat cells were either untreated (top left) or treated with H2O2 (top right, bottom) and labeled with rabbit anti-ZAP-70 [pY315/pY319] (2 µg/ml). Alexa Fluor® 488 goat anti-rabbit at 1:1,000 was used as secondary antibody. Pre-incubation of treated cells with phosphopeptide immunogen decreased signal (bottom right) while pre-incubation with non-phosphopeptide did not (bottom right). Nucleus is stained with Hoescht (blue), AF488 signal (ZAP-70 [pY315/pY319], green).
Product
Quantity Species Reactivity
Applications
Cat. No.
Mnk1 [pT197/pT202] ABfinity™ Recombinant Rabbit Monoclonal Antibody
100 µg
Hu, B,* Ch,* Cn,* Cp,* Eq,* Mk,* Ms,* P,* Rt,* Sw,* X,* Z*
F, IF/ICC, WB
700242
T-bet ABfinity™ Recombinant Rabbit Monoclonal Antibody
100 µg
Hu, Cp,* Mk*
F, IF/ICC, IHC, WB
700059
ERK1/2 [pT185/pY187] ABfinity™ Recombinant Rabbit Monoclonal Antibody
100 µg
Hu, B,* Ch,* Cp,* Ms,* Rt,* X,* Z*
E, IHC, WB
700012
JNK1-2 [pTpY183/185] ABfinity™ Recombinant Rabbit Monoclonal Antibody (clone D12H7L17)
100 µg
Hu, B,* Ch,* Cn,* Cp,* Eq,* Gf,* Mk,* Ms,* Ne*, P,* Or,* Rt,* Sw,* X,* Z*
E, F, IF/ICC,    IHC, WB
700031
JAK1 [pY1022/pY1023] ABfinity™ Recombinant Rabbit Monoclonal Antibody - Purified
100 µg
Hu, B,* Ca,* Cp,* Eq,* Mk,* Ms,* Rt,* Sw,* X,* Z*
WB
700028
ZAP-70 [pY315/pY319] ABfinity™ Recombinant Rabbit Monoclonal Antibody
100 µg
Ms, Ha,* Rt*
WB, F, ICC
700177
*Reactivity predicted but not tested.
Reactivity: B = bovine; Ch = chicken; Cn = canine; Cp = chimpanzee; Eq = equine; Gf = goldfish; Mk = monkey (rhesus); Ms = mouse; Ne = nematode; P = primate; Or = orangutan; Rt = rat; Sw = swine; X = Xenopus; Z = zebrafish.
Applications: E = ELISA; F = flow cytometry; ICC = immunocytochemistry; IF = immunofluorescence; IHC = immunohistochemistry; WB = western blotting.
Recombinant Mouse Sonic Hedgehog C25II
  • High biological activity—more results with less protein
  • High purity—no interference from other proteins or contaminants
The Hedgehog signaling pathway is known for its role in development, but has also been shown to play a role in TCR signaling. In mature peripheral T cells, events induced by the Hedgehog pathway have shown to inhibit TCR-related activation and proliferation.
The Hedgehog pathway has also been shown to influence TCR repertoire selection in the thymus. We offer bioactive recombinant mouse sonic hedgehog C25II for use in cell culture applications.
Description Quantity Cat. No.
Recombinant mouse sonic hedgehog C25II
10 µg
PMC8034
Order 
Recombinant mouse sonic hedgehog C25II
100 µg
PMC8031 Order
Measure IL-2 Production in T Cells with ELISA—Easy-to-Use Invitrogen™ ELISA kits
  • Easy to use—simple, reliable assays
  • Versatile—quantify TNF-α in serum, plasma, or supernatant
  • Compatible—kits available for a variety of species
The T cell receptor (TCR) is a complex of integral membrane proteins that participates in the activation of T cells in response to the presentation of antigen. IL 2, formerly called T cell growth factor, is a 14–16 kDa glycosylated polypeptide produced by activated CD4+ TH cells which acts within an autocrine way to promote growth of T cells and NK cells. T cells respond to IL 2 via binding to high-affinity IL 2 receptors.
Precisely measure IL-2 in a variety of species, including human, mouse, rat, and monkey using precoated Invitrogen™ ELISA kits. ELISA kits provide an easy and reliable way to quantify the amount of IL-2 in your serum, plasma, or supernatant sample. Economy pack sizes are available for your convenience, and for additional savings, matched antibody pairs are available.
  • Learn More about Invitrogen™ ELISA & phosphoELISA™ Kits
Figure 3. Invitrogen™ ELISA kits provide an easy, reliable method for quantifying IL-2.
Figure 4. Mouse IL-2 parallelism using the IL-2 Mouse ELISA Kit (Cat. No. KMC0021). IL-2 was serially diluted in standard diluent buffer. The optical density of each dilution was plotted against the standard curve. Parallelism between the natural and recombinant protein was demonstrated, indicating that the standard accurately reflects natural Ms IL-2 content in samples.
 Sample  Average
Con-A (5 µg/mL), 6 hr
605 pg/mL
PMA (50 ng/mL), Ionophore (250 ng/mL), 6 hr
555 pg/mL
PMA (50 ng/mL), Ionophore (250 ng/mL), 24 hr
2,724 pg/mL
LPS (1 µg/mL), 24 hr
827 pg/mL
Table 1. Sixteen sera and sixteen plasma (heparin) samples were evaluated in this assay. All samples measured <15.6 pg/mL (the lowest Ms IL-2 standard). Mouse splenocytes were cultured and the supernatants were assayed for Ms IL-2 released.
Product
Range
Sample Volume Needed
Incubation Time
Quantity Cat. No.
ELISA Kits
IL-2 Human ELISA Kit
15.6–1,000 pg/mL
15.6–1,000 pg/mL
3 hrs
96 tests
KHC0021 Order Now
IL-2 Human ELISA Kit
15.6–1,000 pg/mL
15.6–1,000 pg/mL
3 hrs 192 tests
KHC0022 Order Now
IL-2 Human ELISA Kit
15.6–1,000 pg/mL
15.6–1,000 pg/mL
3 hrs 480 tests
KHC0021C Order Now
IL-2 Mouse ELISA Kit
15.6–1,000 pg/mL
15.6–1,000 pg/mL 3 hrs 96 tests
KMC0021 Order Now
IL-2 Mouse ELISA Kit
15.6–1,000 pg/mL
15.6–1,000 pg/mL
3 hrs 192 tests
KMC0022 Order Now
IL-2 Mouse ELISA Kit
15.6–1,000 pg/mL
15.6–1,000 pg/mL
3 hrs 480 tests
KMC0021C Order Now
IL-2 Rat ELISA Kit
23.4–1,000 pg/mL
23.4–1,000 pg/mL
3 hrs 96 tests
KRC0021 Order Now
IL-2 Rat ELISA Kit
23.4–1,000 pg/mL
23.4–1,000 pg/mL
3 hrs 192 tests
KRC0022 Order Now
IL-2 Monkey ELISA Kit 3.9–250 pg/mL
3.9–250 pg/mL
4 hrs 96 tests
KPC0021 Order Now
IL-2 Monkey ELISA Kit 3.9–250 pg/mL 3.9–250 pg/mL
4 hrs 192 tests
KPC0022 Order Now
Antibody Pairs (User Flexibility)
IL-2 Human Antibody Pair
--
--
--
10 Plates
CHC1243 Order Now
For research use only.  Not intended for human or animal therapeutic or diagnostic use.