Quicklinks
Related Product Information
Instructions for handling fresh hepatocytes after receipt of shipment
Important—Please read!
GIBCO® Fresh Hepatocytes are shipped in cold preservation medium designed to keep cells viable at 4°C. This shipping medium is not suitable for maintenance of hepatocytes in culture and should be exchanged with new medium, using the following instructions, immediately upon receipt.
Precautions
  • Treat all hepatocytes as potentially hazardous material
  • Wipe surfaces, and exteriors of bottles and hepatocyte containers with 70% isopropanol
  • Perform all operations using aseptic handling technique in a sterile environment
Step 1—Prepare supplemented medium
Option 1: Suspension use (if plating cells, see option 2 below)
For experiments where the hepatocytes will be used in suspension, supplement Williams Medium E without Phenol Red (Cat. No. CM6000), or suitable alternative basal medium, with the appropriate amount of Maintenance Supplement Pack, Serum-free (Cat. No. CM4000). See Table 1. Filtering is recommended, but not required. Use a water bath to warm the supplemented medium to 37°C prior to use with hepatocytes.
Contents of Maintenance Supplement Pack, Serum-free:
  • Dexamethasone, 10 mM in DMSO solution, 1 x 250 μL (supplied in excess)
  • Maintenance Cocktail solution, 1 x 20 mL, containing:
  • 2.5 mL penicillin-streptomycin, 10,000 U/mL
  • 5.0 mL ITS+ (insulin, transferrin, selenium complex, BSA, linoleic acid)
  • 5.0 mL GlutaMAX™, 100X
  • 7.5 mL HEPES, pH 7.4; 1M
Table 1
—Volumes for supplementing medium with Maintenance Supplement Pack, Serum-free (Cat. No. CM4000) for use with hepatocytes in suspension.
Medium 50.0 mL 100 mL 200 mL 500 mL
Dexamethasone 0.50 μL 1.00 μL 2.50 μL 5.00 μL
Maintenance cocktail 2.00 mL 4.00 mL 10.0 mL 20.0 mL (entire tube)
Option 2: Plated use (if using cells in suspension, see option 1 above)
For experiments where the hepatocytes will be used in a plated format, supplement Williams Medium E without Phenol Red (Cat. No. CM6000), or suitable alternative basal medium, with the appropriate amount of Plating Supplement Pack Serum-containing (Cat. No. CM3000). See Table 2. Filtering is recommended, but not required. Use a water bath to warm the supplemented medium to 37°C prior to use with hepatocytes.
Contents of Plating Supplement Pack, Serum-containing:
  • Fetal Bovine Serum, pre-qualified for use with primary hepatocytes, 1 x 25 mL
  • Dexamethasone, 10 mM in DMSO solution, 1 x 250 μL (supplied in excess)
  • Thawing/Plating Cocktail solution, 1 x 18 mL, containing:
  • 5.0 mL penicillin-streptomycin, 10,000 U/mL
  • 500 μL bovine insulin, 4 mg/mL
  • 5.0 mL GlutaMAX™, 100X
  • 7.5 mL HEPES, pH 7.4; 1M
Table 2
—Volumes for supplementing medium with Plating Supplement Pack, Serum-containing (Cat No. CM3000) for use of hepatocytes in a plated format. Each plate requires 12 mL of supplemented media daily.
Medium 50.0 mL 100 mL 250 mL 500 mL
FBS 2.50 mL 5.00 mL 12.5 mL 25.0 mL (entire tube)
Dexamethasone 5.00 μL 10.0 μL 25.0 μL 50.0 μL
Maintenance cocktail 1.80 mL 3.60 mL 9.00 mL 18.0 mL (entire tube)
TOP
Step 2—Centrifuge and resuspend hepatocytes
  1. Remove tube(s) of hepatocytes from shipping container; record lot number.
  2. Slowly invert tube a few times to mix cell suspension.
  3. Centrifuge hepatocytes at 4°C :
    • Human hepatocytes - 100 x g for 10 min
    • Animal hepatocytes - 76 x g for 6 min
  4. Aspirate supernatant carefully, leaving approximately 1 mL of medium in tube.
  5. Gently resuspend pellet in either incubation or plating media - target 1 million cells/mL unless experiment requires higher concentration of cells.
TOP
Step 3— Determine viability and viable cell yield by Trypan Blue exclusion count
Prepare cell counting vial
  1. Mix 400 μL buffer (PBS or similar) with 50 μL 0.4% Trypan Blue stain (Invitrogen Cat. No. T10282) in a 1.5 mL microcentrifuge tube (this is the cell counting vial).
  2. Before adding a portion of the hepatocyte suspension to the counting vial, gently invert the tube of hepatocytes several times to ensure a homogeneous solution. This step is crucial to obtaining an accurate cell count.
  3. Using a wide-bore tip, pipette 50 μL of the hepatocyte suspension into the cell counting vial by drawing the solution into the pipette tip once and depressing the contents into the cell counting vial. Do not rinse out the tip. This results in a 10-fold dilution of the original cell suspension.
  4. Gently flick/tap the counting vial to ensure a homogeneous mixture. Do not shake or agitate vigorously, as this will cause cell death and result in an inaccurate viability determination.
    Prepare hemocytometer
  5. Wait one minute, gently flick/tap the counting vial again to mix, and then slowly pipette 10 μL of the mixture into the V-shaped groove on one side of the hemocytometer. Capillary action will pull the cell suspension into the hemacytometer.
  6. Before counting, quickly scan the quadrants under the microscope to ensure the cells are evenly distributed. To ensure an accurate cell count, aim to have approximately 80 to 140 cells per quadrant. Make note if you change dilution factors, and reload if the distribution appears uneven or contains bubbles.
    Count hepatocytes
  7. Immediately count the viable hepatocytes (clear or yellow color) and dead hepatocytes (blue color) in the four outer quadrants of the hemocytometer.
  8. Determine viability of hepatocytes (live cells/total cells x 100).
  9. Determine total cell density (cells/mL = average number of live cells x 10,000 x 10). Where 10,000 is the surface area of the hemacytometer and 10 is the dilution factor.
    Adjust cell density
  10. To use hepatocytes in suspension assays, adjust cell density to 0.5-2.0 x 106 viable cells/mL as experiment requires.
  11. To use hepatocytes in a plated format, adjust viable cell density according to plate format and species. See tables per species per plate format for recommendations. Note: It is recommended to do a visual density check to ensure your seeding density is accurate.
Table 3a—Seeding density (million/mL)
Plate format Human Rat Dog  Mouse Cynomolgus
6-well 0.850 0.750 0.950 0.380 1.00
12-well 0.670 0.670 0.670 0.340 0.900
24-well 0.600 0.600 0.600 0.300 0.800
96-well 0.600 0.600 0.600 0.300 0.800
48-well 0.500 0.500 0.500 0.270 0.700
Table 3b—Cells per plate (millions)
Plate format Human Rat Dog  Mouse Cynomolgus
6-well 10.2 9.00 11.4 4.56 12.0
12-well 8.04 8.04 8.04 4.08 10.8
24-well 7.20 7.20 7.20 3.60 9.60
96-well 3.60 3.60 3.60 1.80 4.80
48-well 6.00 6.00 6.00 3.24 8.40
Table 3c—Cells per well (millions)
Plate format Human Rat Dog  Mouse Cynomolgus
6-well 1.70 1.50 1.90 0.760 2.00
12-well 0.670 0.670 0.670 0.340 0.900
24-well 0.300 0.300 0.300 0.150 0.400
96-well 0.0375 0.0375 0.0375 0.0188 0.050
48-well 0.125 0.125 0.125 0.0675 0.175
TOP
Technical support
For questions related to this protocol, contact us at:
Email: hepaticproducts@lifetech.com
Phone: +1 919 237 4500 (Toll)
Phone: +1 866 952 3559 (U.S. Toll-free)
LT139                        3-Mar-2011