Quicklinks
Related Product Information
Introduction
Dynabeads® M-270 Carboxylic Acid are uniform, monosized superparamagnetic beads composed of highly cross-linked polystyrene with magnetic material precipitated in pores evenly distributed throughout the particles. The particles are further coated with a hydrophilic layer of glycidyl ether, concealing the iron oxide inside the Dynabeads Carboxylic acid groups are then introduced on the surface. Their hydrophilic surface ensures low non-specific binding, excellent dispersion abilities and easy handling in a wide variety of buffers. Dynabeads M-270 Carboxylic Acid are supplied in an aqueous suspension.
Principle
The Dynabeads M-270 Carboxylic Acid is designed to act as a solid support for a wide variety of biomagnetic separations and manipulations. Their size makes them particularly suitable for isolation of proteins. They can be applied to protein applications such as sample preparation, bioassays or the selection of affinity binders. The ver  rapid and gentle coupling chemistry of the ligand-immobilization reaction make them very useful in coupling labile proteins, peptides and functional enzymes for the isolation of a wide variety of targets (e.g. hormones, receptors, disease markers, bacteriophages etc). Due to the gentle pull to the magnet, the 2.8 μm Dynabeads can also be used for the isolation of fragile cells. For cell separation in general, Invitrogen Dynal recommends the use of the larger 4.5 μm Dynabeads. Activation of the Dynabeads M-270 Carboxylic Acid can be performed with a carbodiimide followed by coupling of an amine containing ligand, resulting in a stable amide bond between the bead and the ligand. The mechanism and optimization of carbodiimide mediated amide bond formation is extensively discussed in the literature (1,2,3,4). Alternatively, a bifunctional cross-linker may be used to introduce other functional groups like thiol, amine, maleimide etc. If the ligand to be bound is an oligonucleotide, it does not contain a primary amino function. This can be introduced by e.g. using 5'-amino modified oligonucleotides. Please note that other aminogroups in the oligonucleotide might to some degree react with the carboxylic acid groups on the beads, resulting in coupling via the internal bases. Once coupled with your ligand, the Dynabeads can be added to a cell lysate or other suspensions containing your target molecule. After a short incubation allowing affinity capture of the target, the Dynabeads are pulled to the side of the test-tube by the use of a magnet (Dynal® MPC™) allowing aspiration of unbound material. Furthermore, the magnetic separation facilitates washing and concentration of the isolated target bound to the beads. Dynabeads with bound target molecule can be used directly in downstream bioassays, or can be boiled in application buffer and analyzed on SDS-PAGE. Alternatively, the target molecule can be eluted off the Dynabeads with conventional elution methods such as high salt, low pH etc.
TOP
Protocol
For coating of a ligand to Dynabeads M-270 Carboxylic Acid, Invitrogen recommends to use 20 μg pure protein or ~700 pmol oligonucleotides/peptides per mg Dynabeads and a final concentration of 10-30 mg beads per ml during incubation. The suggested protocols described in sections below, illustrate an example using 3 mg of Dynabeads, but should be scaled up or down to suit specific needs. It is recommended that the protocols are optimized to meet your requirements (e.g. sample volume, concentration of ligand/ beads/EDC, MES buffer volume and pH).
Calculations:
The concentration of the supplied beads is 30 mg/ml. 3 mg beads = 100 μl. Protein concentration is in this example set to 1 mg/ml. Given that 20 μg protein is required per mg Dynabeads, the required amount of protein in this case is 60 μg. This corresponds to a volume of 60 μl ligand. The required amount of EDC varies depending on the performed coating procedure in below. The final sample volume should be 100 μl to meet the recommendation of final Dynabeads concentration of 30 mg/ml. In brief, according to the calculation above you require: 100 μl of washed Dynabeads (see the washing step under each protocol) 60 μl ligand. MES Buffer to adjust the final volume to 100 μl.
Activation and Coupling of Ligand
The traditional procedure for ligand coupling is the formation of an amide bond between a primary amino group of the ligand and the carboxylic acid groups on the surface of the Dynabeads, mediated by carbodiimide activation. The intermediate product of the reaction between the carboxylic acid and the carbodiimide is very labile and will hydrolyse quickly. To get the desired immobilization of the ligand it is therefore important to have the ligand immediately available. Alternatively, the activated Dynabeads can be captured as a less labile intermediate, like an N-hydroxyl succinimide ester (use NHS, MW 115.1 or sulfo-NHS, MW 217.1), and then react with the ligand over a longer period. There are several alternative protocols for carbodiimide- mediated immobilization of ligand by amide bond formation:
One-step protocol (see protocol One-Step Coating Procedure below):
  1. Mix ligand and Dynabeads.
  2. Add carbodiimide and incubate the reaction mixture.
  3. Wash the Dynabeads and resuspend in buffer.
Two-step protocol (see Two-Step Coating Procedure (without NHS) below):
  1. Add carbodiimide to the Dynabeads.
  2. After 30 minutes incubation, quickly wash the Dynabeads in cold water followed by cold buffer.
  3. Add the ligand to the Dynabeads and incubate further.
  4. Wash the Dynabeads and resuspend in buffer.
Two-step protocol with NHS (see protocol Two-Step Coating Procedure using NHS below):
  1. Add a mixture of carbodiimide and N-hydroxy succinimide to the Dynabeads.
  2. After 30 minutes incubation, wash the Dynabeads to remove excess carbodiimide.
  3. Add the ligand to the Dynabeads and incubate further.
  4. Wash the Dynabeads and resuspend in buffer. The one-step protocol is recommended when using ligands that do not contain carboxylic acid groups (e.g. oligonucleotides). If the ligand does contain carboxylic acid groups  these may react with the carbodiimide and cause polymerization of the ligand. Since this method is less laborious and generally gives higher yields, it may however still be advantageous to use this method if it is possible to add ligand in excess to compensate for the loss due to polymerization. The two-step protocol is preferred when the ligand contains carboxylic acid groups and you have limited amounts of the ligand available. The two-step protocol without NHS requires a very fast wash of the beads in cold buffer after the activation. The two-step protocol with NHS should be used if the ligand is in an alkaline buffer or a buffer with high phosphate concentration. 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride (EDC, MW 191.7) is the carbodiimide most commonly used. It is very soluble in water and relatively stable as an aqueous solution. For some applications, other carbodiimides may slightly improve the results. As an example: When coupling IgG or IgM, without using NHS, the more hydrophobic carbodiimide N-cyclohexyl-N'-(2-morpholinoethyl) carbodiimide methyl-p-toluensulfonate (CMC, MW 423.6) generally improves yield and orientation of the immobilized antibody.
One-Step Coating Procedure
TOP
  1. Wash Dynabeads M-270 Carboxylic Acid twice with 25 mM MES, pH 5, using the equal volume of Dynabeads (100 μl) pipetted out of the vial, for ten minutes with good mixing (en-over-end or similar).
  2. Add the required amount of ligand (60 μg) in 25 mM MES, pH 5 (60 μl) to the washed Dynabeads. Mix well and incubate with slow tilt rotation at room temperature for 30 minutes.
  3. Immediately before use dissolve EDC in cold 100 mM MES, pH 5 to a concentration of 100 mg/ml.
  4. Add 30 μl EDC solution (3 mg) to the Dynabeads/ligand suspension. Mix well.
  5. Add 10 μl of 25 mM MES, pH 5 to final volume of 100 μl.
  6. Incubate for 2 hours or longer (over night) at 4°C with slow tilt rotation.
  7. Wash the coated Dynabeads as described below.
Two-Step Coating Procedure (without NHS)
I) Activation with EDC
  1. Wash Dynabeads M-270 Carboxylic Acid twice with 0.01 M NaOH, using the equal volume of Dynabeads (100 μl) pipetted out of the vial, for ten minutes with good mixing (en-over-end or similar).
  2. Wash three times with 100 μl de-ionized water in the same manner. Remove excess liquid.
  3. Dissolve the EDC in cold, de-ionized water to 19-76 mg/ml (0.1-0.4M). Add 100-200 μl of EDC-solution to the Dynabeads. Vortex to mix properly.
  4. Incubate for 30 minutes at room temperature with slow tilt rotation.
  5. After incubation, place the tube on the magnet for 4 minutes and remove the supernatant. Wash once with cold, de-ionized water and once with 50 mM MES, pH 5, as quickly as possible to avoid hydrolysis of the activated carboxylic acid groups.
The Dynabeads are now activated and ready for coating with a ligand containing primary amine groups. Activated beads cannot be stored and you should proceed directly to the next step:
II) Immobilization of ligand after activation
TOP
  1. Remove the wash solution used in step 5 above. Add the required amount of ligand (60 μg) in 50 mM MES, pH 5 (60 μl) to activated Dynabeads.
  2. Add 40 μl of 50 mM MES, pH 5 to final volume of 100 μl. Vortex to ensure good mixing.
  3. Incubate for at least 30 minutes at room temperature, or 2 hours at 4°C, with slow tilt rotation.
  4. After incubation, place the tube on the magnet for 4 minutes and remove the supernatant.
  5. Wash the coated Dynabeads as described below
Two-Step Coating Procedure using NHS
I) Activation with EDC and NHS:
  1. Wash Dynabeads M-270 Carboxylic Acid twice with 25 mM MES, pH 5, using the equal volume of Dynabeads (100 μl) pipetted out of the vial, for ten minutes with good mixing (en-over-end or similar).
  2. Immediately before use dissolve EDC in cold 25 mM MES, pH 5 to a concentration of 50 mg/ml.
  3. Similarly, prepare a 50 mg/ml solution of NHS in 25 mM MES, pH 5.
  4. Add 50 μl of EDC solution and 50 μl of NHS solution to the washed Dynabeads. Mix well and incubate with slow tilt rotation at room temperature for 30 minutes.
  5. After incubation, place the tube on the magnet for 4 minutes and remove the supernatant. Wash twice with 100 μl of 25 mM MES, pH 5.
The Dynabeads are now activated and ready for coating with a ligand containing primary amine groups.
TOP
II) Immobilization of ligand after activation
  1. Add the required amount of ligand (60 μg) in 25 mM MES, pH 5 (60 μl) to activated Dynabeads.
  2. Add 40 μl 25 mM MES, pH 5 to final volume of 100 μl. Vortex to ensure good mixing.
  3. Incubate for at least 30 minutes at room temperature, or 2 hours at 4°C, with slow tilt rotation.
  4. After incubation, place the tube on the magnet for 4 minutes and remove the supernatant.
  5. Wash the coated Dynabeads as described below
Washing of Coated Beads
All immobilzation procedures require washing of the coated Dynabeads to remove excess ligand and to block un-reacted surface. NOTE: In order to quench the non reacted activated carboxylic acid groups, incubate the Dynabeads coated with ligand with either 50 mM Tris pH 7.4 for 15 minutes or 50 mM ethanolamine in PBS pH 8.  for 60 minutes, both at room temperature with slow tilt rotation.
  1. Wash the coated Dynabeads a total of four times with 100 μl PBS or 50 mM Tris. Blocking protein like BSA or skimmed milk powder may be added to a concentration of 0.1 - 0.5 % when it does not interfere with downstream applications of the Dynabeads. Also 0.1% Tween-20 or Triton X-100 can be added during washes to reduce nonspecific binding.
  2. Resuspend the coated Dynabeads to the desired concentration in PBS or a Tris storage buffer. The Dynabeads are now ready for use.
Store the coated Dynabeads at 2-8°C. Addition of 0.1 - 0.5 % protein (BSA) and/or 0.01 - 0.1 % Tween-20 or Triton X-100 is recommended to stabilize the immobilized ligand and increase the ease of handling. Coated Dynabeads can usually be stored for several months at this temperature, depending on the stability of the immobilized ligand. A final concentration of 0.02% (w/v) sodium azide (NaN3) may be added as a bacteriostatic agent. If the coated Dynabeads are stored for more than two weeks, they should be washed twice for five minutes with a buffer suitable for the application prior to use.
Isolation of Target Molecule
Efficient isolation of target molecules using Dynabeads is dependent on the bead-concentration, target molecule concentration, the ligand's affinity for the target molecule and time. Binding is performed from 10 minutes to 1 hour, at a recommended concentration of 1-10 x 109 beads/ml. Target-ligand equilibrium is reached after approximately 1 hour.
  1. Add sample containing target molecule to the coated Dynabeads (3 mg beads). For a 100 kD protein, use a volume containing approximate 25 μg target molecule to assure an excess of this molecule.
  2. Incubate the mixture with tilting and rotation for one hour to capture the target (incubation times as low as 10 minutes can be used with concentrated protein samples in volumes close to what was originally pipetted from the vial).
  3. Place the tube on the magnet for 4 minutes to collect the Dynabeads at the tube wall. For viscous samples, increase the time on the magnet. Pipette off the supernatant.
  4. Wash the Dynabeads 3 times using 1 ml PBS each time and exchanging buffers by the use of the magnet. 
Efficient isolation of target molecules using Dynabeads is dependent on the bead-concentration, target molecule concentration, the ligand's affinity for the target molecule and the specific binding kinetics involved.
The concentration of required Dynabeads will depend on the size of your specific molecule. Also the salt-concentration and pH of the chosen binding, washing and elution buffers can be varied depending on the type of molecule to be immobilized. Similarly, the selected buffer used in the downstream application should be optimized for the specific application. The size of the Dynabeads M-270 Carboxylic Acid presents a high surface area per mg beads and a corresponding high capacity for the target molecule. The effective binding capacity will depend on the size of the specific molecules to be immobilized. As the Dynabeads M-270 Carboxylic Acid will not inhibit enzymatic activity, bead-bound material can be used directly in downstream analysis. Alternatively, the target molecule can be eluted off the Dynabeads following conventional elution methods.
Target Protein Elution Procedure
TOP
Conventional elution methods can be applied for the elution of target protein from the Dynabeads. Low pH (2.8-3.5), change in ionic strength, affinity elution, electrophoresis, polarity reducing agents, deforming eluants ca  be applied, or even boiling the bead-target complex in SDS-PAGE application buffer for direct characterization of protein on SDS-PAGE. The method of choice depends on the affinity of the specific target molecule to the ligand coated onto the Dynabeads, the stability of the target molecule and the downstream application and detection method. Most proteins will be eluted off at pH 3.1 following the procedure described below, but some protein functionality might be lost under such harsh conditions. If maintaining functionality of the target molecule is important, try milder elution conditions first such as high salt (e.g. 2M NaI) or stepwise elution reducing pH from 6 down to 3. This is also recommended if the beadbound ligand must remain functional to allow reuse of the Dynabeads.
  1. Add 30 μl 0.1 M citrate (pH 3.1) to the Dynabeads with immobilized target.
  2. Mix well by tilting and rotation for 2 minutes.
  3. Place the test tube on the magnet and transfer the supernatant, containing purified target, to a clean tube.
  4. Add additional 30 μl 0.1 M citrate (pH 3.1) to the Dynabeads to elute any remaining target.
  5. Mix well by tilting and rotation for 2 minutes.
  6. Place the test tube on the magnet, pipette off the eluate and pool the supernatants containing pure target molecule.
Total collected volume = 60 μl
To ensure reuse of the Dynabeads and functionality of the isolated target molecule, bring both the Dynabeads and the target molecules back to physiological pH (7.4) immediately after elution.
TOP
General Information
Invitrogen Dynal AS complies with the Quality System Standards ISO 9001:2000 and ISO 13485:2003.
Product Characteristics
Typical characteristics for any given batch of this product:
Diameter: 2.8 μm
Density: 1.6 g/cm3
Specific surface area: 2-5 m2/g beads
Active chemical functionality: 150 μmol/g beads
Concentration: 2 x 10 9 beads/ml (approx. 30 mg/ml)
Certificate of Analysis (CoA) is available upon request. Material Safety Data Sheet (MSDS) is available at ‘ www.invitrogen.com’.
TOP
References
  1. Nakajima N and Ikade Y, "Mechanism of Amide Formation by Carbodiimide for Bioconjugation in Aqueous Media", Bioconjugate Chem. 1995, 6(1),123-130.
  2. Gilles MA, Hudson AQ and Borders CL Jr, "Stability of water-soluble carbodiimides in aqueous solution", Anal Biochem. 1990 Feb 1;184(2):244-248.
  3. Sehgal D and Vijay IK, "A method for the high efficiency of water-soluble carbodiimide-mediated amidation", Anal Biochem. 1994 Apr;218(1): 87-91.
  4. Szajani B et al, "Effects of carbodiimide structure on the immobilization of enzymes", Appl Biochem Biotechnol. 1991 Aug;30(2):225-231.
TOP
143.05D_06D.indd          5-May-2007