protocol
Quicklinks
Related Product Information
Overview for Electrophoresis and Western Blotting
Comparison of Buffer Systems for Protein Separations
Dissociating/Non-Dissociating Native Buffer Systems
Many studies employing zone electrophoresis of proteins in a polyacrylamide gel use a buffer system designed to dissociate all proteins into their individual polypeptide subunits. The most common dissociating agent is sodium dodecyl sulfate (SDS), an ionic detergent which denatures proteins by wrapping around the polypeptide backbone. By heating the protein sample at 100°C in the presence of excess SDS and thiol reagent, disulfide bonds are cleaved and the protein is fully dissociated into its subunits. Under these conditions most polypeptides bind SDS in a constant weight ratio (1.4 g of SDS:1 g of polypeptide). The intrinsic charges of the polypeptide are insignificant compared to the negative charges provided by the bound detergent so that the SDS-polypeptide complexes have essentially the same negative charge and shape and migrate through the gel strictly according to polypeptide size. The simplicity and speed of this method, plus the fact that only microgram quantities of protein are required, have made SDS-PAGE the most widely used method for determination of molecular mass in a polypeptide sample. Proteins from almost any source are readily solubilized by SDS so the method is generally applicable.
In contrast, non-dissociating native buffer systems, where proteins are electrophoresed in their native form, are designed to fractionate a protein mixture such that subunit interaction, native protein conformation, and biological activity are preserved. Proteins in non-dissociating native systems are separated based on their charge-to-mass ratio.
Continuous Buffer Systems
In continuous buffer systems, the composition of the gel buffer and running buffer are essentially identical throughout the gel. They consist of a single separating gel and use the same buffer ions at the same pH throughout the sample, gel, and electrode reservoirs. In continuous systems, molecular charge density and gel pore size are the only factors that have any effect on the stacking, or zone sharpening, of a molecule. The proteins in a sample applied to a continuous system will move faster in the sample well than in the gel.
A concentrated band of sample will form where the molecules are slowed down at the interface of the buffer and the restrictive gel. This type of stacking has limitations, particularly when separating molecules which vary widely in size. The smaller molecules have less of a difference between their free solution mobility and their mobility in a gel so the sharpening factor is not as favorable as for the larger molecules.
Discontinuous Buffer Systems
The discontinuous buffer systems employ different buffer ions and pH in the gel and in the electrode reservoirs. Samples are loaded onto a non-restrictive large pore gel, called a stacking gel, which overlays a smaller pore resolving gel. The major advantages of discontinuous buffer systems are that relatively large volumes of dilute protein samples can be applied to the gel and resolution is much greater than that obtained with continuous systems. This increased resolution is a direct result of the way proteins concentrate, or stack, into narrow zones during migration through the large-pore stacking gel, and destack or resolve, under the given conditions in the small pore resolving gel. The production of thin starting zones by discontinuous systems is described by Ornstein and Davis (1,2).
Ion Mobility and Protein Stacking
Ion mobility is dependent on the charge density of a molecule and the voltage gradient. At a given pH only part of a population of protein molecules will be dissociated (i.e., charged) at any time. The velocity at which these molecules migrate will be dependent upon the effective mobility and the voltage gradient. Therefore, an ion of lower mobility can migrate as fast as one with higher mobility if the products of voltage and effective mobility are equal.  In the Ornstein and Davis system, the sample and stacking gel contain Tris-HCl buffer whereas the upper electrode reservoir contains  Tris-Glycine buffer. At the pH of the sample buffer and stacking gel (pH 6.7), glycine is weakly ionized and therefore, its mobility is low. Chloride is completely ionized and has a much higher mobility, while the mobility of proteins are intermediate between that of glycine and chloride. Once a voltage is applied, chloride (leading) ions migrate away from the glycine (trailing) ions leaving behind a zone of lower conductivity, a higher voltage gradient, and higher pH. The zone accelerates the glycine so it keeps up with the chloride ions. As this glycine/chloride boundary moves through the sample and stacking gel, any proteins in front are rapidly overtaken by chloride ions which have a higher velocity. Behind the moving boundary, the proteins have a higher velocity than glycine. Therefore, the moving boundary sweeps up the proteins, concentrating them into thin zones or stacks in order of decreasing mobility.
What Happens Once the Proteins have been Stacked?
First, the stacked proteins may migrate through a low percentage gel, remaining stacked or unresolved behind the moving boundary or dye front. Second, the proteins may unstack or resolve from the moving boundary. Ornstein and Davis described two different methods for unstacking. Unstacking can be achieved by a) slowing down the protein after it has been stacked, or b) by speeding up the trailing ions once the protein has been stacked.
In the first approach, a low percent stacking gel is cast over a higher percentage resolving gel. As the stacked protein enters the higher percentage resolving gel, its mobility is reduced so that it is slower than the glycine ion. Once this occurs, the protein escapes the stack and migrates as if it were in a continuous gel at a similar, lower, local field strength. The second means of unstacking is to use a shift in pH to accelerate the trailing ions. For the glycine/chloride system, a stacking gel is cast so that the glycine zone will regulate to a lower pH than the resolving gel. When the glycine enters the higher pH resolving gel, its net negative charge will increase its effective mobility. Consequently, the glycine overtakes some of the proteins and now migrates directly behind the chloride ions, causing these proteins to resolve. Ornstein and Davis combined both of these unstacking methods in their Tris-chloride/Tris-glycine discontinuous buffer systems by using a low percentage stacking gel at pH 6.8 over a higher percentage separation gel at pH 8.8. Laemmli later adapted the system for SDS electrophoresis of proteins, keeping essentially the same features (3).
The Relative Importance of Stacking and pH in SDS PAGE Systems
We frequently are asked if the Novex® Pre-Cast Gels have a stacking gel and, if so, what is its pH. The answer is yes, there is a stacking gel (for all NuPAGE®, Tris-Glycine, and Tricine gels), but the pH is the same between the stacking gel and the separating gel. The pH varies depending on the formulation of the gel. Please refer to individual products for the pH value. We have found that the pH discontinuity of the SDS system plays a much smaller role in stacking proteins than does the ion species discontinuity of the system. In the Laemmli system, the stacking gel pH of 6.8 may serve as an extra boost for the stacking system but is not actually required to stack SDS/protein complexes. Even at the separating gel pH of the Laemmli system (pH 8.8), glycine is slow enough so that proteins smaller than about 70 kDa will stack in a 4% gel just as well as they would in a pH 6.8 stacking gel. It should be noted that the Ornstein and Davis system was originally developed for separating native proteins. They found that a low pH stacking gel was required to effectively stack some of these proteins. SDS-bound proteins, however, have a considerably greater negative charge density than native proteins. Consequently, the glycine mobility is low enough to stack SDS-bound proteins ahead of the glycine zone even when the stacking gel is set up at the same pH as the resolving gel. Proteins as large as 70 kDa will remain stacked in a 4% gel at pH 8.6–8.8 just as well as in a 4% gel at 6.8–7.2. The larger proteins will also be separated into discrete bands. Stacking occurs at the interface between the free sample solution and the stacking gel, which promotes band sharpness for the larger proteins. Small and large proteins migrate equally in free solution, but when the larger proteins contact the stacking gel they are slowed considerably more than the smaller proteins and are effectively concentrated. With typical sample volumes, this concentrating effect produces bands over the entire range which are as sharp as those obtained with a pH 6.8 stacking gel.
References:
  1.    Ornstein, L. (1964) Ann. NY Acad. Sci., 121: 321.
  2.    Davis, B.J. (1964) Ann. NY Acad. Sci.: 121: 404.
  3.    Laemmli, U.K. (1970) Nature 227: 680-685.
Comparison of Discontinuous Buffer Systems
SDS-PAGE utilizes a discontinuous buffer system to concentrate, or “stack,” samples into a very sharp zone in the stacking gel at the beginning of the run. In a discontinuous buffer system, the primary anion in the gel is different (or discontinuous) from the primary anion in the running buffer. Both the NuPAGE® systems (Bis-Tris and Tris-Acetate gels) and the Laemmli (Tris-Glycine) system are examples of discontinuous buffer systems and work in a similar fashion. However, the NuPAGE® system operates at a lower pH as a result of different ions in the system. In the case of the Tris-Glycine system (Figure 1), three ions are primarily involved: a) Chloride (-), supplied by the gel buffer, serves as the leading ion because it has the highest attraction to the anode relative to other anions in the system. b) Glycine (-), the primary anion provided by the running buffer, serves as the trailing ion, because it is only partially negatively charged and remains behind the more highly charged chloride ions in a charged environment. c) Tris Base (+) is a common ion present in both the gel and the running buffers. During electrophoresis, the gel and buffer ions in the Tris-Glycine system form an operating pH of 9.5 in the separating region of the gel.
In the case of the NuPAGE® Bis-Tris system (Figure 2), three ions are primarily involved: a) Chloride (-) supplied by the gel buffer, serves as the fast-moving leading ion. b) MES or MOPS (-) (depending on the running buffer choice) serves as the trailing ion. MES: 2-(N-morpholino) ethane sulfonic acid MOPS: 3-(N-morpholino) propane sulfonic acid c) Bis-tris (+) acts as the common ion present in the gel while Tris(+) is provided by the running buffer. The combination of a lower pH gel buffer (pH 6.4) and running buffer (pH 7.3 - 7.7) leads to a significantly lower operating pH (pH 7.0) during electrophoresis. In the case of the NuPAGE® Tris-Acetate system, three ions are primarily involved:
a) Acetate (-), the leading ion from the gel buffer
b) Tricine (-), the trailing ion from the running buffer
c) Tris (+), the common ion (in both gel and running buffer)
This system also operates at a significantly lower pH than the Tris-Glycine system.
Accurate Calibration of Molecular Weight Standards for Different Buffer Systems
Generally, protein mobility in SDS gels is a function of the length of the protein in its fully denatured state (1). By constructing a standard curve with protein standards of known molecular weights, the molecular weight of a sample protein can be calculated based upon its relative mobility. However, the same molecular weight standard may have slightly different mobility and therefore, different apparent molecular weight when run in different SDS-PAGE buffer systems.
The Effects of Secondary Structure
When using SDS-PAGE for molecular weight calibration, slight deviations from the “true” molecular weight of a protein (definitively calculated from the known amino acid sequence) can occur mostly because of the retention of varying degrees of secondary structure in the protein, even in the presence of SDS. This phenomenon is more prevalent in proteins with highly organized secondary structures (such as collagens, histones, or highly hydrophobic membrane proteins) and in peptides, where the effect of local secondary structure becomes magnified relative to the total size of the peptide (2).
The pH Factor
It has also been observed that slight differences in protein mobilities occur when the same proteins are run in different SDS-PAGE buffer systems (2). Each SDS PAGE buffer system has a different pH, which affects the charge of a protein and its binding capacity for SDS. The degree of change in protein mobility is usually small in natural proteins but is more pronounced with “atypical” or chemically modified proteins, such as pre-stained standards.
Demonstrated Protein Shifts
An example of a situation where there is a shift in protein migration occurs when running the MultiMark® Multi-Colored Standard on a Tris-Glycine Gel in comparison to the same standard run on a NuPAGE® Novex Bis-Tris Gel run with either MES SDS or MOPS SDS Running Buffer. The two bands of myoglobin (red and blue) in the MultiMark® Standard are reversed in the NuPAGE® Bis-Tris buffer system, compared to the Tris-Glycine system. This phenomenon can be addressed by applying the recalculated values for the MultiMark® standard run on the respective NuPAGE® System.
Calculated and Assigned Molecular Weights
The apparent molecular weight values currently provided with the Novex® molecular weight standards were derived from the construction of a calibration curve in the Tris-Glycine SDS-PAGE System. We have calculated and assigned apparent molecular weights for the Novex® pre-stained markers in several electrophoresis gel/buffer systems including the Novex® Tricine gel system and the NuPAGE® Novex systems. Remember to use the one which matches your gel for the most accurate calibration of your sample proteins. You may generate additional calibration curves in your own lab with the Novex® standards run in a buffer system of choice.
Lot-to-Lot Molecular Weight Consistency
The important point to remember is that all the Invitrogen standards are manufactured and quality controlled to migrate consistently from one lot of standards to the next and from one gel to the next, given that the gels are using the same buffer system. This makes each of the Novex® standards equally useful in all the available buffer systems.
References:
  1.    Rodbard, D. (1976) Meth. Protein Separation 2: 145-218.
  2.    Patton, W.F. et al. (1991) Anal. Biochemistry, 200: 25-30.
  3.    Wyckoff, M. et al. (1977) Anal Biochem. 78: 459-482.
TOP
Troubleshooting
Electrophoresis Problem Possible Cause Suggested Solution
Run taking longer time with recommended voltage.  Running buffer too dilute.  Make fresh running buffer and use a 1X dilution. 
Current too high and excessive heat generated with recommended voltage.  Running buffer too concentrated.  Make fresh running buffer and use a 1X dilution. 
Current too low or no current with recommended voltage.  Incomplete circuit.  Remove the tape from the bottom of the gel cassette prior to electrophoresis; Make sure the buffer covers sample wells; check the wire connections on the buffer core. 
Streaking of proteins.  Sample overload. High salt concentration in sample.  Load less amount of protein. Decrease the sample salt concentration by dialysis or gel filtration. 
Streaking of proteins.  Sample precipitates.  Increase the concentration of SDS in the sample. 
Streaking of proteins.  Contaminants such as lipids or DNA complexes in sample.  Centrifuge or clarify the sample to remove particulate contaminants. 
Streaking of proteins.  Poorly poured gel.  Make sure the gel is poured evenly and all at once. 
Fuzzy bands.  Protein sample only partially denatured.  Fully denature the protein. 
Fuzzy bands.  Protein sample only partially reduced.  Make sure sufficient amount of DTT or ß-mercaptoethanol is added. 
Fuzzy bands.  Gel run for too long.  Watch the front dye as an indicator for proper running time. 
Dumbbell shaped bands or “smiling” bands.  Loading a large volume of sample causes incomplete stacking.  Load appropriate volume of sample. If the sample is too dilute, concentrate it using ultrafiltration. 
Dumbbell shaped bands or “smiling” bands.  Uneven electric field during run.  Try to make sure the loading is symmetrical if the protein concentration is known. 
Dumbbell shaped bands or “smiling” bands.  Uneven surface of the resolving gel.  Try to make the resolving gel surface even while pouring the gel. 
Dumbbell shaped bands or “smiling” bands.  Expired gels.  Use the gels before the specified expiration date. 
TOP
Running Condition
Gel Type Voltage for Electophoresis Expected Current (gel) Gel Runtime
DNA Retardation  100 Volts constant  12-15 mA/gel (start), 6-15 mA/gel (end)  90 minutes 
IEF 1  100 Volts for 1 hour, 200 Volts for 1hour, 500 Volts for 30 minutes  5 mA (start), 6 mA (end)  2.5 hours 
IEF 2  100 Volts for 1 hour, 200 Volts for 1hour, 500 Volts for 30 minutes  5 mA (start), 6 mA (end)  2.5 hours 
NuPAGE® Bis-Tris SDS-PAGE (Denaturing, non-reducing)  200 Volts constant  100-125 mA (start), 60-70 mA (end)  35-50 minutes 
NuPAGE® Bis-Tris SDS-PAGE (Denaturing, reducing)  200 Volts constant  110-125 mA (start), 70-80 mA (end)  35-50 minutes 
NuPAGE® Tris-Acetate SDS-PAGE (Denaturing, non-reducing)  150 Volts constant  40-55 mA (start), 25-40 mA (end)  60 minutes 
NuPAGE® Tris-Acetate SDS-PAGE (Denaturing, reducing)  150 Volts constant  40-55 mA (start), 25-40 mA (end)  60 minutes 
NuPAGE® Tris-Acetate SDS-PAGE (Native)  20-150 Volts constant  40-55 mA (start), 25-40 mA (end)  1-12 hours 
TBE  200 Volts constant  10-18 mA (start), 4-6 mA (end)  30-90 minutes 
TBE-Urea  180 Volts constant  10-20 mA (start), 6-14 mA (end)  50-75 minutes 
Tricine SDS-PAGE (Denaturing, non-reducing)  125 Volts constant  80 mA (start), 40 mA (end)  90 minutes 
Tris-Glycine SDS-PAGE (Denaturing, non-reducing)  125 Volts constant  30-40 mA (start), 8-12 mA (end)  90 minutes 
Tris-Glycine SDS-PAGE (Denaturing, reducing)  125 Volts constant  30-40 mA (start), 8-12 mA (end)  90 minutes 
Tris-Glycine SDS-PAGE (Native)  20-125 Volts constant  6-12 mA (start), 3-6 mA (end)  1-12 hours 
ZOOM® IPGRunner™  Protocol A, 175 V for 15 minutes, 175-2000 V ramp for 45 minutes, 2000 V for 20-30 minutes; Protocol B, 200 V for 20 minutes, 450 V for 15 minutes, 750 V for 15 minutes, 2000 V for 30 minutes  50 uA per strip  90 minutes 
Zymogram  125 Volts constant  30-40 mA (start), 8-12 mA (end)  90 minutes 
TOP
Transfer Condition
Gel Type Transfer Buffer Transfer Membrane Voltage for Transfer Expected Current (transfer) Transfer Time
DNA Retardation  TBE Running buffer used at 0.5X concentration.  Nylon  30 V constant  Start: 390 mA, End: 350 mA  1-2 hours 
IEF 1  0.7% acetic acid, pH 3.0 (Assemble the gel/membrane sandwich in reverse order so that the membrane is on the cathode side (-) of the gel.  Nitrocellulose or PVDF  25 V constant  Start: 65-85 mA  1 hour 
IEF 2  Novex® Tris-Glycine Transfer Buffer with 20% methanol (equilibrate gel before transfer).  Nitrocellulose or PVDF  10 V constant  Start: 65-85 mA  1 hour 
NuPAGE® Bis-Tris SDS-PAGE (Denaturing, non-reducing)  NuPAGE® Transfer Buffer with 10% methanol for transfer of one gel.  Nitrocellulose or PVDF  30 V constant  Start: 170 mA, End: 110 mA  1 hour 
NuPAGE® Bis-Tris SDS-PAGE (Denaturing, reducing)  NuPAGE® Transfer Buffer with 10% methanol for transfer of one gel., NuPAGE® Antioxidant for reduced samples.  Nitrocellulose or PVDF  30 V constant  Start: 170 mA, End: 110 mA  1 hour 
NuPAGE® Tris-Acetate SDS-PAGE (Denaturing, non-reducing)  NuPAGE® Transfer Buffer with 10% methanol for transfer of one gel.  Nitrocellulose or PVDF  30 V constant  Start: 220 mA, End: 180 mA  1 hour 
NuPAGE® Tris-Acetate SDS-PAGE (Denaturing, reducing)  NuPAGE® Transfer Buffer with 10% methanol for transfer of one gel., NuPAGE® Antioxidant for reduced samples.  Nitrocellulose or PVDF  30 V constant  Start: 220 mA, End: 180 mA  1 hour 
NuPAGE® Tris-Acetate SDS-PAGE (Native)  NuPAGE® Transfer Buffer with 10% methanol for transfer of one gel., NuPAGE® Antioxidant for reduced samples.  Nitrocellulose or PVDF  30 V constant  Start: 220 mA, End: 180 mA  1 hour 
TBE  TBE Running buffer used at 0.5X concentration.  Nylon  30 V constant  Start: 390 mA, End: 350 mA  1-2 hours 
TBE-Urea  TBE Running buffer used at 0.5X concentration.  Nylon  30 V constant  Start: 390 mA, End: 350 mA  1-2 hours 
Tricine SDS-PAGE (Denaturing, non-reducing)  Novex® Tris-Glycine Transfer Buffer with 20% methanol., 1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol. Do not use acid or base to adjust the pH.  Nitrocellulose or PVDF  25 V constant  Start: 100 mA  1-2 hours 
Tris-Glycine SDS-PAGE (Denaturing, non-reducing)  Novex® Tris-Glycine Transfer Buffer with 20% methanol., 1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol. Do not use acid or base to adjust the pH. For basic proteins, use blotting membrane on BOTH sides of the gel. Proteins that are more basic than the pH of the Tris-Glycine transfer buffer will be captured on th cathode side membrane. Those less basic will be captured on the anode side., Increase the pH of the Tris-Glycine transfer buffer to pH 9.2. All proteins below pI 9.2 will transfer toward the anode electrode., Incubate the gel for 15 minutes in Tris-Glycine transfer buffer with 0.1% SDS. The small amount of SDS may give the proteins enough charge to move unidirectionally towards the anode and in most cases should not denature the protein.  Nitrocellulose or PVDF  25 V constant  Start: 100 mA  1-2 hours 
Tris-Glycine SDS-PAGE (Denaturing, reducing)  Novex® Tris-Glycine Transfer Buffer with 20% methanol., 1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol. Do not use acid or base to adjust the pH. For basic proteins, use blotting membrane on BOTH sides of the gel. Proteins that are more basic than the pH of the Tris-Glycine transfer buffer will be captured on th cathode side membrane. Those less basic will be captured on the anode side., Increase the pH of the Tris-Glycine transfer buffer to pH 9.2. All proteins below pI 9.2 will transfer toward the anode electrode., Incubate the gel for 15 minutes in Tris-Glycine transfer buffer with 0.1% SDS. The small amount of SDS may give the proteins enough charge to move unidirectionally towards the anode and in most cases should not denature the protein.  Nitrocellulose or PVDF  25 V constant  Start: 100 mA  1-2 hours 
Tris-Glycine SDS-PAGE (Native)  Novex® Tris-Glycine Transfer Buffer with 20% methanol., 1X Transfer Buffer should be pH 8.3 before addition of SDS or methanol. Do not use acid or base to adjust the pH. For basic proteins, use blotting membrane on BOTH sides of the gel. Proteins that are more basic than the pH of the Tris-Glycine transfer buffer will be captured on th cathode side membrane. Those less basic will be captured on the anode side., Increase the pH of the Tris-Glycine transfer buffer to pH 9.2. All proteins below pI 9.2 will transfer toward the anode electrode., Incubate the gel for 15 minutes in Tris-Glycine transfer buffer with 0.1% SDS. The small amount of SDS may give the proteins enough charge to move unidirectionally towards the anode and in most cases should not denature the protein.  Nitrocellulose or PVDF  25 V constant  Start: 100 mA  1-2 hours 
ZOOM® IPGRunner™  N/A  N/A  N/A  N/A  N/A 
Zymogram  N/A  N/A  N/A  N/A  N/A 
TOP
Gel Selection Guide
Gel GelType MinResolution MaxResolution Denaturing SampleBuffer RunningBuffer TransferBuffer
10% Tricine Gel    6kDa  200kDa  Tricine SDS Sample Buffer  Tricine SDS Running Buffer  Tris-Glycine Transfer Buffer 
10% Tris-Glycine    20kDa  250kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
10% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
10-20% Tricine Gel    2kDa  200kDa  Tricine SDS Sample Buffer  Tricine SDS Running Buffer  Tris-Glycine Transfer Buffer 
10-20% Tris-Glycine    6kDa  200kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
10-20% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
12% Tris-Glycine    20kDa  250kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
12% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
14% Tris-Glycine    3kDa  60kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
14% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
16% Tricine Gel    <2kDa  200kDa  Tricine SDS Sample Buffer  Tricine SDS Running Buffer  Tris-Glycine Transfer Buffer 
16% Tris-Glycine    3kDa  60kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
16% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
18% Tris-Glycine    3kDa  60kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
18% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
4% Tris-Glycine    116kDa  500kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
4% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
4-12% Tris-Glycine    6kDa  200kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
4-12% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
4-20% Tris-Glycine    6kDa  200kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
4-20% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
6% Tris-Glycine    116kDa  500kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
6% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
8% Tris-Glycine    20kDa  250kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
8% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
8-16% Tris-Glycine    6kDa  200kDa  Tris-Glycine SDS Sample Buffer  Tris-Glycine SDS Running Buffer  Tris-Glycine Transfer Buffer 
8-16% Tris-Glycine (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris-Glycine Transfer Buffer 
NuPAGE 10% Bis-Tris (With MES)    2.5 kDa  200 kDa  NuPAGE® LDS Sample Buffer  NuPAGE® MES SDS Running Buffer  NuPAGE® Transfer Buffer 
NuPAGE 10% Bis-Tris (With MOPS)    14kDa  220kDa  NuPAGE® LDS Sample Buffer  NuPAGE® MOPS SDS Running Buffer  NuPAGE® Transfer Buffer 
NuPAGE 12% Bis-Tris (With MES)    2kDa  200kDa  NuPAGE® LDS Sample Buffer  NuPAGE® MES SDS Running Buffer  NuPAGE® Transfer Buffer 
NuPAGE 12% Bis-Tris (With MOPS)    6kDa  200kDa  NuPAGE® LDS Sample Buffer  NuPAGE® MOPS SDS Running Buffer  NuPAGE® Transfer Buffer 
NuPAGE 3-8% Tris-Acetate    31kDa  400kDa  NuPAGE® LDS Sample Buffer  NuPAGE® Tris-Acetate SDS Running Buffer  NuPAGE® Transfer Buffer 
NuPAGE 3-8% Tris-Acetate (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris Glycine Transfer Buffer 
NuPAGE 4-12% Bis-Tris (With MES)    2.5kDa  200kDa  NuPAGE® LDS Sample Buffer  NuPAGE® MES SDS Running Buffer  NuPAGE® Transfer Buffer 
NuPAGE 4-12% Bis-Tris (With MOPS)    12kDa  220kDa  NuPAGE® LDS Sample Buffer  NuPAGE® MOPS SDS Running Buffer  NuPAGE® Transfer Buffer 
NuPAGE 7% Tris-Acetate (Native)    n/a  n/a  Tris-Glycine Native Sample Buffer  Tris-Glycine Native Running Buffer  Tris Glycine Transfer Buffer 
NuPAGE 7%Tris-Acetate    31kDa  400kDa  NuPAGE® LDS Sample Buffer  NuPAGE® Tris-Acetate SDS Running Buffer  NuPAGE® Transfer Buffer 
TOP
Maximum Loading
WellNum-Thickness Max Loading Volume Max Protein Amount Max DNA Amount
10-1.0 mm  25 ul  0.5 ug/band with Coomassie  100 ng/band with EtBr 
10-1.5 mm  37 ul  0.5 ug/band with Coomassie  100 ng/band with EtBr 
1-1.0 mm  700 ul  12 ug/band with Coomassie  2400 ng/band with EtBr 
12-1.0 mm  20 ul  0.5 ug/band with Coomassie  100 ng/band with EtBr 
15-1.0 mm  15 ul  0.5 ug/band with Coomassie  100 ng/band with EtBr 
15-1.5 mm  25 ul  0.5 ug/band with Coomassie  100 ng/band with EtBr 
17-1.0 mm  15 ul  0.5 ug/band with Coomassie  n/a 
2D-1.0 mm  400 ul  12 ug/band with Coomassie  2000 ng/band with EtBr 
2D-1.5 mm  600 ul  12 ug/band with Coomassie  2000 ng/band with EtBr 
5-1.0 mm  60 ul  2 ug/band with Coomassie  400 ng/band with EtBr 
9-1.0 mm  28 ul  0.5 ug/band with Coomassie  100 ng/band with EtBr 
IPG-1 .0 mm  7 cm IPG strip  n/a  n/a 
TOP
Buffer Composition
Buffer 1XComposition Storage
Hi-Density TBE Sample Buffer  Tris Base_18 mM, Boric Acid_18 mM, EDTA (free acid)_0.4 mM, Ficoll™ Type 400_3%, Bromophenol Blue_0.02%, Xylene Cyanol_0.02%, DI Water  +4°C 
IEF Anode Buffer (for both pH ranges) (Lower Buffer Chamber)  Phosphoric Acid 85%_7 mM, DI Water, pH 2.4  Room Temperature 
IEF Cathode Buffer pH 3-10 (Upper Buffer Chamber)  Arginine (free base)_20 mM, Lysine (free base)_20 mM, DI Water, pH 10.1  +4°C 
IEF Cathode Buffer pH 3-7 (Upper Buffer Chamber)  Lysine (free base)_40 mM, DI Water  +4°C 
IEF Sample Buffer pH 3-10  Arginine (free base)_20 mM, Lysine (free base)_20 mM, Glycerol_15%, DI Water  +4°C 
IEF Sample Buffer pH 3-7  Lysine (free base)_40 mM, Glycerol_15%, DI Water  +4°C 
NuPAGE® LDS Sample Buffer  Glycerol_10%, Tris Base_141 mM, Tris HCl_106 mM, LDS_2%, EDTA_0.51 mM, SERVA® Blue G250_0.22 mM, Phenol Red_0.175 mM, pH 8.5  +4°C 
NuPAGE® MES SDS Running Buffer  MES pH 7.2_50 mM, Tris Base_50 mM, SDS_0.1%, EDTA_1 mM, pH 7.3  +4°C 
NuPAGE® MOPS SDS Running Buffer  MOPS_50 mM, Tris Base_50 mM, SDS_0.1%, EDTA_1 mM, pH 7.7  +4°C 
NuPAGE® Transfer Buffer  Bicine_25 mM, Bis-tris (free base)_25 mM, EDTA_1.0 mM, Chlorobutanol_0.05 mM, pH 7.2  +4°C 
NuPAGE® Tris-Acetate SDS Running Buffer  Tris Base_50 mM, Tricine_50 mM, SDS_0.1%, pH 8.24  +4°C 
Prep TBE-Urea Sample Buffer  Tris Base_45mM, Boric Acid_45 mM, EDTA (free acid)_1mM, Ficoll™ Type 400_6%, Urea_3.5 M, DI Water  +4°C 
Sample Rehydration Buffer IPGRunner™  Urea_8 M, CHAPS_2%, Ampholytes_0.5%, Bromophenol Blue_0.002%, DI Water, DTT_20 mM  -20°C 
TBE Running Buffer  Tris Base_89mM, Boric Acid_89 mM, EDTA (free acid)_2mM, DI Water, pH 8.3  Room Temperature 
TBE Transfer Buffer (Use TBE Running Buffer [Equivalent to Cat. No. LC6675] at 1/2X concentration)  Tris Base_89mM, Boric Acid_89 mM, EDTA (free acid)_2mM, DI Water  Room Temperature 
TBE-Urea Sample Buffer  Tris Base_45mM, Boric Acid_45 mM, EDTA (free acid)_1mM, Ficoll™ Type 400_6%, Bromophenol Blue_0.005%, Xylene Cyanol _0.025%, Urea_3.5 M, DI Water  +4°C 
Tricine SDS Running Buffer  Tris Base pH 8.3_100 mM, Tricine_100 mM, SDS_0.1%, DI Water, pH 8.3  Room Temperature 
Tricine SDS Sample Buffer  Tris HCl pH 8.45_450 mM, Glycerol_12%, SDS_4%, Coomassie® Blue G_0.0075%, Phenol Red_0.0025%, DI Water, pH 8.45  +4°C 
Tris-Glycine Native Running Buffer  Tris Base_25 mM, Glycine_192 mM, DI Water, pH 8.3  Room Temperature 
Tris-Glycine Native Sample Buffer  Tris HCl_100 mM, Glycerol_10%, Bromophenol Blue_0.0025%, DI Water, pH 8.6  +4°C 
Tris-Glycine SDS Running Buffer  Tris Base_25 mM, Glycine_192 mM, SDS_0.1%, DI Water, pH 8.3  Room Temperature 
Tris-Glycine SDS Sample Buffer  Tris HCl_63 mM, Glycerol_10%, SDS_2%, Bromophenol Blue_0.0025%, DI Water, pH 6.8  +4°C 
Tris-Glycine Transfer Buffer  Tris Base_12 mM, Glycine_96 mM, DI Water, pH 8.3  Room Temperature 
Zymogram Developing Buffer  Tris Base_50 mM, 6N HCl_40 mM, NaCl_200 mM, CaCl2 2 H2O_5 mM, Brij 35_0.02% (w/v), DI Water  Room Temperature 
Zymogram Renaturing Buffer  Triton® X-100_2.7% (w/v) in H2O, DI Water  Room Temperature 
TOP