Search Thermo Fisher Scientific
Organism Type | Human | Tissue: embryonal carcinoma, malignant; testis; from metastatic site:lung. Temperature sensitive neural cell line expressing SV40 small T at lower temperature, inhibited at high temperature and therefore ceases proliferation and differentiates. |
Tissues | Testes | |
Phenotype | Adherent | |
Primary | yes | |
Application | Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||