Search Thermo Fisher Scientific
Organism Type | Mouse | Primary erythroleukemias were induced following injection of BALB/c mice at birth with clone 57 of F-MuLV helper virus. To establish cell lines from these tumors, erythroleukemic cells from tumor HB60-t were cultured in -MEM supplemented with 15% FBS, 1 U of Epo (Boehringer) per ml, and 100 mg of SCF per ml. After several days of culture in the presence of Epo and SCF, a small population of the HB60 splenic tumor cells survived and proliferated in the presence of 20% FBS. While these cells grow slowly under this condition, a rare and fast-growing population of these cells had emerged after approximately 2 months in culture. The HB60 cells were cloned by limited dilution, and the clone HB60-5 was used in further studies. To induce differentiation, HB60-5 cells were washed twice with phosphate-buffered saline (PBS) and incubated in the presence of 15% FBS and 0.1 U of Epo per ml. |
Tissues | Spleen | |
Phenotype | Adherent | |
Primary | no | |
Application | Stable Cell Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||