Technical Resources - Media Formulations
YM Medium, powder
INTENDED USE: Growth and expression of Agrobacterium. YM Medium improves transformation efficiency for electroporation of Agrobacterium by up to 40-fold when compared with LB broth. Also, no aggregation of Agrobacterium was observed when YM Medium was used in culturing the bacteria for plant infection. INSTRUCTIONS FOR USE: Suspend contents of 1 packet in 1 L of demineralized water. Autoclave for 15 min at 121°C. To prepare YM agar: Suspend contents of 1 packet in 1 L of demineralized water. Add 15 g of SELECT Agar and boil while stirring to dissolve all ingredients completely. Autoclave for 15 min at 121°C. Cool down to 50C before adding antibiotics (optional) and pouring into plates. RECOMMENDED STORAGE CONDITION: 15°C TO 30°C.
  • Catalog Number(s)
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Dipotassium hydrogen phosphate0.5Infinity
Magnesium Sulfate (MgSO4-7H2O)0.2Infinity
Sodium Chloride (NaCl)0.1Infinity
Other Components
Mannitol10.0Infinity
SELECT Yeast Extract0.4Infinity
参照
1. Lin J.J. (1994) Focus® 16, 18.
2. Lin J.J. (1994) Plant Science 10, 11.
3. Lin J.J. Assad-Garcia, N. and Kuo, J. (1994) Focus® 16, 72.
Related Products:
Terrific Broth
Cat No: 22711022
2-YT Broth
Cat No: 22712020
Gibco® BG-11 Media: Optimized for Cyanobacteria
Cat No: a1379901 , A1379902
TAP Growth Media, optimized for Chlamydomonas culture
Cat No: A1379801 , A1379802