Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 100.0 | 0.9009009 |
| Magnesium Chloride (anhydrous) | 95.0 | 47.0 | 0.49473685 |
| Potassium Chloride (KCl) | 75.0 | 200.0 | 2.6666667 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 200.0 | 1.4705882 |
| Sodium Chloride (NaCl) | 58.0 | 8000.0 | 137.93103 |
| Sodium Phosphate dibasic (Na2HPO4) anhydrous | 142.0 | 1150.0 | 8.098592 |
1. Dulbecco, R. and Vogt, M., (1954) Plaque formation and isolation of pure lines with Poliomyelitis viruses. J. Exp. Med., 98:167.
14040 - DPBS, calcium, magnesium
Cat No:
14040083
,
14040091
,
14040117
,
14040133
,
14040141
,
14040166
,
14040174
,
14040182
,
14040216
,
14040224
,
14040224
14080 - DPBS, calcium, magnesium
Cat No:
14080048
,
14080055
,
14080089